Biopsy Pads
Thermo Scientific™ Shandon™ Sponge, 25x31mm Non-distribution product as customer accommodation.
Protect small biopsies from overprocessing with the Thermo Scientific™ Shandon™ Sponge.
Thermo Scientific™ RA Lamb Biopsy Inserts
Prevent the loss of small or friable material while allowing interchange of fluids with Thermo Scientific™ RA Lamb Biopsy Inserts.
Fisherbrand™ Foam Biopsy Pads, Rectangular Available on GSA/VA Contract for Federal Government customers only.
Verified for consistency to achieve optimum solvent flow
Fisherbrand™ Foam Biopsy Pads, Circular Available on GSA/VA Contract for Federal Government customers only.
Verified for consistency to achieve optimum solvent flow
CellPath BioPad Biopsy Foam Pads
Allow fluid to thoroughly and safely transfer around the tissue sample, whilst also holding the tissue specimen safely in place. CellPath Biopsy Foam Pads can be cut to size to fit most major processing cassettes.
Bio Plas Foam Biopsy Pads
For histology specimen protection
Jaece™ Foam Biopsy Pads
Made from uniform polyester reticulated open cell foam, which allows fluids to transfer safely and thoroughly around the tissue while holding the specimen secure. Jaece™ Foam Biopsy Pads have a thin 2mm design which helps eliminate pressure from being applied to the unfixed specimen, reducing the risk of tissue sticking to the pads during embedding.
  spinner