Embedding Accessories
Thermo Scientific™ Shandon™ Nylon Biopsy Bags Available on GSA/VA Contract for Federal Government customers only.
Ensure unrestricted fluid movement around tissue with Thermo Scientific™ Shandon™ Nylon Biopsy Bags.
Thermo Scientific™ Shandon™ Para-Release™ Spray Available on GSA/VA Contract for Federal Government customers only.
Ease the release of paraffin blocks from embedding molds with Thermo Scientific™ Shandon™ Para-Release™ Spray.
Thermo Scientific™ Shandon™ Peel-A-Way Disposable Embedding Molds
Promote rapid paraffin hardening with Thermo Scientific™ Shandon™ Peel-A-Way Disposable Embedding Molds.
Thermo Scientific™ Shandon™ Acid-Resistant Biopsy Bags Available on GSA/VA Contract for Federal Government customers only.
Reduce risk of small specimen loss with Thermo Scientific™ Shandon™ Acid-Resistant Biopsy Bags.
Thermo Scientific™ RA Lamb Biopsy Inserts
Prevent the loss of small or friable material while allowing interchange of fluids with Thermo Scientific™ RA Lamb Biopsy Inserts.
Thermo Scientific™ Shandon™ Sponge, 25x31mm Non-distribution product as customer accommodation.
Protect small biopsies from overprocessing with the Thermo Scientific™ Shandon™ Sponge.
Thermo Scientific™ Richard-Allan Scientific™ Disposable Base Molds and Embedding Rings Available on GSA/VA Contract for Federal Government customers only.
Use Thermo Scientific™ Richard-Allan Scientific™ Disposable Base Molds and Embedding Rings with all standard-size cassettes and embedding rings.
Thermo Scientific™ Richard-Allan Scientific™ Base Mold Release
Coat base molds for easy separation of paraffin from mold with Thermo Scientific™ Richard-Allan Scientific™ Base Mold Release.
Thermo Scientific™ Shandon™ Disposable Base Molds
Promote rapid paraffin hardening with Thermo Scientific™ Shandon™ Disposable Base Molds.
Thermo Scientific™ Shandon™ Mega Base Molds
High-quality, large-sized molds
Thermo Scientific™ Shandon™ Stainless-Steel Base Molds
Optimize tissue embedding with the superior thermal exchange of our high-quality, durable Thermo Scientific™ Shandon™ Stainless-Steel Base Molds.
Fisherbrand™ Nylon Biopsy Bags
Reduce risk of small specimen loss
Fisher Scientific™ Tissue Path™ Disposable Base Molds
Inexpensive enough to be discarded, yet strong enough to be reused
Fisherbrand™ Foam Biopsy Pads, Circular Available on GSA/VA Contract for Federal Government customers only.
Verified for consistency to achieve optimum solvent flow
Fisherbrand™ Foam Biopsy Pads, Rectangular Available on GSA/VA Contract for Federal Government customers only.
Verified for consistency to achieve optimum solvent flow
CellPath Stainless-steel Reusable Base Molds
Ideal for prostrate and breast cores and can be used with both grid and slatted processing cassettes for both horizontal and vertical sectioning. Cellpath Stainless-steel Reusable Base Moulds are manufactured from high quality stainless steel that put an end to extensive trimming of wax.
CellPath TissueWrap Disposable Biopsy Papers
Specifically designed for processing biopsy specimens. CellPath TissueWrap Disposable Biopsy Papers is cut to a convenient ready to use size. This non-absorbent paper allows fluids to flow through freely whilst retaining small tissue fragments.
CellPath Acid Resistant Polyester Biopsy Bags
Holds biopsy sample during processing. CellPath Acid Resistant Polyester Biopsy Bags are great alternatives to BioWrap paper and foam biopsy pads. Particularly helpful when collecting endometrial samples from formalin as it can be used as a sieve.
CellPath BioPad Biopsy Foam Pads
Allow fluid to thoroughly and safely transfer around the tissue sample, whilst also holding the tissue specimen safely in place. CellPath Biopsy Foam Pads can be cut to size to fit most major processing cassettes.
Cell Safe Biopsy Capsule
Fits inside any cassette
Simport™ Scientific Stainless-Steel Base Molds
For applications in specimen embedding with Simport embedding rings and cassettes
Simport™ Scientific Disposable Base Molds
Inexpensive enough to be discarded after use, yet strong enough to be reused
Cen-Med Paper Biopsy Bags
Available in three sizes, these bags are made from porous paper to provide maximum tissue protection at a very affordable price
Cancer Diagnostics Nylon Mesh Biopsy Bags
Reduce cross-contamination
Cancer Diagnostics Cleanerol™
Base mold release agent
Bio Plas Foam Biopsy Pads
For histology specimen protection
Bio Plas HistoPlas™ Tissue Processing Capsules
The integral lid fits tightly—you never need to worry about losing your tissues
Jaece™ Foam Biopsy Pads
Made from uniform polyester reticulated open cell foam, which allows fluids to transfer safely and thoroughly around the tissue while holding the specimen secure. Jaece™ Foam Biopsy Pads have a thin 2mm design which helps eliminate pressure from being applied to the unfixed specimen, reducing the risk of tissue sticking to the pads during embedding.
  spinner