Embedding Cassettes
Fisherbrand™ Histosette II™ Tissue and Biopsy Cassettes
Open and close as often as necessary — cassettes will always relock securely
Fisher Scientific™ Tissue Path™ Microsette™ Biopsy Cassettes
Innovative, disposable, plastic biopsy cassettes will make biopsy pads obsolete
Fisher Scientific™ Tissue Path™ IV Lids and Bases Available on GSA/VA Contract for Federal Government customers only.
For SHUR/Mark™ and other automatic labeling systems
Fisherbrand™ Tissue Path™ IV Tissue Cassettes Available on GSA/VA Contract for Federal Government customers only.
Contain no jagged edges or uneven surfaces
Fisherbrand™ TRUFLOW™ Tissue and Biopsy Cassettes Available on GSA/VA Contract for Federal Government customers only.
Just bend, snap apart, and conveniently lock
Fisher Scientific™ Tissue Path™ MACROSETTE™ Processing/Embedding Cassettes
For holding large tissue specimens during processing and embedding
Fisherbrand™ Tissue Path™ II Cassettes Available on GSA/VA Contract for Federal Government customers only.
Standard lids fit securely onto cassettes
Fisher Scientific™ Tissue Path™ IV Biopsy Cassettes Available on GSA/VA Contract for Federal Government customers only.
Finely perforated to prevent loss of small biopsy specimens during processing
Fisherfinest™ Taped Biopsy Cassettes for Leica IP C and Sakura Cassette Printers
Pre-stacked, quick-load cassettes designed with efficient MicrOflow II™ reagent exchange technology
CellPath System II Hex™ Tissue Processing Cassettes (Hoppered)
Pre-loaded into hoppers for use with automated and carousel type printers. CellPath System II Hex Tissue Processing Cassettes (Hoppered) feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.
CellPath Supa Mega Tissue Processing Cassettes
Widely adopted in research and toxicology laboratories. CellPath Supa Mega Tissue Processing Cassettes are ideal for embedding and processing larger tissue samples of prostrate, brain, breast and eye.
General Data Healthcare™ SHUR/Track™ Process and Embedding Cassettes with Channel Track Hinge
Unique Track Hinge design provides easy one-handed installation of lid cover
CellPath System III Hex™ Biopsy Cassettes
To hold tissue specimens during embedding, processing, and storage. CellPath System III Hex Biopsy Cassettes with lid feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.
CellPath System III Hex™ Cassettes for Tissue Processing
To hold tissue specimens during embedding, processing, and storage. CellPath System III Hex Tissue Processing Cassettes with lid feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.
CellPath System II Hex™ Tissue Processing Cassettes
To hold tissue specimens during embedding, processing, and storage. CellPath System II Hex Tissue Processing Cassettes feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.
General Data Healthcare™ SHUR/Mark™ Processing and Embedding Cassette Taped Sleeves
Prepackaged SHUR/Track™ cassette sleeves for direct insert into the SHUR/Mark Cassette Labeler taped sleeve dispensing hopper (SM-TSDH)
CellPath Stainless-Steel Supa Mega Base Moulds
Reusable base moulds. CellPath Stainless-Steel Supa Mega Base Moulds have extra deep side walls allowing wax to be decanted into the cassette during embedding without the risk of the usual ‘leakage’ of wax.
CellPath CellSafe 5 Core Biopsy Cassettes with Five Wells
Reduces the pressure on laboratory staff by enabling them to perform five core biopsies in numbered wells sectioned together for quick, cost effective preparation. CellPath CellSafe Biopsy cassettes are used for breast and prostrate samples.
General Data Healthcare™ SHUR/Track™ Processing and Embedding Cassettes with Channel Track Hinge
Specifically designed and quality controlled for use with both automated labeling instruments and hand marking devices
CellPath ProMarc Supa Mega Mother Ship Tissue Processing Cassettes
Provides an innovative solution for the processing and embedding of large tissue specimens. CellPath ProMarc Supa Mega Mother Ship Tissue Processing Cassettes feature an internal chamber a third of the depth of the conventional Supa Mega Cassette.
CellPath CellSafe+ Blue Biopsy Capsules
Offers high security and easy to use alternative to traditional biopsy handling techniques such as tissue wrap and foam pads which are proving to be unsatisfactory and time consuming. CellPath CellSafe+ Blue Biopsy Capsules features new blue mesh that provides better contrast between the biopsy and the CellSafe.
CellPath HEX™ Cassette Lid
Lid for HEX tissue processing casette
General Data Healthcare™ SHUR/Mark™ Biopsy Cassette Taped Sleeves
Prepackaged SHUR/Track™ cassette sleeves for direct insert into the SHUR/Mark Cassette Labeler taped sleeve dispensing hopper (SM-TSDH)
CellPath CellSafe+ White Biopsy Capsules
Offers high security and easy to use alternative to traditional biopsy handling techniques such as tissue wrap and foam pads. CellPath CellSafe+ White Biopsy Capsules features two hinged interlocking frames with an integral nylon mesh that ensures high level of specimen security, zero tissue damage, and low reagent carry-over.
CellPath Tissue Processing System II Cassette Lid, Stainless- Steel
For use in place of plastic lid on System II tissue processing cassettes. CellPath Tissue Processing System II Cassette Lid is made of stainless-steel.
  spinner