Embedding Cassette Storage
Thermo Scientific™ Plastic Block Filing Cabinets
Simple filing of up to 1500 blocks. Thermo Scientific™ Plastic Block Filing Cabinets feature smooth sliding action and rigid construction.
Thermo Scientific™ Modular Cassette Storage Drawers
Permanently store up to 165 embedding rings or 250 cassettes per drawer with modular cassette storage drawers.
Thermo Scientific™ Modular Filing System Available on GSA/VA Contract for Federal Government customers only.
Hold a combination of microscope slides, tissue cassettes and 35mm slides in one cabinet with the Thermo Scientific™ Modular Filing System.
Thermo Scientific™ Stackable Slide Storage Cabinet Available on GSA/VA Contract for Federal Government customers only.
Hold up to 5000 standard 3 × 1″ (75 × 26mm) microscope slides in the Thermo Scientific™ Stackable Slide Storage Cabinet. Finished in beige baked enamel and complete with drawers.
Thermo Scientific™ Slide File Storage System
Save space by storing materials in the stackable Thermo Scientific™ Slide File Storage System.
Thermo Scientific™ Slide and Block Cabinets
Form an integrated storage system with stackable Thermo Scientific™ Slide and Block Cabinets, constructed of steel and finished in solvent-proof paint.
Accessories for the Syntri™ Arcos™ Block Management System
Improve capacity and organization with these Thermo Scientific™ Acccessories for the Syntri™ Arcos™ Block Management System.
Syntri™ Arcos™ Block Management System
Take control of your archives with the Thermo Scientific™ Syntri™ Arcos™ Block Management System.
Fisherbrand™ Storage File Systems Available on GSA/VA Contract for Federal Government customers only.
The best balance between economy and superior long-term protection
Fisherbrand™ Cassette Storage Cabinet
Economical option for secure storage of tissue cassettes
Fisher Scientific™ Tissue Path™ Universal Modular Filing System Available on GSA/VA Contract for Federal Government customers only.
Stores 22% more slides or 32% more specimen blocks than standard units
Fisherbrand™ HistoPrep™ Modular File Drawer Available on GSA/VA Contract for Federal Government customers only.
To transport, just remove drawer and go
Fisher Scientific™ Tissue Path™ Tissue Cassette/Embedding Ring Storage Cabinets Available on GSA/VA Contract for Federal Government customers only.
Six-drawer model with six insert trays holds 900 embedding rings or 1800 tissue cassettes
Fisher Scientific™ Tissue Path™ Tissue Cassette Storage Box
Multiple stacking for maximum use of storage space
Fisher Scientific™ Tissue Path™ Tissue Cassette/Embedding Ring File Drawers
Multiple stacking for maximum use of storage space
  spinner