Glass Staining Dishes
Fisherbrand™ Glass Staining Dish for 10 Slides, with Cap Available on GSA/VA Contract for Federal Government customers only.
Hold up to 10 standard 3 × 1 in. (75 × 25mm) slides back-to-back
Fisherbrand™ Glass Staining Dishes for 16 Slides Available on GSA/VA Contract for Federal Government customers only.
Staining jar holds 3 × 1 in. (75 × 25mm) slides vertically
DWK Life Sciences Wheaton™ Accessories and Replacement Parts for 20-Slide Glass Staining Dish Available on GSA/VA Contract for Federal Government customers only.
Featuring an open bottom to facilitate rapid immersion and drainage, reducing carryover. Wheaton™ Accessories and Replacement Parts for 20-Slide Glass Staining Dish are manufactured from soda-lime glass.
DWK Life Sciences Wheaton™ Glass Staining Dish and Slide Rack Available on GSA/VA Contract for Federal Government customers only.
Mix-and-match components offer greater flexibility in meeting laboratory slide staining requirements. Made of sode-lime glass.
Thermo Scientific™ RA Lamb Glass Coplin Jar, holds 5 slides (10 back to back)
Hold up to 10 slides back-to-back with Thermo Scientific™ RA Lamb molded glass coplin jars. Jars have a drop-on lid and integral large diameter base.
Fisherbrand™ Glass Staining Dishes for 20 Slides
Staining dish with 10 grooves for carrying up to 20 standard microscope slides back-to-back
DWK Life Sciences Wheaton™ Staining Dishes, 8-16 Slide Unit
Can be used for staining or as a TLC developing chamber. Wheaton™ Staining Dishes, 8-16 Slide Unit are manufactured from soda lime glass.
DWK Life Sciences Wheaton™ Screw Caps for 900570 Slide Staining Jar Available on GSA/VA Contract for Federal Government customers only.
Screw caps for Wheaton Glass Staining Jar (900570). Coplin staining jar holds up to five single slides or ten slides 3 x 1 in. (75 x 25mm) back-to-back vertically
DWK Life Sciences Wheaton™ Staining Jars, 8-10 Slide Unit Available on GSA/VA Contract for Federal Government customers only.
Featuring a wide top designed for convenience in transferring slides. Wheaton™ Staining Jars, 8-10 Slide Unit are especially suitable for staining slides that are inscribed on one end.
DWK Life Sciences Wheaton™ Columbia Jars for Cover Slips Available on GSA/VA Contract for Federal Government customers only.
For staining applications, cleaning cover slips, as well as coating them with materials such as poly-lysine or silane. Wheaton™ Columbia Jars for Cover Slips are manufactured from soda lime glass.
Fisherbrand™ Glass Staining Dishes for 10 Slides, with Ground Glass Cover Available on GSA/VA Contract for Federal Government customers only.
Hold up to 10 standard 3 × 1 in. (75 × 25mm) slides back-to-back
DWK Life Sciences Wheaton™ Columbia Jars for Cover Slips
For staining applications, cleaning cover slips, as well as coating them with materials such as poly-lysine or silane. Wheaton™ Columbia Jars for Cover Slips are manufactured from soda lime glass.
DWK Life Sciences Wheaton™ Glass Staining Dish for 10 or 20 Slides Available on GSA/VA Contract for Federal Government customers only.
Grooved staining dish holds up to 10 single slides or 20 slides back-to-back
Horizontal Staining Dish Products for Science Education
Horizontal staining dish with removable tray and cover.
United Scientific Supplies Coplin Staining Jar Products for Science Education
Features domed and shallow thread screw cap.
Eisco™ Glass Staining Dish with Cover Products for Science Education
Eisco glass staining dish with 9 grooves, can hold up to 10 slides (20 slides back to back). Molded glass lid included.
Eisco™ Glass Coplin Jar Products for Science Education
Eisco™ Glass Coplin Jar with a molded glass cover that accommodates 5 slides is for staining and washing slides.
Coplin Staining Dish Products for Science Education
Large capacity.
  spinner