Indigo Violet Purple Stains Dyes and Indicators
Crystal Violet (Certified Biological Stain), Fisher Chemical
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 MDL Number: MFCD00011750 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M Synonym: Gentian Violet, Basic Violet 3 PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Bromocresol Purple, pure, indicator grade, ACROS Organics™
CAS: 115-40-2 Molecular Formula: C21H16Br2O5S Molecular Weight (g/mol): 540.222 InChI Key: ABIUHPWEYMSGSR-UHFFFAOYSA-N Synonym: Bromcresol Purple, BCP, 5', 5''-Dibromo-o-cresolsulfonephthalein PubChem CID: 8273 ChEBI: CHEBI:86154 IUPAC Name: 2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol SMILES: CC1=C(C(=CC(=C1)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)C)Br)O
Cresyl Violet acetate, pure, high purity biological stain, ACROS Organics™
CAS: 10510-54-0 Molecular Formula: C18H15N3O3 Molecular Weight (g/mol): 321.336 MDL Number: MFCD00013151 InChI Key: UEWSHLFAYRXNHZ-UHFFFAOYSA-N PubChem CID: 44134641 IUPAC Name: acetic acid;5-iminobenzo[a]phenoxazin-9-amine SMILES: CC(=O)O.C1=CC=C2C(=C1)C(=N)C=C3C2=NC4=C(O3)C=C(C=C4)N
Cresyl Violet acetate, pure, certified, ACROS Organics™
CAS: 10510-54-0 Molecular Formula: C18H15N3O3 Molecular Weight (g/mol): 321.336 InChI Key: UEWSHLFAYRXNHZ-UHFFFAOYSA-N PubChem CID: 44134641 IUPAC Name: acetic acid;5-iminobenzo[a]phenoxazin-9-amine SMILES: CC(=O)O.C1=CC=C2C(=C1)C(=N)C=C3C2=NC4=C(O3)C=C(C=C4)N
Crystal Violet, pure, high purity biological stain, ACROS Organics™
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 MDL Number: MFCD00011750 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M Synonym: Basic Violet 3, C.I. 42555, Gentian Violet PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Bromocresol Purple (Certified), Fisher Chemical
CAS: 115-40-2 Molecular Formula: C21H16Br2O5S Molecular Weight (g/mol): 540.222 MDL Number: MFCD00011681 InChI Key: ABIUHPWEYMSGSR-UHFFFAOYSA-N Synonym: 5, 5'dibromo-o-cresolsulfonphthalein PubChem CID: 8273 ChEBI: CHEBI:86154 IUPAC Name: 2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol SMILES: CC1=C(C(=CC(=C1)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)C)Br)O
Crystal Violet, certified, ACROS Organics™
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 MDL Number: MFCD00011750 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M Synonym: Basic Violet 3, C.I. 42555, Gentian Violet PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Alfa Aesar™ m-Cresol, 99%
CAS: 108-39-4 Molecular Formula: C7H8O Molecular Weight (g/mol): 108.14 MDL Number: MFCD00002302 InChI Key: RLSSMJSEOOYNOY-UHFFFAOYSA-N Synonym: 3-Methylphenol PubChem CID: 342 ChEBI: CHEBI:17231 IUPAC Name: 3-methylphenol SMILES: CC1=CC(=CC=C1)O
Cresyl violet (acetate), For Microscopy, Certistain™, EMD Millipore™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 10510-54-0 Molecular Formula: C18H15N3O3 Molecular Weight (g/mol): 321.336 InChI Key: UEWSHLFAYRXNHZ-UHFFFAOYSA-N PubChem CID: 44134641 IUPAC Name: acetic acid;5-iminobenzo[a]phenoxazin-9-amine SMILES: CC(=O)O.C1=CC=C2C(=C1)C(=N)C=C3C2=NC4=C(O3)C=C(C=C4)N
Crystal Violet Solution R, Ricca Chemical
CAS: 64-19-7 Molecular Formula: C2H4O2 Molecular Weight (g/mol): 60.052 InChI Key: QTBSBXVTEAMEQO-UHFFFAOYSA-N PubChem CID: 176 ChEBI: CHEBI:15366 IUPAC Name: acetic acid SMILES: CC(=O)O
Indigo Carmine, ACROS Organics™
CAS: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Molecular Weight (g/mol): 466.346 MDL Number: MFCD00005723 InChI Key: KHLVKKOJDHCJMG-QDBORUFSSA-L Synonym: Acid Blue 74, C.I. 73015, 5, 5'-Indigodisulfonic acid, disodium salt PubChem CID: 5284351 IUPAC Name: disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate SMILES: C1=CC2=C(C=C1S(=O)(=O)[O-])C(=O)C(=C3C(=O)C4=C(N3)C=CC(=C4)S(=O)(=O)[O-])N2.[Na+].[Na+]
Crystal violet (C.I. 42555), For Microscopy, Certistain™, EMD Millipore™
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M Synonym: C.I. 42555 PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Crystal Violet Gram Solution EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Crystal Violet
Alfa Aesar™ m-Cresol Purple sodium salt
CAS: 62625-31-4 Molecular Formula: C21H17NaO5S MDL Number: MFCD00010177 Synonym: m-Cresol Purple, water soluble; m-Cresolsulfonephthalein sodium salt
Indigo, synthetic, ACROS Organics™
CAS: 482-89-3 Molecular Formula: C16H10N2O2 Molecular Weight (g/mol): 262.268 MDL Number: MFCD00005722 InChI Key: COHYTHOBJLSHDF-BUHFOSPRSA-N Synonym: C.I. 73000, Vat Blue 1 PubChem CID: 5318432 IUPAC Name: (2E)-2-(3-oxo-1H-indol-2-ylidene)-1H-indol-3-one SMILES: C1=CC=C2C(=C1)C(=O)C(=C3C(=O)C4=CC=CC=C4N3)N2
Indigo Carmine, High Purity Biological Stain, ACROS Organics™
CAS: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Molecular Weight (g/mol): 466.346 MDL Number: MFCD00005723 InChI Key: KHLVKKOJDHCJMG-QDBORUFSSA-L Synonym: Acid Blue 74, C.I. 73015, 5, 5'-Indigodisulfonic acid, disodium salt PubChem CID: 5284351 IUPAC Name: disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate SMILES: C1=CC2=C(C=C1S(=O)(=O)[O-])C(=O)C(=C3C(=O)C4=C(N3)C=CC(=C4)S(=O)(=O)[O-])N2.[Na+].[Na+]
Cresyl Violet Acetate High Purity Biological Stain MP Biomedicals
CAS: 10510-54-0 Molecular Formula: C18H15N3O3 Molecular Weight (g/mol): 321.336 InChI Key: UEWSHLFAYRXNHZ-UHFFFAOYSA-N Synonym: Cresyl Fast Violet, 9-Amino-5-imino-5H-benzo[a]phenoxazine acetate salt PubChem CID: 44134641 IUPAC Name: acetic acid;5-iminobenzo[a]phenoxazin-9-amine SMILES: CC(=O)O.C1=CC=C2C(=C1)C(=N)C=C3C2=NC4=C(O3)C=C(C=C4)N
Crystal Violet, pure, indicator, ACROS Organics™
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 MDL Number: MFCD00011750 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M Synonym: Basic Violet 3, C.I. 42555, Gentian Violet PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Crystal Violet Indicator, 0.1% (w/v) in Glacial Acetic Acid, for Nonaqueous Titrations, Ricca Chemical
CAS: 64-19-7 Molecular Formula: C2H4O2 Molecular Weight (g/mol): 60.052 InChI Key: QTBSBXVTEAMEQO-UHFFFAOYSA-N PubChem CID: 176 ChEBI: CHEBI:15366 IUPAC Name: acetic acid SMILES: CC(=O)O
Alfa Aesar™ Crystal Violet, ACS, 90+%
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 MDL Number: MFCD00011750 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M Synonym: Basic Violet 3; C.I. 42555 PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Alfa Aesar™ Cresyl Violet acetate
CAS: 10510-54-0 Molecular Formula: C18H15N3O3 Molecular Weight (g/mol): 321.336 MDL Number: MFCD00013151 InChI Key: UEWSHLFAYRXNHZ-UHFFFAOYSA-N Synonym: 9-Amino-5-imino-5H-benzo[a]phenoxazine acetate salt; Cresyl echt violet PubChem CID: 44134641 IUPAC Name: acetic acid;5-iminobenzo[a]phenoxazin-9-amine SMILES: CC(=O)O.C1=CC=C2C(=C1)C(=N)C=C3C2=NC4=C(O3)C=C(C=C4)N
Indigo Carmine (Certified Biological Stain), Fisher Chemical
CAS: 860-22-0 Molecular Formula: C16H8N2Na2O8S2 Molecular Weight (g/mol): 466.346 MDL Number: MFCD00005723 InChI Key: KHLVKKOJDHCJMG-QDBORUFSSA-L Synonym: Acid Blue 74, FD&C Blue No. 2 PubChem CID: 5284351 IUPAC Name: disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate SMILES: C1=CC2=C(C=C1S(=O)(=O)[O-])C(=O)C(=C3C(=O)C4=C(N3)C=CC(=C4)S(=O)(=O)[O-])N2.[Na+].[Na+]
Gentian Violet, 2% (w/v) Aqueous Solution, Ricca Chemical
CAS: 548-62-9 Molecular Formula: C25H30ClN3 Molecular Weight (g/mol): 407.986 InChI Key: ZXJXZNDDNMQXFV-UHFFFAOYSA-M PubChem CID: 11057 ChEBI: CHEBI:41688 IUPAC Name: [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride SMILES: CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Bromocresol Purple, sodium salt, pure, water soluble, indicator, ACROS Organics™
CAS: 62625-30-3 Molecular Formula: C21H15Br2NaO5S Molecular Weight (g/mol): 562.204 MDL Number: MFCD00148896 InChI Key: KVSJYIXUWNOWBD-UGAWPWHASA-M Synonym: BCP, Bromcresol Purple, sodium salt, 5';, 5';';-Dibromo-o-cresolsulfonephthalein, sodium salt PubChem CID: 23665568 IUPAC Name: sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate SMILES: CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+]
  spinner