Tissue Markers and Dyes
Thermo Scientific™ Richard-Allan Scientific™ Mark-It™ Tissue Marking Dyes Available on GSA/VA Contract for Federal Government customers only.
For a variety of tissue identification purposes use Thermo Scientific™ Richard-Allan Scientific™ Mark-It™ Tissue Marking Dyes.
eBioscience™ Fixable Viability Dye eFluor™ 660 Non-distribution product as customer accommodation.
Can be used to irreversibly label dead cells prior to cryopreservation, fixation and/or permeabilization procedures. Unlike 7-AAD and PI, cells labeled with Fixable Viability Dyes can be washed, fixed, permabilized and stained for intracellular antigens without loss of staining intensity.
Cancer Diagnostics, Inc.™ Tissue Marking Dyes
Formulated to be used without a mordant
Thermo Scientific™ Shandon™ Tissue-Marking Dyes
Permanently mark the margins of excised surgical specimens for exact orientation with Thermo Scientific™ Shandon™ Tissue-Marking Dyes.
General Data Healthcare™ TMD™-7 Tissue Marking Dyes Refills
Refills for the TMD-7 Tissue Marking Dyes 7 Color Kit
EMD Millipore™ Weigert's Iron Hematoxylin kit Available on GSA/VA Contract for Federal Government customers only.
For staining nuclei of tissue sections
EMD Millipore™ HARLECO™ Nuclear Fast Red Available on GSA/VA Contract for Federal Government customers only.
For use in immunohistochemistry
EMD Millipore™ Hematoxylin Weigert No.1 Available on GSA/VA Contract for Federal Government customers only.
Used in conjunction with Weigert iron chloride
EMD Millipore™ HARLECO™ Schiff Reagent Available on GSA/VA Contract for Federal Government customers only.
For identification of carbohydrates functional groups, fungi, mucins, glycogen and immune complexes in tissue
Bradley Products™ Davidson™ Marking System Dyes
Visible, reliable and safe dyes for marking peripheral tumor or surgical margins
General Data Healthcare™ TMD™-7 Tissue Marking Dyes 7 Color Kit
Provides superior adherence to tissue surfaces
Novus Biologicals™ Mini Elite PAP Pen
Circle made by PAP pen is insoluble in alcohol and acetone
  spinner