Tissue Decalcifiers
Thermo Scientific™ Richard-Allan Scientific™ Cal-Rite™ Decalcifying/Fixation Solution Available on GSA/VA Contract for Federal Government customers only.
Streamline processes with Thermo Scientific™ Richard-Allan Scientific™ Cal-Rite™ Decalcifying/Fixation Solution, which allows simultaneous decalcification and fixation.
Thermo Scientific™ Richard-Allan Scientific™ Decalcifying Solution Edge Available on GSA/VA Contract for Federal Government customers only.
Remove calcium from histological specimens with Thermo Scientific™ Richard-Allan Scientific™ Decalcifying Solution.
Thermo Scientific™ Shandon™ TBD-1™ Decalcifier
Process bone marrow biopsies in as little as 15 to 30 minutes with this rapid hydrochloric acid decalcifier.
National Diagnostics™ Calci-Clear™ Rapid Decalcifying Solution Non-distribution product as customer accommodation.
High-speed decalcifier that permits the processing of tissue samples in considerably less time
National Diagnostics™ Calci-Clear™ Decalcifying Solution Non-distribution product as customer accommodation.
A solution of efficient chelating and sequestering agents used with tissue samples to remove and bind all calcium and other metal ions
  spinner