Tissue Fixatives
Thermo Scientific™ Richard-Allan Scientific™ Neutral Buffered Formalin (10%) Available on GSA/VA Contract for Federal Government customers only.
Reduce exposure to toxic fumes with the premixed Thermo Scientific™ Richard-Allan Scientific™ Neutral Buffered Formalin (10%). AFIP formulated.
Thermo Scientific™ Richard-Allan Scientific™ Cytoseal™ 60 and 280 Available on GSA/VA Contract for Federal Government customers only.
Prevent cracking, fading and discoloration of stained specimens with Thermo Scientific™ Richard-Allan Scientific™ Cytoseal™ 60 and 280 mounting media.
Invitrogen™ Molecular Probes™ ProLong™ Gold Antifade Mountant Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Liquid mountant applied directly to fluorescently labeled cell or tissue samples on microscope slides
Thermo Scientific™ Richard-Allan Scientific™ Cytoseal™ XYL Available on GSA/VA Contract for Federal Government customers only.
Inhibit the fading or yellowing of stained specimens with specially formulated XYL mounting media.
Thermo Scientific™ Richard-Allan™ Parapel™ Paraffin Repellent Available on GSA/VA Contract for Federal Government customers only.
Protect against excess buildup of wax on countertops and instrumentation with Thermo Scientific™ Richard-Allan Scientific™ Parapel™ Paraffin Repellent.
Invitrogen™ Molecular Probes™ Image-iT™ FX Signal Enhancer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Apply directly to slides or coverslips containing fixed and permeabilized cell or tissue samples prior to staining with fluorescent probes
Invitrogen™ eBioscience™ Intracellular Fixation and Permeabilization Buffer Set Non-distribution product as customer accommodation.
Designed for use in intracellular staining and flow cytometric analysis and has been specially formulated to reduce non-specific staining of fluorochrome-labelled antibodies and increase fluorescence signal-to-noise ratios.
Thermo Scientific™ ClearVue™ Mountants
Enjoy compatibility with a variety of stainers to accommodate any lab setting with Thermo Scientific™ ClearVue™ Mountants.
Invitrogen™ Molecular Probes™ SlowFade™ Gold Antifade Mountant Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Protects fluorescent dyes from fading (photobleaching) during fluorescence microscopy experiments
BD Phosflow™ Fix Buffer I Available on GSA/VA Contract for Federal Government customers only.
For simultaneous fixation and permeabilization of cells prior to intracellular staining
Thermo Scientific™ Richard-Allan Scientific™ Signature Series™ Pen-Fix™ Available on GSA/VA Contract for Federal Government customers only.
Achieve fixation and dehydration of specimens simultaneously with Thermo Scientific™ Richard-Allan Scientific™ Signature Series™ Pen-Fix™
Thermo Scientific™ Shandon™ Glyo-Fixx™ Available on GSA/VA Contract for Federal Government customers only.
Obtain superior results to neutral buffered formalin with Thermo Scientific™ Shandon™ Glyo-Fixx™, a noncarcinogenic, nonsensitizing, and environmentally friendly substitute.
Thermo Scientific™ Richard-Allan Scientific™ Buffered Zinc Formalin Available on GSA/VA Contract for Federal Government customers only.
Enhance nuclear detail in immunohistochemistry techniques with Thermo Scientific™ Richard-Allan Scientific™ Buffered Zinc Formalin.
Dynamic Diagnostics TransPak™ Pre-filled 10% Neutral Buffered Formalin (NBF)
Histology reagent - Pre-filled 10% NBF (Available in different volumes)
Thermo Scientific™ Shandon™ Xylene Substitute Mountant Available on GSA/VA Contract for Federal Government customers only.
Employ all routine xylene-substitute clearing agents with the Thermo Scientific™ Shandon™ Xylene Substitute Mountant.
Scigen O.C.T. Compound Cryostat Embedding Medium
Provides convenient specimen matrix for cryostat sectioning at temperatures of -10°C and below. It is a formulation of 10.24% polyvinyl alcohol, 4.26% polyethylene glycol and 85.5% nonreactive ingredients, being water soluble; it leaves no residue during staining.
Invitrogen™ Molecular Probes™ SlowFade™ Antifade Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Protects dyes from fading (photobleaching) during fluorescence microscopy experiments
Thermo Scientific™ Shandon™ Formal-Fixx™ 10% Neutral Buffered Formalin Available on GSA/VA Contract for Federal Government customers only.
Produce consistent results and extend life of rotary valve in tissue processors with Thermo Scientific™ Shandon™ Formal-Fixx™ 10% Neutral Buffered Foramalin.
Thermo Scientific™ Shandon™ Rapid-Fixx™ Formalin Available on GSA/VA Contract for Federal Government customers only.
Obtain quick results with this formalin. Excellent for antigenic preservation, avoids the need for enzyme digestion, and rapidly penetrates into tissue.
Thermo Scientific™ Shandon™ Shandon-Mount™ Available on GSA/VA Contract for Federal Government customers only.
Perform quick, high-quality conventional coverslipping with Thermo Scientific™ Shandon™ Shandon-Mount™ permanent mounting medium.
Thermo Scientific™ Richard-Allan Scientific™ Fast Flex™ Holding Solution
Employ a fixative designed for use in the grossing room with the Thermo Scientific™ Richard-Allan Scientific™ Fast Flex™ Holding Solution.
Invitrogen™ Molecular Probes™ ProLong™ Antifade Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Allows longer-term storage of the sample
Fisherbrand™ SlideRite™ Cytology Fixative Available on GSA/VA Contract for Federal Government customers only.
A water soluble fixative for use in exfoliative cytology
Azer Scientific Pre-Filled 10% Neutral Buffered Formalin
Designed specifically for the collection and transport of various sizes of tissue samples
BD Pharmingen™ IHC Zinc Fixative Available on GSA/VA Contract for Federal Government customers only.
An alternative to formalin fixation of tissue for paraffin embedded sections
Thermo Scientific™ Richard-Allan Scientific™ Fast-Flex™ Processing Reagents
Improve the quality of processed tissue with these processing reagents, which can either be ordered with a xylene-based or a xylene-substitute-based clearant.
Thermo Scientific™ Shandon™ Consul-Mount™ Available on GSA/VA Contract for Federal Government customers only.
Designed specifically for use with the Thermo Scientific Shandon Consul™ coverslipper
Thermo Scientific™ Shandon™ Zinc Formal-Fixx™ Available on GSA/VA Contract for Federal Government customers only.
Acquire amazing results with Thermo Scientific™ Shandon™ Zinc Formal-Fixx™, which is excellent for immunochemistry.
  spinner