Tissue Pretreatment Reagents
Thermo Scientific™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (10X)
Improve the reactivity of many antibodies in formalin-fixed tissues with the Thermo Scientific™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (10X).
Thermo Scientific™ Lab Vision™ Dewax and HIER Buffer L,M,H (IVD)
Thermo Scientific Dewax and Heat Induced Epitope Retrieval (HIER) Buffers simultaneously automate deparaffinization, rehydration and epitope retrieval in the Thermo Scientific PT Module.
Thermo Scientific™ Lab Vision™ EDTA Buffer for Heat-Induced Epitope Retrieval (10X)
Improve the reactivity of many antibodies in formalin-fixed tissues with Thermo Scientific™ Lab Vision™ EDTA Buffer for Heat-Induced Epitope Retrieval (10X).
Thermo Scientific™ Lab Vision™ HIER Buffer L (100X)
Perform heat induced epitope retrieval (HIER) on formalin-fixed paraffin-embedded tissue sections using the Thermo Scientific™ Lab Vision™ HIER Buffer L (100X).
Thermo Scientific™ Lab Vision™ Pepsin Solution for Enzyme-Induced Epitope Retrieval
Improve the immunoreactivity of many antigens in formaldehyde fixed tissues with Thermo Scientific™ Lab Vision™ Pepsin Solution for Enzyme-Induced Epitope Retrieval.
BD Retrievagen Antigen Retrieval Systems
Recommended for zinc-fixed and formalin-fixed paraffin embedded tissue sections that show suboptimal or no reactivity with antibodies for immunohistochemical staining
R&D Systems™ Antigen Retrieval Reagent, Basic Available on GSA/VA Contract for Federal Government customers only.
Antigen retrieval system using heat-induced recovery of cell and tissue antigens
R&D Systems™ Antigen Retrieval Reagent, Universal Available on GSA/VA Contract for Federal Government customers only.
An antigen-retrieval system using heat-induced recovery of cell and tissue antigens
Thermo Scientific™ Lab Vision™ Protease XXV for Enzyme-Induced Epitope Retrieval
Improve the immunoreactivity of certain antigens in formaldehyde-fixed tissues with Thermo Scientific™ Lab Vision™ Protease XXV for Enzyme-Induced Epitope Retrieval.
Thermo Scientific™ Lab Vision™ Tris-HCl Buffer for Heat-Induced Epitope Retrieval, (10X)
Improve the reactivity of certain antibodies in formalin-fixed tissues with Thermo Scientific™ Lab Vision™ Tris-HCl Buffer for Heat-Induced Epitope Retrieval (10X).
Thermo Scientific™ Lab Vision™ HIER Buffer M (100X)
Perform heat induced epitope retrieval (HIER) on formalin-fixed paraffin-embedded tissue sections using the Thermo Scientific™ Lab Vision™ HIER Buffer M (100X)
Thermo Scientific™ Lab Vision™ HIER Buffer H (100X)
Perform heat induced epitope retrieval (HIER) on formalin-fixed paraffin-embedded tissue sections using the Thermo Scientific™ Lab Vision™ HIER Buffer H (100X).
R&D Systems™ Antigen Retrieval Reagent, Acidic
An antigen retrieval system using heat-induced recovery of cell and tissue antigens
R&D Systems™ Antigen Retrieval Reagent Sampler Available on GSA/VA Contract for Federal Government customers only.
Contains three antigen retrieval reagents in a single pack
  spinner