Salmonella Testing Supplies
Thermo Scientific™ Wellcolex™ Color Salmonella Rapid Latex Agglutination Test Kit Available on GSA/VA Contract for Federal Government customers only.
Save up to two days over traditional screening methods with this easy-to-use, cost effective latex agglutination test for rapid, accurate identification of Salmonella.
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O Available on GSA/VA Contract for Federal Government customers only.
Used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H Available on GSA/VA Contract for Federal Government customers only.
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Thermo Scientific™ Remel™ Novobiocin Disk Available on GSA/VA Contract for Federal Government customers only.
Use for differentiation of Staphylococcus saprophyticus from other coagulase-negative staphylococci.
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O Polyvalent Available on GSA/VA Contract for Federal Government customers only.
Used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O/Vi
Used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
  spinner