Media Additives
Thermo Scientific™ Oxoid™ Bolton Broth Selective Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid Bolton Broth Selective Supplement is used for the selective pre-enrichment of Campylobacter spp. in food samples.
BD™ Bacto™ Dehydrated Agar Available on GSA/VA Contract for Federal Government customers only.
Purified solidifying agent.  BD Difco&trade Bacto™ Dehydrated Agar is purified to remove extraneous matter. 
Thermo Scientific™ Remel™ Egg Yolk Suspension 50% Available on GSA/VA Contract for Federal Government customers only.
Remel Egg Yolk Suspension 50% is used with McClung-Toabe Agar Base.
BD BBL™ Middlebrook OADC Enrichment
Used as a growth supplement for microbiology research.  BD Difco™ BBL™ Middlebrook OADC Enrichment is recommended for isolation and cultivation of mycobacteria and other microbes.
Thermo Scientific™ Oxoid™ Legionella BCYE Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid Legionella BCYE Supplement is used for the isolation of Legionella spp.
Thermo Scientific™ Oxoid™ Perfringens TSC Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid Perfringens TSC Supplement (D-cycloserine 200mg/vial) is used for the recovery of Clostridium perfringens.
BD Difco™ Dehydrated Culture Media: Granulated Agar
Used as a solidifying agent for culture media. BD Difco™ Dehydrated Culture Media: Granulated Agar is carefully monitored for cultural response, solubility and gelation temperature.
Thermo Scientific™ Oxoid™ Bacillus Cereus Selective Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid Bacillus Cereus Selective Supplement (Polymyxin B Supplement) is supplied 50,000 IU/Vial for inhibition of Gram-negative bacteria.
Thermo Scientific™ Remel™ Moxalactam Selective Supplement Available on GSA/VA Contract for Federal Government customers only.
Remel Moxalactam Selective Supplement is used with Modified Oxford Base.
BD Difco™ Peptone Water
For cultivation of nonfastidious organisms and studying carbohydrate fermentation
Thermo Scientific™ Oxoid™ Oxford Listeria Selective Supplement Available on GSA/VA Contract for Federal Government customers only.
The Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.
BD™ Difco™ Dehydrated Culture Media: Noble Agar
Used as a solidifying agent essentially free of impurities.  Difco™ Dehydrated Culture Media: Noble Agar is perfect for electrophoretic and nutritional procedures wherever increased purity is required.
Thermo Scientific™ Oxoid™ Legionella MWY Selective Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid Legionella MWY Selective Supplement is used for the isolation of Legionella pneumophila from potable water samples.
BD Difco™ Leptospira Enrichment EMJH
Used with Leptospira Medium Base EMJH in cultivating Leptospira. BD™ Difco™ Leptospira Enrichment EMJH is a lyophilized media additive. The solution contains albumin, polysorbate 80 and additional growth factors for Leptospira.
Thermo Scientific™ Remel™ Iodine for Tetrathionate
Thermo Scientific™ Remel Iodine for Tetrathionate is used with Tetrathionate Broth.
BD Butterfields Buffer Solution
For isolation and/or differentiation of microorganisms from sampls or specimens
BD BBL™ IsoVitaleX™ Enrichment
BD BBL™ IsoVitaleX™ Enrichment is a chemically defined supplement used as an additive to media for cultivation of nutritionally fastidious microorganisms.
Thermo Scientific™ Oxoid™ C-T Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid C-T Supplement is a chromogenic medium for the identification of Escherichia coli O157:H7.
BD Bacto™ Casamino Acids Available on GSA/VA Contract for Federal Government customers only.
Suitable for use in diphtheria toxin production.   BD Difco™ Bacto™ Casamino Acids comprise acid hydrolyzed casein having low sodium chloride and iron concentrations.
Thermo Scientific™ Oxoid™ Novobiocin Selective Supplement Available on GSA/VA Contract for Federal Government customers only.
Add to EC Broth, Part No. CM0990B; Muller-Kauffmann Tetrathionate-Novobiocin Broth Base, Brilliant Green Agar, Modified Tryptone Soya Broth and Lysine Iron Agar.
BD BBL™ Dehydrated Culture Media: Middlebrook ADC Enrichment
Used as a growth supplement for microbiology research.  BD Difco™ BBL™ Middlebrook ADC Enrichment is recommended for isolation and cultivation of mycobacteria and other microbes
Thermo Scientific™ Oxoid™ Legionella GVPC Selective Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid Legionella GVPC Selective Supplement is used for the isolation of Legionella spp. from environmental samples.
  spinner