C. difficile Testing
Thermo Scientific™ Xpect™ C. difficile Toxin A/B Test Available on GSA/VA Contract for Federal Government customers only.
Rapidly detect Clostridium difficile toxin A and B from human fecal samples with a simple, three-step procedure and results in 20 minutes.
Thermo Scientific™ ProSpecT™ C. difficile Toxin A/B Microplate Assay Available on GSA/VA Contract for Federal Government customers only.
Rapidly detect Clostridium difficile toxin A and B from human fecal samples.
Quidel Solana C. difficile AssayKit Available on GSA/VA Contract for Federal Government customers only.
in vitro diagnostic test for the direct, qualitative detection of the Clostridium difficile Toxin A gene (tcdA) in unformed stool specimens
Thermo Scientific™ Remel™ PRO Disk with Reagent Available on GSA/VA Contract for Federal Government customers only.
Used for identification of yeast and anaerobic microorganisms primarily Candida albicans and Clostridium difficile.
BD™ MAX™ Cdiff
BD MAX™ Cdiff is a test for patients suspected of having CDI through the detection of the C. difficile toxin B gene (tcdB) from liquid or soft stool.
Meridian Bioscience™ illumigene™ C. difficile Test Kit
Molecular assay for the detection of C. difficiles in stool
Quidel AmpliVue™ C. difficile Assay Kit
For qualitative detection and identification of toxigenic Clostridium difficile bacterial nucleic acids extracted from stool samples
Alere C. Diff Quik Chek™ Complete Available on GSA/VA Contract for Federal Government customers only.
For good infection control measures
Alere TOX A/B Quik Chek™ Kit Available on GSA/VA Contract for Federal Government customers only.
Rapid enzyme immunoassay cassette for detecting C. difficile toxins A and B in fecal specimens. Specimens can be run individually or batched.
Quidel Molecular C. difficile Control Set Available on GSA/VA Contract for Federal Government customers only.
To be used as full process controls in molecular testing
Quidel Solana RSV+hMPV Assay Kit
The Solana RSV + hMPV Assay is a rapid qualitative in vitro diagnostic test for the detection and differentiation of Respiratory Syncytial Virus (RSV) and Human Metapneumovirus (hMPV) viral RNA in nasal and nasopharyngeal swabs from patients with signs and symptoms of respiratory infection.
Meridian Bioscience™ ImmunoCard™ Toxins A&B Test Kit Available on GSA/VA Contract for Federal Government customers only.
Rapid EIA for the detection of Clostridium difficile toxins A and B in stool samples
Alere C. difficile Tox A/B Microplate Assay Available on GSA/VA Contract for Federal Government customers only.
Rapid ELISA test for qualitative detection of C. difficile antigens
Alere C. difficile Toxin and Antitoxin Kit
Uses a tissue culture format to detect the presence of cytotoxic activity (cell rounding) in fecal specimens and confirms the identification of C. difficile toxin by using specific antitoxin.
Meridian Bioscience™ Premier™ C. difficile Toxins A and B Microplate Assay Available on GSA/VA Contract for Federal Government customers only.
ELISA for the detection of C. difficile toxins A and B in stool specimens
Alere C. difficile Quik Chek™ Available on GSA/VA Contract for Federal Government customers only.
Rapid enzyme immunoassay for detecting glutamate dehydrogenase (GDH), also known as the C. difficile antigen, in stool specimens. Does not distinguish between toxigenic strains and those that cannot produce toxin.
Quidel Solana C. diff Startup Kit
The Quidel Solana C. difficile Assay is available with Virena, near real-time surveillance information to provide assessment of incidence throughout a health system to monitor C. diff infections.
Meridian Bioscience™ Premier™ C. difficile GDH
Enzyme immunoassay for detection of Clostridium difficile common antigen (GDH)
Meridian Bioscience™ Immuno Card™ C. difficile GDH
A rapid enzyme immunoassay for the detection of Clostridium difficile common antigen in stool
  spinner