H. pylori Testing
Fisher Healthcare™ Sure-Vue™ H. pylori Test Kit
For qualitative detection of antibodies to H. pylori in whole blood, serum and plasma
Thermo Scientific™ Remel™ Selective Rapid Urea Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Selective Rapid Urea is used for detection of Helicobacter pylori in gastric mucosal biopsy specimens.
Quidel QuickVue™ H. pylori g II™ Test Available on GSA/VA Contract for Federal Government customers only.
Rapid immunoassay for the simple, accurate and qualitative detection of IgG antibodies specific to H. pylori
Otsuka America BreathTek™ UBT H. pylori Test Kit
No special training needed — CLIA nonregulated
Meridian Bioscience™ Premier™ Platinum HpSA™ Plus Available on GSA/VA Contract for Federal Government customers only.
Patented technique uses a truly non-invasive method of detecting H. pylori antigens in human stool samples
Alere H PYLORI QUIK CHEK™ Stool Antigen Test
Alere H PYLORI QUIK CHEK™ Stool Antigen Test test detects H. pylori antigens in human stool and provides accurate results within 30 minutes
Sekisui Diagnostics OSOM™ H. pylori Tests
For the detection of H. Pylori antibodies
Alere Clearview™ H. pylori Test
Innovative test uses rapid, two-step technology for the direct detection of Helicobacter pylori antibodies
Alere Acceava™ H. pylori Kit Available on GSA/VA Contract for Federal Government customers only.
Helps to diagnose infection without an invasive procedure. Simple test for the diagnosis of Helicobacter pylori
Meridian Bioscience™ ImmunoCardHelicobacter pylori Test Kit Available on GSA/VA Contract for Federal Government customers only.
Easily convertible to a batchable system as test volume increases
Thermo Scientific™ Remel™ Clearview™ H. pylori
Rapid test for the qualitative detection of IgG antibodies to Helicobacter pylori (H. pylori) in whole blood, serum and plasma.
Polymedco™ Poly stat™ H. pylori test
For rapid qualitative detection of IgG antibodies specific to Helicobacter pylori in human serum, plasma and whole blood specimens
LifeSign Status H. Pylori Test Kit
Rapid immunoassay for the qualitative detection of the anti-helicobacter pylori antibody in human whole blood, serum or plasma
Alere H. PYLORI CHEK™ Stool Antigen ELISA Test
Alere H. PYLORI CHEK™ Stool Antigen ELISA Test detects H. pylori specific antigen in human stool and provides excellent performance in determining active infection before and after eradication therapy
Germaine™ Laboratories AimStep™ H. Pylori Rapid Cassette Test
Germaine™ Laboratories AimStep™ H. Pylori Rapid Cassette Test is for the qualitative detection of Helicobabacter pylori in serum, plasma, or whole blood
Diamedix™ Immunosimplicity™ H. pylori IgG Test Kit
Detects IgG antibodies to Helicobacter pylori antigen in human sera
  spinner