Haemophilis Testing
Thermo Scientific™ Oxoid™ V Factor Discs Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Oxoid V Factor Disc is used for identification of Haemophilus spp.
Thermo Scientific™ Oxoid™ X + V Factor Disc Available on GSA/VA Contract for Federal Government customers only.
Oxoid X + V Factor Disc is used for identification of Haemophilus spp.
Thermo Scientific™ Oxoid™ X Factor Discs Available on GSA/VA Contract for Federal Government customers only.
Perform identification of Haemophilus spp. with Thermo Scientific™ Oxoid X Factor Discs.
Thermo Scientific™ Remel™ Agglutinating Sera, Haemophilus influenzae Available on GSA/VA Contract for Federal Government customers only.
Used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Thermo Scientific™ Remel™ Microring™ XV System Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Microring XV is an identification system for Haemophilus spp. using X, V and XV factors plus porphyrin test.
Thermo Scientific™ Remel™ Phadebact™ Haemophilus Test
A slide co-agglutination test used for confirmatory identification of Haemophilus influenzae.
Thermo Scientific™ Oxoid™ V Factor Discs Available on GSA/VA Contract for Federal Government customers only.
For identification of Haemophilus sp.
Thermo Scientific™ Remel™ Haemophilus Identification Test Kit Available on GSA/VA Contract for Federal Government customers only.
Use for determination of growth requirements and porphyrin production by Haemophilus spp.
BD BBL™ Hemo ID Quad Plate, with Growth Factors Available on GSA/VA Contract for Federal Government customers only.
Separates various growth factors by media type. Isolates, differentiates and identifies Haemophilus species based on the X- and V-factor growth requirements and the hemolysis of horse blood.
Thermo Scientific™ Remel™ ALA Disk™ Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel ALA Disk is used for demonstration of porphyrin synthesis in Haemophilus spp.
Thermo Scientific™ Oxoid™ X & V Factor Discs
For identification of Haemophilus spp.
  spinner