Meningitis Testing
BD Directigen™ Meningitis Latex Test System Available on GSA/VA Contract for Federal Government customers only.
For qualitative detection of antigens to H. influenzae type b, S. pneumoniae, N. meningitidis groups A, B, C, Y or W135 and Escherichia coli K1 in CSF, serum or urine. Can also be used for qualitative detection of antigens to group B Streptococcus in CSF and serum.
  spinner