Salmonella Testing Supplies
Thermo Scientific™ Wellcolex™ Color Salmonella Rapid Latex Agglutination Test Kit Available on GSA/VA Contract for Federal Government customers only.
Save up to two days over traditional screening methods with this easy-to-use, cost effective latex agglutination test for rapid, accurate identification of Salmonella.
Thermo Scientific™ Remel™ Novobiocin Disk Available on GSA/VA Contract for Federal Government customers only.
Use for differentiation of Staphylococcus saprophyticus from other coagulase-negative staphylococci.
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O Polyvalent Available on GSA/VA Contract for Federal Government customers only.
Used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H Available on GSA/VA Contract for Federal Government customers only.
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O Available on GSA/VA Contract for Federal Government customers only.
Used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
LifeSign Status Mono Test Kit
Status Mono immunoassay for the qualitative detection of infectious mononucleosis heterophile antibodies in whole blood, serum or plasma
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O/Vi Available on GSA/VA Contract for Federal Government customers only.
Used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
Thermo Scientific™ Denka Seiken™ Agglutinating Sera Salmonella H-G
Denka Seiken Agglutinating Sera are packaged 2mL per vial.
Thermo Scientific™ Denka Seiken™ Agglutinating Sera Salmonella O Factor
Denka Seiken Agglutinating Sera are packaged 2mL per vial.
Thermo Scientific™ Denka Seiken™ Agglutinating Sera Salmonella V, Set 1
Denka Seiken Agglutinating Sera are packaged 2mL per vial.
Thermo Scientific™ Denka Seiken ™Agglutinating Sera, Salmonella H Slide Method
Denka Seiken™ Agglutinating Sera are packaged 2mL per vial.
Thermo Scientific™ Denka Seiken™ Agglutinating Sera Salmonella Polyvalent O
Denka Seiken Agglutinating Sera are packaged 2mL per vial.
  spinner