Dehydrated Media
Thermo Scientific™ Remel™ Buffered Peptone Water Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Buffered Peptone Water is used for the non-selective pre-enrichment of Salmonella spp. from foods.
Fisher BioReagents™ Agar, Powder / Flakes, Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
Used as a solidifying agent for culture media
BD Difco™ Dehydrated Culture Media: Demi-Fraser Broth Base
For use with Fraser broth supplement in selectively and differentially enriching Listeria, reduced Acriflavine and Nalidixic Acid levels
Fisher BioReagents™ Microbiology Media: LB Broth, Miller Available on GSA/VA Contract for Federal Government customers only.
Used for growing molecular biology strains of E. coli-containing recombinant plasmids
BD Bacto™ Tryptic Soy Broth (Soybean-Casein Digest Medium) Available on GSA/VA Contract for Federal Government customers only.
Conforms with USP and EP specifications. BD™ Bacto™ Tryptic Soy Broth (Soybean-Casein Digest Medium) is a dehydrated, general purpose media for isolation, detection and cultivation of wide variety of microorganisms.
BD BBL™ Trypticase™ Soy Broth (Soybean-Casein Digest Medium)
Conforms with USP and EP specifications. BD™ BBL™ Trypticase™ Soy Broth (Soybean-Casein Digest Medium) is a dehydrated, general purpose media for isolation, detection and cultivation of wide variety of microorganisms.
BD Difco™ Dehydrated Culture Media: UVM Modified Listeria Enrichment Broth
Selective enrichment for culturing L. monocytogenes
BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar) Available on GSA/VA Contract for Federal Government customers only.
Use dehydrated media for isolation, detection and cultivation of a wide variety of microorganisms. BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar) is supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.
BD Difco™ Dehydrated Culture Media: Potato Dextrose Agar Available on GSA/VA Contract for Federal Government customers only.
For cultivation of Yeasts and molds.  BD Difco™ Dehydrated Culture Media: Potato Dextrose Agar is supplied in dehydrated format.
Fisher BioReagents™ Microbiology Media: LB Agar, Miller (Powder) Available on GSA/VA Contract for Federal Government customers only.
Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
BD Difco™ Dehydrated Culture Media: Dey/Engley (D/E) Neutralizing Broth
Tests efficiency of disinfectants and sanitation.  BD Difco™ Dehydrated Culture Media: Dey/Engley (D/E) Neutralizing Broth is part of a large selection of dehydrated media.
BD Difco™ Dehydrated Culture Media: Lactobacilli MRS Agar
For cultivation of lactobacilli.  BD Difco™ Dehydrated Culture Media: Lactobacilli MRS Agar is part of a large selection of dehydrated media.
BD Difco™ Dehydrated Culture Media: Fluid Lactose Medium
For cultivation of Salmonella and coliform organisms in water, foods, dairy and pharmaceutical products
BD Difco™ Sabouraud Dextrose Agar
Rendered more selective for fungi by the addition of antimicrobics. BD™ Difco™ Sabouraud Dextrose Agar is a general-purpose medium used in the cultivation of yeasts, molds and aciduric microorganisms. It is also recommended for the testing of cosmetics and food.
Thermo Scientific™ Perfringens TSC Agar Base (Dehydrated) Available on GSA/VA Contract for Federal Government customers only.
Selectively isolate and presumptively identify Clostridium perfringensin food samples with this basal medium.
BD BBL™ Dehydrated Culture Media: Mueller Hinton II Broth (Cation-Adjusted)
For antimicrobial susceptibility testing of microorganisms by broth dilution methods.   BD Difco™ BBL™ Dehydrated Culture Media: Mueller Hinton II Broth (Cation-Adjusted) is part of a large selection of dehydrated media.
Fisher BioReagents™ Microbiology Media Additives: Tryptone Available on GSA/VA Contract for Federal Government customers only.
Additive for preparing bacterial nutrient media
BD Difco™ Nutrient Agar Available on GSA/VA Contract for Federal Government customers only.
For the cultivation of bacteria and for the enumeration of organisms in water, sewage, feces and other materials.
BD BBL™ Fluid Thioglycollate Dehydrated Culture Media
Used for sterility testing of viscous materials
BD Difco™ Dehydrated Culture Media: Fluid Thioglycollate Medium
For sterility testing of biologics and cultivation of anaerobes, aerobes and microaerophiles
Thermo Scientific™ Brilliance™ Cronobacter Sakazakii Agar (DFI formulation) (Dehydrated) Available on GSA/VA Contract for Federal Government customers only.
Differentiate and enumerate Cronobacter sakazakii from infant formula and other food samples with this chromogenic medium.
BD BBL™ Dehydrated Culture Media: Mannitol Salt Agar
For isolation of pathogenic staphylococci
BD™ Difco™ Dehydrated Culture Media: TAT Broth Base
For sterility testing of viscous materials, adapted to sterility of cosmetics
Fisher BioReagents™ Microbiology Media Additives: Yeast Extract Available on GSA/VA Contract for Federal Government customers only.
Additive for preparing bacterial nutrient media
BD BBL™ Prepared Bottled Media: Trypticase Soy Agar (Soybean-Casein Digest Agar)
Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms
Thermo Scientific™ Remel™ Tryptic Soy Broth Available on GSA/VA Contract for Federal Government customers only.
Remel Tryptic Soy Broth (TSB) (Soybean Casein Digest) (Dehydrated) is used for the cultivation of a wide variety of nonfastidious microorganisms.
BD Difco™ Yeast Nitrogen Base without Amino Acids
Used for classifying yeasts based on amino acid and carbohydrate requirements. BD™ Difco™ Yeast Nitrogen Base without Amino Acids is a dehydrated culture medium used in many molecular genetics applications. Prepared according to the formulas of Wickerham.
BD BBL™ Dehydrated Culture Media: Brain Heart Infusion
Used in cultivation of fastidious and nonfastidious microorganisms, including aerobic and anaerobic bacteria,from variety of clinical and nonclinical materials
Thermo Scientific™ Bolton Broth Base (Dehydrated) Available on GSA/VA Contract for Federal Government customers only.
Selectively pre-enrich Campylobacter spp. in food samples with this medium.
  spinner