Inoculating Loops and Needles
Thermo Scientific™ Nunc™ Disposable Loops and Needles Available on GSA/VA Contract for Federal Government customers only.
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Fisherbrand™ Disposable Inoculating Loops and Needles, Flexible Needle and Loop
For quantitative procedures such as sampling, serial dilutions and urine counts, as well as for bacterial inoculation. Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 9 in. (23cm) are molded from a specially formulated, biologically inert polymer.
Fisherbrand™ Disposable Inoculating Loops and Needles
Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation. Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 8 in. (20cm) are precision molded from a specially formulated, biologically inert polymer.
BD Difco™ Disposable Inoculating Needles and Loops Available on GSA/VA Contract for Federal Government customers only.
Sterile, disposable inoculating needles do not require flaming
Copan Diagnostics Calibrated Disposable Inoculating Loops and Needles in Zip Lock Pack Available on GSA/VA Contract for Federal Government customers only.
Calibrated 1 and 10μL plastic disposable inoculating loops and needles
Fisherbrand™ Nichrome/Aluminum Inoculating Loops
Fisherbrand Nichrome/Aluminum Inoculating Loops consist of a nichrome wire loop mounted to an aluminum handle. Inoculating loops are used to introduce microorganisms into a culture medium.
Thermo Scientific™ Nunc™ Disposable Loops and Needles, needle Available on GSA/VA Contract for Federal Government customers only.
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Globe Scientific Sterile Rigid Inoculating Loops with Needles
These disposable inoculation loops and needles offer the most convenient and safest way of streaking culture media. Molded from a special medical grade of polystyrene (PS), they offer a strong, rigid feel to provide comfortable, stable and uniform streaking.
Troemner™ Inoculating Turntable Available on GSA/VA Contract for Federal Government customers only.
Hand-operated inoculating turntables
Decon™ LeStreak™ Microstreakers Available on GSA/VA Contract for Federal Government customers only.
Perfect for inoculating or transferring microbiological samples
Bio Plas™ Astral™ Inoculation Device System
Economical, disposable inoculation system designed for safety and dependable sterility
Globe Scientific Sterile Rigid 10uL & 1uL Combo Inoculation Loops
These disposable inoculation loops and needles offer the most convenient and safest way of streaking culture media. Molded from a special medical grade of polystyrene (PS), they offer a strong, rigid feel to provide comfortable, stable and uniform streaking.
Thermo Scientific™ Nunc™ Disposable Loops and Needles, needle Available on GSA/VA Contract for Federal Government customers only.
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Leica Microsystems Covidien No. 26 Platinum-iridium Loops
Loop, Platinum-iridium, Covidien; Fused, 3mm I.D., 2-3/4in. long, Without holder
Decon™ LeLoop™ Inoculating Loops Available on GSA/VA Contract for Federal Government customers only.
More economical than platinum or disposable plastic loops
Thermo Scientific™ Reusable Calibrated Inoculating Loops Available on GSA/VA Contract for Federal Government customers only.
Select, inoculate or transfer a well-defined microbiological sample volume on media plates with these reusable calibrated inoculating loops.
Pro-Lab Diagnostics™ Pro-Loops™ Insulated Inoculating Loop Wire Holders
Available in nickel-plated brass or lightweight aluminum
Corning™ Sterile Polystyrene Inoculating Loops Available on GSA/VA Contract for Federal Government customers only.
Constructed of flexible polystyrene for optimum retrieval of an inoculum from a culture. Gosselin™ Sterile Polystyrene Inoculating Loops provide a superior solution for transferring microscopic organisms, inoculum, or streaking.
Decon™ LeStab™ Inoculating Microneedles Available on GSA/VA Contract for Federal Government customers only.
Perfectly balanced for easy manipulation
Thermo Scientific™ Remel™ Calibrated Inoculating Loops Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Calibrated Inoculating Loops come in a variety of materials: platinum, twisted nichrome and plastic.Some with color coded with insulated handle.
Decon™ LeLoop™ Holder Available on GSA/VA Contract for Federal Government customers only.
Holders for LeLoop Inoculating Loops
Bel-Art™ SP Scienceware™ Replica-Plating Tool and Replacement Velveteen Squares Available on GSA/VA Contract for Federal Government customers only.
Utilize a locking ring which secures sterile velveteen squares onto a PVC cylinder for easy control of the plating device
Leica Microsystems Inoculating Loops and Holders
Loops and holders designed for culturing, transferring, streaking or isolating tissue cultures in cytology or histology
Thermo Scientific™ Remel™ Microstreaker Loop
Thermo Scientific™ Remel Microstreaker is a twisted nichrome loop with handle.
Thermo Scientific™ Remel™ Microloop
Thermo Scientific™ Remel Microloop is a twisted nichrome loop without handle.
Thermo Scientific™ Remel™ Microstreaker Needle
Thermo Scientific™ Remel Microstreaker Needle is a straight nichrome needle with handle.
  spinner