Media Additives
Thermo Scientific™ Oxoid™ Bolton Broth Selective Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid Bolton Broth Selective Supplement is used for the selective pre-enrichment of Campylobacter spp. in food samples.
BD Bacto™ Dehydrated Agar Available on GSA/VA Contract for Federal Government customers only.
Purified solidifying agent.  BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter. 
BD Difco™ Peptone Water
For cultivation of nonfastidious organisms and studying carbohydrate fermentation
Thermo Scientific™ Oxoid™ Perfringens TSC Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid Perfringens TSC Supplement (D-cycloserine 200mg/vial) is used for the recovery of Clostridium perfringens.
BD BBL™ Middlebrook OADC Enrichment
Used as a growth supplement for microbiology research.  BD Difco™ BBL™ Middlebrook OADC Enrichment is recommended for isolation and cultivation of mycobacteria and other microbes.
BD Difco™ Dehydrated Culture Media: Granulated Agar
Used as a solidifying agent for culture media. BD Difco™ Dehydrated Culture Media: Granulated Agar is carefully monitored for cultural response, solubility and gelation temperature.
Thermo Scientific™ Remel™ Moxalactam Selective Supplement Available on GSA/VA Contract for Federal Government customers only.
Remel Moxalactam Selective Supplement is used with Modified Oxford Base.
BD Difco™ Dehydrated Culture Media: Noble Agar
Used as a solidifying agent essentially free of impurities.  Difco™ Dehydrated Culture Media: Noble Agar is perfect for electrophoretic and nutritional procedures wherever increased purity is required.
Thermo Scientific™ Remel™ Egg Yolk Suspension 50% Available on GSA/VA Contract for Federal Government customers only.
Remel Egg Yolk Suspension 50% is used with McClung-Toabe Agar Base.
Thermo Scientific™ Remel™ Iodine for Tetrathionate
Thermo Scientific™ Remel Iodine for Tetrathionate is used with Tetrathionate Broth.
Thermo Scientific™ Remel™ Pancreatic Digest of Casein Peptone Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Pancreatic Digest of Casein Peptone is a peptone used as a nutritional supplement in microbiological culture media.
Gibco™ CultureOne™ Supplement (100X) Non-distribution product as customer accommodation.
Easily added to conventional differentiation medium for differentiating NSCs to neurons. Gibco™ CultureOne™ Supplement enables superior neuronal cell cultures that improve downstream assays, accelerate maturation and can be seamlessly maintained for 5 weeks or longer.
BD Bacto™ Casamino Acids Available on GSA/VA Contract for Federal Government customers only.
Excellent supplement for media formulations where nitrogen requirements are minimal, including diphtheria toxin production. BD Bacto™ Casamino Acids is an animal-origin, acid hydrolysis of casein having low sodium chloride and iron concentrations.
Thermo Scientific™ Oxoid™ Legionella BCYE Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid Legionella BCYE Supplement is used for the isolation of Legionella spp.
Thermo Scientific™ Oxoid™ Bacillus Cereus Selective Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid Bacillus Cereus Selective Supplement (Polymyxin B Supplement) is supplied 50,000 IU/Vial for inhibition of Gram-negative bacteria.
BD BBL™ IsoVitaleX™ Enrichment
BD BBL™ IsoVitaleX™ Enrichment is a chemically defined supplement used as an additive to media for cultivation of nutritionally fastidious microorganisms.
Thermo Scientific™ Oxoid™ Chloramphenicol Selective Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid Chloramphenicol Selective Supplement assists in the isolation of yeast and molds associated with spoilage.
Thermo Scientific™ Oxoid™ Listeria Selective Enrichment Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid Listeria Selective Enrichment Supplement is used for the enrichment of Listeria spp.
Thermo Scientific™ Remel™ Beef Extract
Remel Beef Extract provides nutritional media supplementation, ideal for general bacteriological use.
BD BBL™ Middlebrook OADC Enrichment Available on GSA/VA Contract for Federal Government customers only.
Used in qualitative procedures for isolation and cultivation of mycobacteria.  BD Difco™ BBL™ Middlebrook OADC Enrichment is a dehydrated culture media.
BD BBL™ Dehydrated Culture Media: Middlebrook ADC Enrichment
Used as a growth supplement for microbiology research.  BD Difco™ BBL™ Middlebrook ADC Enrichment is recommended for isolation and cultivation of mycobacteria and other microbes
Thermo Scientific™ Oxoid™ Modified Bolton Broth Selective Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid Modified Bolton Broth Selective Supplement is for the selective pre-enrichment of Campylobacter spp. in food samples.
Thermo Scientific™ Oxoid™ Brilliance™ Listeria Differential Supplement
Oxoid Brilliance Listeria Differential Supplement is added to Brilliance Listeria Agar Base to isolate/differentiate Listeria spp. in food.
  spinner