Prepared Media Plates
Thermo Scientific™ Blood Agar (TSA with Sheep Blood) Medium Available on GSA/VA Contract for Federal Government customers only.
Grow a wide variety of microorganisms on this general purpose non-selective medium that includes sheep blood to promote haemolysis.
Thermo Scientific™ MacConkey Agar Medium Available on GSA/VA Contract for Federal Government customers only.
Detect and enumerate coliform organisms, E. coli, while isolating and detecting Salmonella and Shigella spp. in both pathological and food specimens.
Thermo Scientific™ Blood Agar (TSA with 5% Sheep Blood) / MacConkey Agar Plate Available on GSA/VA Contract for Federal Government customers only.
Grow fastidious organisms, detect and enumerate coliforms, and isolate and detect Salmonella and Shigella spp. on a single plate.
Thermo Scientific™ Remel™ Contact Plate Sterile Tryptic Soy Agar w/Lecithin, Polysorbate 80 (Irradiated) Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Contact Plate Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 (Irradiated) is for the cultivation of bacteria, yeast and fungi.
Thermo Scientific™ Burkholderia Cepacia Selective Agar Available on GSA/VA Contract for Federal Government customers only.
Selectively isolate and differentiate Burkholderia cepacia complex from respiratory secretions of patients with cystic fibrosis with this medium.
Thermo Scientific™ Columbia CNA Agar with Sheep Blood Medium Available on GSA/VA Contract for Federal Government customers only.
Isolate Gram-positive organisms from clinical samples with this selective media traditionally used to identify staphylococci, streptococci, and S. pneumoniae.
BD BBL™ Prepared Plated Media: Trypticase™ Soy Agar (TSA II™) with Sheep Blood Available on GSA/VA Contract for Federal Government customers only.
Used for cultivating fastidious microorganisms and for visualization of hemolytic reactions produced by many bacterial species
Thermo Scientific™ Mitchison 7H11 Selective Agar with Carbenicillin, Polymyxin B, Amphotericin B and Trimethoprim Available on GSA/VA Contract for Federal Government customers only.
Isolate and cultivate Mycobacterium species from specimen containing mixed flora with this selective medium.
Thermo Scientific™ Chocolate Agar Medium Available on GSA/VA Contract for Federal Government customers only.
Obtain fast and accurate results with a nutritious medium for the isolation of fastidious organisms Neisseria, Haemophilus, and Gardnerella vaginalis.
BD BBL™ Prepared Plated Media: CHROMagar™ MRSA II
For the qualitative direct detection of nasal colonization by methicillin-resistant Staphylococcus aureus (MRSA)
BD BBL™ Prepared Plated Media: MacConkey II Agar Available on GSA/VA Contract for Federal Government customers only.
Selective and differential media for the detection of coliform organisms and enteric pathogens
Thermo Scientific™ Remel™ HE Agar (Hektoen Enteric Agar) Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel HE Agar (Hektoen Enteric Agar) facilitates isolation and differentiation of gram-negative enteric microorganisms.
Thermo Scientific™ Middlebrook 7H11 Agar Available on GSA/VA Contract for Federal Government customers only.
Isolate and cultivate Mycobacterium species with this selective, ready-to-use medium.
Thermo Scientific™ Spectra™ MRSA Medium Available on GSA/VA Contract for Federal Government customers only.
Obtain fast and accurate results with a selective chromogenic medium for screening clinical samples for the presence of Methicillin Resistant Staphylococcus aureus.
BD BBL™ Prepared Plated Media: Mueller Hinton II Agar Available on GSA/VA Contract for Federal Government customers only.
For antimicrobial disc diffusion susceptibility testing of common, rapidly growing bacteria by the Bauer-Kirby method
Thermo Scientific™ Columbia Blood Agar with Sheep Blood Medium Available on GSA/VA Contract for Federal Government customers only.
Isolate and cultivate fastidious microorganisms with clearly visible haemolytic reactions (staphylococci and streptococci) with a medium containing sheep blood.
BD BBL™ Prepared Plated Media: Chocolate II Agar (GC II Agar with Hemoglobin and IsoVitaleX™) Available on GSA/VA Contract for Federal Government customers only.
For the isolation and cultivation of fastidious microorganisms, especially Neisseria and Haemophilus species, from a variety of clinical specimens
BD BBL™ Prepared Plated Media: Trypticase™ Soy Agar (Soybean-Casein Digest Agar) Available on GSA/VA Contract for Federal Government customers only.
Used for isolation and cultivation of non-fastidious and fastidious microorganisms
Thermo Scientific™ Sabouraud Dextrose Agar Available on GSA/VA Contract for Federal Government customers only.
Isolate, cultivate and differentiate dermatophytes, fungi and yeasts, such as Candida albicans, on a ready-to-use prepared media plate.
BD BBL™ Prepared Plated Media: Columbia CNA Agar with 5% Sheep Blood Available on GSA/VA Contract for Federal Government customers only.
Selective and differential media used for the isolation and differentiation of gram-positive microorganisms from clinical and nonclinical materials
Thermo Scientific™ Remel™ Baird-Parker Agar Available on GSA/VA Contract for Federal Government customers only.
Easily isolate and enumerate Staphylococcus aureus with highly selective and sensitive, ready-to-use media.
Thermo Scientific™ Remel™ Settling Plate Sterile R2A Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Settling Plate R2A Sterile is used for performing heterotrophic plate count on treated potable water.
Thermo Scientific™ Remel™ MEP Agar (Mannitol, Egg Yolk, Polymyxin B Agar, MYP Agar) Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel MEP Agar (Mannitol, Egg Yolk, Polymyxin B Agar, MYP Agar) facilitates selective and differential isolation of Bacillus species.
BD BBL™ Trypticase™ Soy Agar with Lecithin and Polysorbate 80 Sterile Pack Media - RODAC Plates
Plated media in gamma-irradiated packs for isolator systems—multiwrap packaging protects media from vaporized hydrogen peroxide exposure
Thermo Scientific™ Remel™ Settling Plate Tryptic Soy Agar Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Settling Plate Tryptic Soy Agar is used for the cultivation of bacteria, yeast and fungi.
Thermo Scientific™ Remel™ Contact I Tryptic Soy Agar w/Lecithin, Polysorbate 80 Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Contact I Tryptic Soy Agar w/Lecithin, Polysorbate 80 is used for the cultivation of bacteria, yeast and fungi.
Thermo Scientific™ Mueller Hinton Agar Available on GSA/VA Contract for Federal Government customers only.
Perform effortless Antimicrobial Susceptibility Testing (AST) using this medium which may be used in internationally recognized standard procedures.
BD Difco™ Buffered Listeria Enrichment Broth Base
Enrichment broth for cultivation of Listeria spp. from food
Thermo Scientific™ Remel™ Settling Plate Sterile Tryptic Soy Agar (Irradiated) Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Settling Plate Sterile Tryptic Soy Agar (Irradiated) is used for the cultivation of bacteria, yeast and fungi.
BD BBL™ Prepared Plated Media: CIN Agar (Yersinia Selective) Available on GSA/VA Contract for Federal Government customers only.
For the isolation of Yersinia enterocolitica from a variety of clinical and nonclinical specimens
  spinner