Prepared Medial Tubes
Thermo Scientific™ Saline 0.45% Solution Tubes Available on GSA/VA Contract for Federal Government customers only.
Reduce sample preparation time for automated identification and susceptibility systems using convenient, pre-filled and ready-to-use 0.45% saline tubes.
Thermo Scientific™ Remel™ Lysine Iron Agar (LIA) Available on GSA/VA Contract for Federal Government customers only.
Use for the differentiation of enteric microorganisms based on their ability to deaminate or decarboxylate lysine and produce hydrogen sulfide.
Thermo Scientific™ Remel™ Thioglycollate Medium w/Dextrose, Hemin, Vitamin K Available on GSA/VA Contract for Federal Government customers only.
General-purpose growth medium for cultivation of aerobic and anaerobic microorganisms.
EMD Millipore™ Nutrient Media Available on GSA/VA Contract for Federal Government customers only.
Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs
Thermo Scientific™ Remel™ Triple Sugar Iron Agar Available on GSA/VA Contract for Federal Government customers only.
Provides differentiation of gram-negative enteric bacilli based on carbohydrate fermentation and hydrogen sulfide production.
Thermo Scientific™ Remel™ Lowenstein-Jensen Gruft (LJ) w/penicillin, nalidixic acid Available on GSA/VA Contract for Federal Government customers only.
The Thermo Scientific™ Remel Lowenstein-Jensen Gruft (LJ) with penicillin, nalidixic acid for the selective isolation of Mycobacterium spp.
BD BBL™ Mycoflask™ Trypticase™ Soy Broth Prepared Media Available on GSA/VA Contract for Federal Government customers only.
Used for the cultivation of fastidious and nonfastidious microorganisms from a variety of clinical and nonclinical specimens
Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium Available on GSA/VA Contract for Federal Government customers only.
Reduce prep time while isolating aerobes, anaerobes and microaerophilic organisms with this liquid medium with formats available that meet international industry guidelines.
BD Difco™ Modified TSB Prepared Media with Casamino Acids
Used in qualitative procedures for the sterility test and for the enrichment and cultivation of aerobic microorganisms
EMD Millipore™ m-ColiBlue24™ Broth Culture Media Available on GSA/VA Contract for Federal Government customers only.
Simultaneously detects total coliforms and E. coli in water and beverages using standard membrane filtration equipment
BD BBL™ Mycoflask™ Prepared Lim Broth (Todd Hewitt with CNA) Media Available on GSA/VA Contract for Federal Government customers only.
For selective enrichment of group B Streptococci (Streptococcus agalactiae), especially from genital specimens
Thermo Scientific™ Todd Hewitt Broth with Colistin and Nalidixic Acid (LIM Broth) Available on GSA/VA Contract for Federal Government customers only.
Reduce media prep time while selectively isolating group B streptococci from clinical specimens containing mixed flora with pre-filled, ready-to-use tubes.
Thermo Scientific™ Remel™ Mueller Hinton Broth Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Mueller Hinton Broth is for the cultivation of a wide variety of microorganisms.
Thermo Scientific™ Saline 0.85% Solution Tubes Available on GSA/VA Contract for Federal Government customers only.
Reduce diluent preparation time using convenient, pre-filled and ready-to-use 0.85% saline tubes.
Thermo Scientific™ Tryptic Soy Broth Available on GSA/VA Contract for Federal Government customers only.
Cultivate and isolate microorganisms with ease using this general purpose enrichment broth with formats available that meet USP guidelines for sterility testing.
BD BBL™ Prepared Lowenstein-Jensen Media Available on GSA/VA Contract for Federal Government customers only.
Used for cultivation of Mycobacterium tuberculosis and other mycobacterial species
BD BBL™ Mycoflask™ Seven H11 Agar Prepared Media Slants Available on GSA/VA Contract for Federal Government customers only.
Used in qualitative procedures for isolation and cultivation of mycobacteria
Thermo Scientific™ Remel™ Yeast Malt Extract Agar
Thermo Scientific™ Remel Yeast Malt Extract Agar provides isolation, detection, and enumeration of yeast and molds.
BD BBL™ Mycoflask™ Phosphate Buffer, pH 6.8, 600mL, P80
Used as a buffered diluent for microbiological procedures
BD BBL™ Mycoflask™ Prepared Tubed Media: Rappaport Vassiliadis Soy Salmonella Broth
For isolation and identification of microorganisms and for cultures requiring prolonged incubation
Thermo Scientific™ Remel™ Regan-Lowe Semi-Solid Transport Medium Available on GSA/VA Contract for Federal Government customers only.
Use for the transport and selective isolation of Bordetella pertussis and Bordetella parapertussis.
BD BBL™ Mycoflask™ Thioglycollate (Fluid) Prepared Media Available on GSA/VA Contract for Federal Government customers only.
Used for the sterility testing of biologics and for the cultivation of anaerobes, aerobes and microaerophiles
Thermo Scientific™ Selenite Broth Available on GSA/VA Contract for Federal Government customers only.
Reduce media preparation time and simultaneously enrich and isolate Salmonella spp. using convenient, pre-filled, ready-to-use tubes.
Thermo Scientific™ Lowenstein-Jensen Medium (LJ) Available on GSA/VA Contract for Federal Government customers only.
Reduce media preparation time and simultaneously cultivate and isolate Mycobacterium spp. using convenient, pre-filled, ready-to-use tubes.
BD BBL™ Mycoflask™ Prepared Tubed Media: BHI Broth Available on GSA/VA Contract for Federal Government customers only.
General-purpose liquid media used in the cultivation of fastidious and nonfastidious microorganisms, including aerobic and anaerobic bacteria, from a variety of clinical and nonclinical materials
BD BBL™ Mycoflask™ Thioglycollate Prepared Media Enriched with Vitamin K1 and Hemin Available on GSA/VA Contract for Federal Government customers only.
Ideal for use in the isolation and cultivation of fastidious or slow growing, obligately anaerobic microorganisms present in clinical materials
Thermo Scientific™ Remel™ PRAS Chopped Meat Broth (VPI) (Pre-Reduced Anaerobic Sterilized) (Chopped Meat Medium) Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel PRAS Chopped Meat Broth (VPI) (Pre-Reduced Anaerobic Sterilized) (Chopped Meat Medium) is used for cultivation of anaerobic bacteria.
Thermo Scientific™ Remel™ Urea Agar Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Urea Agar (Christensens) facilitates detection of urea hydrolysis by microorganisms.
BD BBL™ Mycoflask™ Phosphate Buffer, pH 6.8, 500mL Available on GSA/VA Contract for Federal Government customers only.
Used as buffered diluent for microbiological procedures
Thermo Scientific™ Remel™ GN Broth (Hajna) Available on GSA/VA Contract for Federal Government customers only.
Contains selective enrichment for isolation of gram-negative bacilli, especially Salmonella and Shigella.
  spinner