Tubes Prepared Microbiology Media
Thermo Scientific™ Saline 0.45% Solution Tubes Available on GSA/VA Contract for Federal Government customers only.
Reduce sample preparation time for automated identification and susceptibility systems using convenient, pre-filled and ready-to-use 0.45% saline tubes.
Thermo Scientific™ Remel™ Triple Sugar Iron Agar Available on GSA/VA Contract for Federal Government customers only.
Provides differentiation of gram-negative enteric bacilli based on carbohydrate fermentation and hydrogen sulfide production.
Thermo Scientific™ Remel™ Lysine Iron Agar (LIA) Available on GSA/VA Contract for Federal Government customers only.
Use for the differentiation of enteric microorganisms based on their ability to deaminate or decarboxylate lysine and produce hydrogen sulfide.
EMD Millipore™ Nutrient Media Available on GSA/VA Contract for Federal Government customers only.
Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs
Thermo Scientific™ Tryptic Soy Broth Available on GSA/VA Contract for Federal Government customers only.
Cultivate and isolate microorganisms with ease using this general purpose enrichment broth with formats available that meet USP guidelines for sterility testing.
Thermo Scientific™ Remel™ Lowenstein-Jensen Gruft (LJ) w/penicillin, nalidixic acid Available on GSA/VA Contract for Federal Government customers only.
The Thermo Scientific™ Remel Lowenstein-Jensen Gruft (LJ) with penicillin, nalidixic acid for the selective isolation of Mycobacterium spp.
Thermo Scientific™ Lowenstein-Jensen Medium (LJ) Available on GSA/VA Contract for Federal Government customers only.
Reduce media preparation time and simultaneously cultivate and isolate Mycobacterium spp. using convenient, pre-filled, ready-to-use tubes.
Thermo Scientific™ Saline 0.85% Solution Tubes Available on GSA/VA Contract for Federal Government customers only.
Reduce diluent preparation time using convenient, pre-filled and ready-to-use 0.85% saline tubes.
BD BBL™ Mycoflask™ Trypticase™ Soy Broth Prepared Media Available on GSA/VA Contract for Federal Government customers only.
Used for the cultivation of fastidious and nonfastidious microorganisms from a variety of clinical and nonclinical specimens
BD Difco™ Modified TSB Prepared Media with Casamino Acids
Used in qualitative procedures for the sterility test and for the enrichment and cultivation of aerobic microorganisms
Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium Available on GSA/VA Contract for Federal Government customers only.
Reduce prep time while isolating aerobes, anaerobes and microaerophilic organisms with this liquid medium with formats available that meet international industry guidelines.
BD BBL™ Mycoflask™ Prepared Lim Broth (Todd Hewitt with CNA) Media Available on GSA/VA Contract for Federal Government customers only.
For selective enrichment of group B Streptococci (Streptococcus agalactiae), especially from genital specimens
Thermo Scientific™ Remel™ Mueller Hinton Broth Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Mueller Hinton Broth is for the cultivation of a wide variety of microorganisms.
BD BBL™ Mycoflask™ Seven H11 Agar Prepared Media Slants Available on GSA/VA Contract for Federal Government customers only.
Used in qualitative procedures for isolation and cultivation of mycobacteria
BD BBL™ Prepared Lowenstein-Jensen Media Available on GSA/VA Contract for Federal Government customers only.
Used for cultivation of Mycobacterium tuberculosis and other mycobacterial species
BD BBL™ Mycoflask™ Prepared Tubed Media: BHI Broth Available on GSA/VA Contract for Federal Government customers only.
General-purpose liquid media used in the cultivation of fastidious and nonfastidious microorganisms, including aerobic and anaerobic bacteria, from a variety of clinical and nonclinical materials
EMD Millipore™ m-ColiBlue24™ Broth Culture Media Available on GSA/VA Contract for Federal Government customers only.
Simultaneously detects total coliforms and E. coli in water and beverages using standard membrane filtration equipment
Thermo Scientific™ Remel™ Yeast Malt Extract Agar
Thermo Scientific™ Remel Yeast Malt Extract Agar provides isolation, detection, and enumeration of yeast and molds.
Thermo Scientific™ Remel™ Rappaport-Vassiliadis Salmonella Enrichment Broth MLT Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Rappaport-Vassiliadis Salmonella Enrichment Broth MLT facilitates selective enrichment of Salmonella spp.
Thermo Scientific™ Remel™ Regan-Lowe Semi-Solid Transport Medium Available on GSA/VA Contract for Federal Government customers only.
Use for the transport and selective isolation of Bordetella pertussis and Bordetella parapertussis.
Thermo Scientific™ Remel™ GN Broth (Hajna) Available on GSA/VA Contract for Federal Government customers only.
Contains selective enrichment for isolation of gram-negative bacilli, especially Salmonella and Shigella.
BD BBL™ Mycoflask™ Thioglycollate Prepared Media Enriched with Vitamin K1 and Hemin Available on GSA/VA Contract for Federal Government customers only.
Ideal for use in the isolation and cultivation of fastidious or slow growing, obligately anaerobic microorganisms present in clinical materials
Thermo Scientific™ Remel™ PRAS Chopped Meat Broth (VPI) (Pre-Reduced Anaerobic Sterilized) (Chopped Meat Medium) Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel PRAS Chopped Meat Broth (VPI) (Pre-Reduced Anaerobic Sterilized) (Chopped Meat Medium) is used for cultivation of anaerobic bacteria.
BD Tubed Media Lauryl Sulfate Broth
For detection of coliform organisms in materials of sanitary importance like microbiological examination of water
BD BBL™ Mycoflask™ Thioglycollate (Fluid) Prepared Media Available on GSA/VA Contract for Federal Government customers only.
Used for the sterility testing of biologics and for the cultivation of anaerobes, aerobes and microaerophiles
Thermo Scientific™ Remel™ Motility Test Medium w/TTC Indicator Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Motility Test Medium with TTC Indicator is for motility testing of microorganisms.
Thermo Scientific™ Selenite Broth Available on GSA/VA Contract for Federal Government customers only.
Reduce media preparation time and simultaneously enrich and isolate Salmonella spp. using convenient, pre-filled, ready-to-use tubes.
Thermo Scientific™ Remel™ Tryptic Soy Broth w/15% Glycerol Available on GSA/VA Contract for Federal Government customers only.
For long-term cryopreservation of stock cultures
  spinner