Prepared Media Bottles
Thermo Scientific™ Remel™ Fluid K
Use as diluting or rinsing fluid for sterility testing on samples containing petrolatum.
Thermo Scientific™ Remel™ Fluid A Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Fluid A with 0.1% peptone (Peptone Fluid) is used as diluting or rinsing fluid for sterility testing.
Thermo Scientific™ Remel™ Butterfield's Phosphate Buffer Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Butterfield's Phosphate Buffer (pH 7.2) is used as a diluent in microbial limit testing of pharmaceutical products and food testing.
Thermo Scientific™ Remel™ Butterfield's 99mL Phosphate Buffer Available on GSA/VA Contract for Federal Government customers only.
Formulation is used in food, dairy and pharmaceutical testing as referenced in AOAC, APHA, FDA, USDA, and USP test methods.
Thermo Scientific™ Remel™ Tryptic Soy Agar Available on GSA/VA Contract for Federal Government customers only.
The Thermo Scientific™ Remel Tryptic Soy Agar (TSA) (Soybean Casein Digest Agar) is a general purpose medium for cultivation and isolation of microorganisms.
Corning™ Tryptic Soy Broth, Animal-Free, Liquid
For an accurate assessment of sterility throughout the manufacturing process
Thermo Scientific™ Remel™ Sabouraud Dextrose Broth 2% Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Sabouraud Dextrose Broth 2% (pH 5.6) (Fluid Sabouraud Medium) is used for cultivation and isolation of yeast and fungi.
BD Bottled Media Trypticase Soy Broth
Used in qualitative procedures for detection, isolation, and cultivation of fastidious and nonfastidious microorganisms
Thermo Scientific™ Remel™ Tryptic Soy Broth Available on GSA/VA Contract for Federal Government customers only.
The Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.
BD Sterility Bottles
Glass sterility bottles available with a variety of media
Thermo Scientific™ Remel™ R2A Agar Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel R2A Agar is used for performing heterotrophic plate count of treated potable water.
Thermo Scientific™ Remel™ MacConkey Broth Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel MacConkey Broth (MLT) is used for presumptive identification of coliforms using harmonized pharmacopial guidelines.
BD Prepared Bottled Media: Tryptic Soy Broth, Wide Mouth Bottle Edge
For the detection of microorganisms from pharmaceutical, food, environmental, or water testing samples
Thermo Scientific™ Remel™ Phosphate Buffer w/MgCl 99mL
Thermo Scientific™ Remel Phosphate Buffer w/MgCl 99mL (pH 7.2) formulation is used in water and wastewater testing as referenced in APHA and EPA test methods.
Thermo Scientific™ Remel™ Fluid D w/0.1% peptone, 0.1% Polysorbate 80
Thermo Scientific™ Remel Fluid D with 0.1% peptone, 0.1% Polysorbate 80 is used as diluting or rinsing fluid for sterility testing.
BD BBL™ Prepared Bottled Media: Buffered Sodium Chloride Peptone Solution (pH 7.0)
Used for dissolving, suspending and diluting test samples
Thermo Scientific™ Remel™ BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin Available on GSA/VA Contract for Federal Government customers only.
The Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Chloramphenicol and Gentamicin is used for selective isolation of fastidious fungi and yeast.
BD Bottled Media Thioglycollate
For the sterility testing of biologics
Thermo Scientific™ Remel™ Tryptic Soy Broth with 4% Polysorbate 20 and 0.5% Lecithin
Thermo Scientific™ Remel Tryptic Soy Broth with 4% Polysorbate 20 and 0.5% Lecithin is a general-purpose enrichment broth for cultivation/isolation of microorganisms.
BD Prepared Bottled Media: Reinforced Clostridial
For the detection of microorganisms from pharmaceutical, food, environmental, or water testing samples
Thermo Scientific™ Remel™ SP4 Glucose Broth Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel SP4 Glucose Broth (w/thallium acetate, penicillin) is used for isolation of Mycoplasma species.
Thermo Scientific™ Remel™ TAT Broth
Thermo Scientific™ Remel TAT Broth (w/Polysorbate 20) is used for sterility testing of highly viscous substances, such as salves, ointments and cosmetics.
Thermo Scientific™ Remel™ Thayer Martin Agar Improved Available on GSA/VA Contract for Federal Government customers only.
Use for isolation of Neisseria gonorrhoeae and Neisseria meningitidis.
Thermo Scientific™ Remel™ Tetrathionate Broth Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Tetrathionate Broth is used for selective enrichment and isolation of Salmonella species.
Thermo Scientific™ Remel™ Sabouraud Dextrose Agar, Emmons w/Chloramphenicol Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Sabouraud Dextrose Agar, Emmons with Chloramphenicol is used for selective isolation of yeast and fungi.
Thermo Scientific™ Remel™ Reinforced Medium for Clostridia MLT Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Reinforced Medium for Clostridia MLT facilitates cultivation and enumeration of Clostridium species and other anaerobes.
Thermo Scientific™ Remel™ Dermatophyte Test Medium Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Dermatophyte Test Medium (DTM) provides selective and differential isolation of pathogenic fungi from cutaneous sources.
Thermo Scientific™ Remel™ Phosphate Buffer w/1% Peptone Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Phosphate Buffer w/1% Peptone is used as a pre-enrichment for isolation of Salmonella from various food sources.
  spinner