Histological and Cytological Stains
Thermo Scientific™ Richard-Allan Scientific™ Signature Series Hematoxylin 7211 Available on GSA/VA Contract for Federal Government customers only.
Yield results that exceed those of Gill or Harris Hematoxylins with this uniquely formulated product.
Thermo Scientific™ Richard-Allan Scientific™ Signature Series Eosin-Y 7111 Available on GSA/VA Contract for Federal Government customers only.
Create an ideal visual contrast to Hematoxylin with the fast-acting Thermo Scientific™ Richard-Allan Scientific™ Signature Series Eosin-Y counterstain.
Thermo Scientific™ Richard-Allan Scientific™ Signature Series Hematoxylin 2 7231 Available on GSA/VA Contract for Federal Government customers only.
Yield visually similar results to Gills 2 and 3 with this uniquely formulated product.
Thermo Scientific™ Richard-Allan Scientific™ Signature Series Hematoxylin 1 7221 Available on GSA/VA Contract for Federal Government customers only.
Yield visually similar results to Gills 1 and 2 with this uniquely formulated product.
Thermo Scientific™ Richard-Allan Scientific™ Eosin-Y Alcoholic, Eosin-Y with Phloxine, Eosin-Y Saturated Available on GSA/VA Contract for Federal Government customers only.
Enjoy strict quality control of raw materials and finished product and a high quality product every time with the Thermo Scientific™ Richard-Allan Scientific™ Eosin-Y Alcoholic, Eosin-Y with Phloxine, and Eosin-Y Saturated.
Thermo Scientific™ Richard-Allan Scientific™ Masson Trichrome Kit Available on GSA/VA Contract for Federal Government customers only.
Aids in differentiating collagen and smooth muscle in tumors. Comes with all stains and solutions necessary to complete the staining procedure.
Thermo Scientific™ Richard-Allan Scientific™ Three-Step Stain Available on GSA/VA Contract for Federal Government customers only.
Obtain results on par with Wright-Giemsa with Thermo Scientific™ Richard-Allan Scientific™ Three Step Stain.
Thermo Scientific™ Shandon™ Kwik-Diff™ Stains Available on GSA/VA Contract for Federal Government customers only.
Enjoy a quicker alternative to traditional Wright's and Giemsa Staining procedures with Thermo Scientific™ Shandon™ Kwik-Diff™ Stains.
Invitrogen™ Trypan Blue Stain (0.4%) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
For use with the Countess™ Automated Cell Counter
Thermo Scientific™ Richard-Allan Scientific™ Modified Harris Hematoxylin
Use Thermo Scientific™ Richard-Allan Scientific™ Modified Harris Hematoxylin, a regressive nuclear stain, for both histology and cytology.
Thermo Scientific™ Richard-Allan Scientific™ Gill™ 1, 2, 3 Hematoxylin
With three strengths available, the Thermo Scientific™ Richard-Allan Scientific™ Gill™ 1, 2, 3 Hematoxylin allows complete control of staining intensities.
Thermo Scientific™ Richard-Allan Scientific™ Signature Series Cyto-Stain™ Available on GSA/VA Contract for Federal Government customers only.
Achieve results similar to conventional Papanicolaou stains with Thermo Scientific™ Richard-Allan Scientific™ Signature Series Cyto-Stain™, a uniquely formulated cytoplasmic stain
Azer Scientific 10% Neutral Buffered Formalin
Available in three convenient size configurations
Thermo Scientific™ Richard-Allan Scientific™ Modified Mayer's Hematoxylin, 1 pt. (0.47L) Available on GSA/VA Contract for Federal Government customers only.
Produce crisp nuclear staining with Thermo Scientific™ Richard-Allan Scientific™ Modified Mayer's Hematoxylin, a progressive nuclear stain.
Fisher HealthCare™ PROTOCOL™ Histological/Cytological Stains Available on GSA/VA Contract for Federal Government customers only.
Manufactured with strict quality control to ensure lot-to-lot uniformity
Thermo Scientific™ Richard-Allan Scientific™ Reticulin Silver Available on GSA/VA Contract for Federal Government customers only.
Identify and visualize reticular fibers in tissue sections with the Reticulin Silver Stain, a Gordon and Sweet's modification.
Thermo Scientific™ Shandon™ Bluing Reagent Available on GSA/VA Contract for Federal Government customers only.
Employ a gentle, pH-controlled solution for bluing hematoxylin with the Thermo Scientific™ Shandon™ Bluing Reagent. 
Thermo Scientific™ Shandon™ CytoRich™ Red Collection Fluid Available on GSA/VA Contract for Federal Government customers only.
Allow for more accurate evaluation of cytological features with Thermo Scientific™ Shandon™ CytoRich™ Red Collection Fluid, which lyses RBCs and solubilizes proteins.
Thermo Scientific™ Shandon™ Eosin Y Cytoplasmic Counterstain Available on GSA/VA Contract for Federal Government customers only.
Produce brilliant staining with Thermo Scientific™ Shandon™ Eosin-Y, a fast-acting cytoplasmic counterstain.
Sigma Diagnostics™ Poly-L-lysine Solution
Can be used in promoting cell adhesion to solid substrates
Fisher HealthCare™ Pinnacle Portfolio Clarifier™ 1, 2 Available on GSA/VA Contract for Federal Government customers only.
Unique acetic acid reagents, developed to supplement our family of Hematoxylins
Thermo Scientific™ Richard-Allan Scientific™ Gomori Trichrome (Blue Collagen) Available on GSA/VA Contract for Federal Government customers only.
Save time with Thermo Scientific™ Richard-Allan Scientific™ Gomori Trichrome (Blue Collagen), a one-step trichrome stain that achieves color differentiation similar to the Masson Trichrome.
Thermo Scientific™ Richard-Allan Scientific™ Modified Wright Stain Pack Available on GSA/VA Contract for Federal Government customers only.
Achieve optimum performance on Hema-Tek™ Slide Stainers with the Thermo Scientific™ Richard-Allan Scientific™ Stain Pack.
Thermo Scientific™ Shandon™ Harris Hematoxylin Available on GSA/VA Contract for Federal Government customers only.
Take advantage of acidified or non-acidified formulations with the Thermo Scientific™ Shandon™ Harris Hematoxylin. This product does not employ a mercury-based oxidizing agent, thus removing a possible source of environmental contamination.
BD Propidium Iodide Staining Solution
Designed for use in two-color Annexin V flow cytometric assays.
EMD Millipore™ PAS Staining Kit Available on GSA/VA Contract for Federal Government customers only.
For the detection of aldehyde and mucosubstances. PAS (periodic acid-Schiff) staining is one of the most widely used chemical methods in histology and is specific in staining non-substituted polysaccharides, neutral mucopolysaccharides, muco- and glycoproteins, as well as glycol- and phospholipids.
Sigma Diagnostics™ Bouin's Solution
Fixative for preserving soft and delicate structures
  spinner