Red Stains and Dyes and Indicators
Fuchsin Basic (Certified Biological Stain), Fisher Chemical
CAS: 632-99-5 Molecular Formula: C20H20ClN3 Molecular Weight (g/mol): 337.851 MDL Number: MFCD00001657 InChI Key: AXDJCCTWPBKUKL-UHFFFAOYSA-N Synonym: Magenta, Pararosaniline PubChem CID: 12447 IUPAC Name: 4-[(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride SMILES: CC1=CC(=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N)C=CC1=N.Cl
Phenol Red, Free Acid (Certified ACS), Fisher Chemical
CAS: 143-74-8 Molecular Formula: C19H14O5S Molecular Weight (g/mol): 354.376 MDL Number: MFCD00003552 InChI Key: BELBBZDIHDAJOR-UHFFFAOYSA-N Synonym: 4,4'-(3H-2,1-Benzoxathiol-3-yildene)bisphenol S,S-Dioxide, 4, 4'-(3H-2, 1-Benzoxathiol-3-yildene)bisphenol S PubChem CID: 4766 ChEBI: CHEBI:31991 IUPAC Name: 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol SMILES: C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)O
Nile Red 99%, ACROS Organics™
CAS: 7385-67-3 Molecular Formula: C20H18N2O2 Molecular Weight (g/mol): 318.376 MDL Number: MFCD00011639 InChI Key: VOFUROIFQGPCGE-UHFFFAOYSA-N Synonym: Nile blue A oxazone PubChem CID: 65182 ChEBI: CHEBI:52169 IUPAC Name: 9-(diethylamino)benzo[a]phenoxazin-5-one SMILES: CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=O)C=C3O2
Eosin Y (1% Solution in Water), Fisher Chemical
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 745.904 MDL Number: MFCD00005040 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
Alizarin Red S, pure, certified, ACROS Organics™
CAS: 130-22-3 Molecular Formula: C14H7NaO7S Molecular Weight (g/mol): 342.253 MDL Number: MFCD00013049 InChI Key: HFVAFDPGUJEFBQ-UHFFFAOYSA-M Synonym: 9,10-Dihydro-3,4-dihydroxy-9,10-dioxo-2-anthracenesulfonic acid, sodium salt, 9, 10-Dihydro-3, 4-dihydroxy-9, Mordant Red 3, Sodium alizarinesulfonate PubChem CID: 3955344 ChEBI: CHEBI:87358 IUPAC Name: sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate SMILES: C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]
Eosin Y (Certified Biological Stain), Fisher Chemical
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 745.904 MDL Number: MFCD00005040 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L Synonym: D&C Red No. 22, Acid Red 87 PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
Methyl Red, Sodium Salt (Certified ACS), Fisher Chemical
CAS: 845-10-3 Molecular Formula: C15H14N3NaO2 Molecular Weight (g/mol): 291.286 MDL Number: MFCD00002426 InChI Key: GNTPCYMJCJNRQB-UHFFFAOYSA-M Synonym: Acid Red 2, Sodium Salt PubChem CID: 4465632 IUPAC Name: sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate SMILES: CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
Congo Red (Certified Biological Stain), Fisher Chemical
CAS: 573-58-0 Molecular Formula: C32H22N6Na2O6S2 Molecular Weight (g/mol): 696.664 InChI Key: IQFVPQOLBLOTPF-UHFFFAOYSA-L Synonym: Diphenyl Bisazonaphthionic Acid, Direct Red 28 PubChem CID: 11313 ChEBI: CHEBI:34653 IUPAC Name: disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate SMILES: C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]
Invitrogen™ SYTOX™ Red Dead Cell Stain, for 633 or 635nm Excitation Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
A simple and quantitative single-step dead-cell indicator for use with red laser-equipped flow cytometers
SynaptoRed™ Reagent, Calbiochem™, EMD Millipore™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 162112-35-8 Molecular Formula: C30H45Br2N3 Molecular Weight (g/mol): 607.519 InChI Key: AFVSZGYRRUMOFH-UHFFFAOYSA-L Synonym: FM4-64, Frie Mao 4-64, N-(3-Triethylammoniumpropyl)-4-(p-diethylaminophenylhexatrienyl)-pyridinium, 2Br PubChem CID: 90489009 IUPAC Name: 3-[4-[(1Z,3E,5Z)-6-[4-(diethylamino)phenyl]hexa-1,3,5-trienyl]pyridin-1-ium-1-yl]propyl-triethylazanium;dibromide SMILES: CCN(CC)C1=CC=C(C=C1)C=CC=CC=CC2=CC=[N+](C=C2)CCC[N+](CC)(CC)CC.[Br-].[Br-]
Congo Red, Certified, Sigma-Aldrich™
CAS: 573-58-0 Molecular Formula: C32H22N6Na2O6S2 Molecular Weight (g/mol): 696.664 InChI Key: IQFVPQOLBLOTPF-UHFFFAOYSA-L Synonym: Cosmos Red PubChem CID: 11313 ChEBI: CHEBI:34653 IUPAC Name: disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate SMILES: C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]
Quinaldine red, pure, ACROS Organics™
CAS: 117-92-0 Molecular Formula: C21H23IN2 Molecular Weight (g/mol): 430.333 MDL Number: MFCD00011968 InChI Key: JOLANDVPGMEGLK-UHFFFAOYSA-M Synonym: 2-[4-(Dimethylamino)styryl]-1-ethylquinolinium iodide PubChem CID: 5702759 IUPAC Name: 4-[(E)-2-(1-ethylquinolin-1-ium-2-yl)ethenyl]-N,N-dimethylaniline;iodide SMILES: CC[N+]1=C(C=CC2=CC=CC=C21)C=CC3=CC=C(C=C3)N(C)C.[I-]
Nile Red Fluorescent stain for intracellular lipid droplets. MP Biomedicals
CAS: 7385-67-3 Molecular Formula: C20H18N2O2 Molecular Weight (g/mol): 318.376 InChI Key: VOFUROIFQGPCGE-UHFFFAOYSA-N Synonym: Nile Blue A Oxazone, 9-diethylamino-5H-benzo-alpha-phenoxazine-5-one PubChem CID: 65182 ChEBI: CHEBI:52169 IUPAC Name: 9-(diethylamino)benzo[a]phenoxazin-5-one SMILES: CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=O)C=C3O2
EOSIN Y, Ricca Chemical
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 745.904 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
Neutral Red, ACROS Organics™
CAS: 553-24-2 Molecular Formula: C15H17ClN4 Molecular Weight (g/mol): 288.779 MDL Number: MFCD00012651 InChI Key: PGSADBUBUOPOJS-UHFFFAOYSA-N Synonym: Basic Red 5, C.I. 50040, 3-Amino-7-dimethylamino-2-methylphenazine hydrochloride PubChem CID: 11105 ChEBI: CHEBI:86370 IUPAC Name: 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride SMILES: CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl
Alfa Aesar™ Brilliant Crocein
CAS: 5413-75-2 Molecular Formula: C22H16N4Na2O7S2 Molecular Weight (g/mol): 558.491 MDL Number: MFCD00003907 InChI Key: BMBHMIGLJQAUTJ-HKWNQDJZSA-N Synonym: C.I. 27290; Crocein Scarlet MOO PubChem CID: 88641610 IUPAC Name: (8Z)-7-oxo-8-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium SMILES: C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC(=CC(=C43)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
Eosin Y, 1% Solution in Water, ACROS Organics™
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 745.904 MDL Number: MFCD00005040 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L Synonym: 2',4',5',7'-Tetrabromofluorescein, disodium salt, Eosin Yellowish, C.I. 45380, Eosin Yellowish, 2', 4', 5' PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
Calcein-AM, Calbiochem™, EMD Millipore™
CAS: 148504-34-1 Molecular Formula: C46H46N2O23 Synonym: Calcein-acetoxymethyl Ester, Diacetate, CAL-AM
Neutral Red (Certified Biological Stain), Fisher Chemical
CAS: 553-24-2 Molecular Formula: C15H17ClN4 Molecular Weight (g/mol): 288.779 MDL Number: MFCD00012651 InChI Key: PGSADBUBUOPOJS-UHFFFAOYSA-N Synonym: Basic Red 5, Toluylene Red PubChem CID: 11105 ChEBI: CHEBI:86370 IUPAC Name: 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride SMILES: CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl
Acriflavine hydrochloride, MP Biomedicals™
CAS: 8063-24-9 Molecular Formula: C13H11N3 · HCl MDL Number: MFCD00069039 Synonym: Euflavin, Trypaflavine
Alfa Aesar™ Sulforhodamine 101 acid chloride
CAS: 82354-19-6 Molecular Formula: C31H29ClN2O6S2 Molecular Weight (g/mol): 625.151 MDL Number: MFCD00012406 InChI Key: MPLHNVLQVRSVEE-UHFFFAOYSA-N Synonym: Texas Red™; Sulforhodamine 101 sulfonyl chloride PubChem CID: 452705 ChEBI: CHEBI:51247 SMILES: C1CC2=C3C(=C4C(=C2)C(=C5C=C6CCC[N+]7=C6C(=C5O4)CCC7)C8=C(C=C(C=C8)S(=O)(=O)Cl)S(=O)(=O)[O-])CCCN3C1
Alfa Aesar™ Oil Red O
CAS: 1320-06-5 Molecular Formula: C26H24N4O Molecular Weight (g/mol): 408.505 MDL Number: MFCD00003898 InChI Key: HJAYEODIXUYVIC-LTFAAIOASA-N Synonym: C.I. 26125; Solvent Red 27 PubChem CID: 6046885 IUPAC Name: (1Z)-1-[[4-[(2,5-dimethylphenyl)diazenyl]-2,5-dimethylphenyl]hydrazinylidene]naphthalen-2-one SMILES: CC1=CC(=C(C=C1)C)N=NC2=C(C=C(C(=C2)C)NN=C3C(=O)C=CC4=CC=CC=C43)C
Eosin Y, pure, high purity, biological stain, ACROS Organics™
CAS: 17372-87-1 Molecular Formula: C20H12Br4Na2O8 Molecular Weight (g/mol): 745.904 MDL Number: MFCD00005040 InChI Key: MASXMTNVNASWNH-UHFFFAOYSA-L Synonym: 2',4',5',7'-Tetrabromofluorescein, disodium salt, Eosin Yellowish, 2', 4', 5' PubChem CID: 91886399 IUPAC Name: disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
Nile red, MP Biomedicals™
CAS: 7385-67-3 Molecular Formula: C20H18N2O2 Molecular Weight (g/mol): 318.376 MDL Number: MFCD00011639 InChI Key: VOFUROIFQGPCGE-UHFFFAOYSA-N Synonym: Nile Blue A Oxazone, 9-Diethylamino-5H-benzo[a]phenoxazine-5-one PubChem CID: 65182 ChEBI: CHEBI:52169 IUPAC Name: 9-(diethylamino)benzo[a]phenoxazin-5-one SMILES: CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=O)C=C3O2
  spinner