Sterilization Verification Indicators
Sartorius™ Sterisart™ NF Sterility Testing System
Provides completely closed system sterility testing of pharmaceuticals, open containers, closed containers, pre-filled syringes, powders, and medical devices. Sartorius™ Sterisart™ NF Sterility Testing System is optimal for use with isolators.
Mesa Labs Prospore™ Self-Contained Biological Indicator
Ideal for validating steam sterilization of liquids
STERIS VERIFY™ Bowie-Dick Test Pack
VERIFY™ Bowie-Dick Test Pack evaluate the performance of prevacuum sterilizers by confirming adequate air removal from the sterilizer chamber.
STERIS Dart™ Daily Air Removal Test
Dart™ Daily Air Removal Test is used in monitoring healthcare steam sterilizers.
Steris Corp™ VERIFY™ Biological Steam Test Pack
Steris Corp™ VERIFY™ Biological Steam Test Pack to confirm the performance of sterility equipment
STERIS Steraffirm™ Bowie-Dick Test Packs
Steraffirm™ Bowie-Dick Test Packs are designed to evaluate the performance of the air removal system of a pre-vacuum equipped sterilizer.
Propper Duo-Spore™ Sporestrips
Contain two test strips and one control strip
Propper Manufacturing Sterilization Autoclave Tape Edge
Propper sterilization tape is designed to distinguish processed and unprocessed items
Propper OK™ Steam Sterilization Monitor Strips Edge
Preprinted standard reference color technology eliminates guesswork
Steris Corp™ VERIFYtrade; Biological Steam Test Pack (SCBI) with Control
Steris Corp™ VERIFY™ Biological/Integrator Challenge Pack used to monitor and qualify autoclave cycles
Steris Corp™ VERIFY™ SCBI Vial Activator Set
Steris Corp™ VERIFY™ SCBI Vial Activator Set ensures proper release of growth medium from ampule, and spore disc
Steris Corp™ Biological Steam Test Pack and Vial Activator Bundle
Steris Corp™ Biological Steam Test Pack and Vial Activator Bundle for use with VERIFY™ Biological Indicators
Sartorius™ Universal Pump for Sterisart™ Sterility Testing System Available on GSA/VA Contract for Federal Government customers only.
Peristaltic pump for sample and media transfer into the Sterisart NF systems - Suitable for isolators
Steris Corp™ VERIFY™ Incubator Bundle
Steris Corp™ VERIFY™ Incubator Bundle for use with VERIFY™ Biological Indicators
  spinner