Sterilizer and Autoclave Repair and Replacement Components
Kinematica HMC Feed Water Connection
Automatic water feed connection.
Kinematica HMC Vertical Autoclave Air Filters
Exhaust air filter. Safety for individuals and the environment. Option for safe, emission-free sterilization of highly infectious germs and genetically modified materials. Not for applications requiring FDA approval.
Kinematica™ Sterilization Box
Sterilization Box for use with Fisherbrand STE40 Autoclave
Kinematica HMC Vertical Autoclave Air Cooling RL
Automatic air cooling decreaes turnaround time for busy applications.
Kinematica HMC Vertical Autoclave Temperature Sensors
For the sterilization of liquids. Extremely flexible, mechanically and chemically resistant. Quick response, encapsulated, steam-tight.
  spinner