Manual Glassware Washers and Dryers
Bel-Art™ SP Scienceware™ Lab-Aire™ II Electric Glassware Dryers: Single-Sided, 120V Available on GSA/VA Contract for Federal Government customers only.
Make labware maintenance easier with the SP Scienceware™ Lab-Aire™ II Electric Dryer&mdashperfect for drying glassware and plasticware quickly.
DWK Life Sciences Kimble™ KIMAX™ 5-Place NMR Tube Washers
These complete five-place washers use flexible PTFE tubing and a vacuum source (not included) to direct a power stream of wash solvent to thoroughly clean the insides of 7 and 8 in. NMR tubes.
Fisherbrand™ Portable Electric Labware Washer
Compact and installs in any sink in seconds with no special plumbing or electrical requirements
Bel-Art™ SP Scienceware™ Lab-Aire II™ Single-Sided Electric Benchtop Labware Dryers
Offer compact footprint and accelerates drying of labware with warm, circulating air.
Bel-Art™ SP Scienceware™ Lab-Aire™ II Electric Glassware Dryers: Double-Sided, 230V
Speed your drying time with the SP Scienceware Lab-Air™ II Electir Dryer.
Bel-Art™ SP Scienceware™ Lab-Aire™ II Electric Glassware Dryers: Double-Sided, 120V Available on GSA/VA Contract for Federal Government customers only.
Electric-powered drying racks for glassware and plasticware
Bel-Art™ SP Scienceware™ Lab-Aire™ II Electric Dryers: Double-Sided, 230 Products for Science Education
Speed your drying time with the SP Scienceware Lab-Air™ II Electir Dryer.
Bel-Art™ SP Scienceware™ Lab-Aire II™ Double-Sided Electric Benchtop Glassware Dryers Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Offers compact footprint and accelerates drying of labware with warm, circulating air.
Bel-Art™ SP Scienceware™ Lab-Aire II™ Single-Sided Electric Benchtop Glassware Dryers Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Offer compact footprint and accelerates drying of labware with warm, circulating air.
  spinner