Susceptibility Media
Invitrogen™ S.O.C. Ready-to-Use Medium Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used in the final step of bacterial cell transformation to obtain maximal transformation efficiency of E. coli
Gibco™ LB Broth, Ready-to-use Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Ready-to-use liquid growth media used for bacterial culture
Invitrogen™ Bluo-Gal, Substrate for β-galactosidase Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Alternative histochemical substrate for β-galactosidase that produces a darker blue color than X-Gal, the more frequently used substrate
Invitrogen™ imMedia™ Growth Medium, Agar, Ampicillin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Microwaveable, premixed, presterilized E. coli growth medium that can be prepared in just five minutes without autoclaving
Invitrogen™ Yeast Nitrogen Base Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Base medium for preparation of minimal and synthetic defined yeast media
Invitrogen™ LB Agar, Powder (Lennox L Agar), Autoclavable Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Formulation per one liter: 10g SELECT Peptone 140, 5g SELECT Yeast Extract, 5g sodium chloride, 12g SELECT Agar
Thermo Scientific™ Oxoid™ TST-RPLA Kit
Oxoid TST-RPLA Kit detects staphylococcal toxic-shock syndrome toxin (TST) by reversed passive latex agglutination.
Invitrogen™ X-Gal Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Histochemical substrate for β-galactosidase
Invitrogen™ LB Broth Base, Autoclavable Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Recommended for the general maintenance and growth of E. coli
Invitrogen™ imMedia™ Growth Medium, Agar, Kanamycin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Microwaveable, premixed, presterilized E. coli growth medium that can be prepared in just five minutes without autoclaving
Invitrogen™ imMedia™ Growth Medium, Ampicillin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Microwaveable, premixed, presterilized E. coli growth medium that can be prepared in just five minutes without autoclaving
Invitrogen™ imMedia™ Growth Medium, Kanamycin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Microwaveable, premixed, presterilized E. coli growth medium that can be prepared in just five minutes without autoclaving
Invitrogen™ imMedia™ Growth Medium, Agar, Ampicillin, X-Gal/IPTG Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Microwaveable, premixed, presterilized E. coli growth medium that can be prepared in just five minutes without autoclaving
Scientific Device McFarland Standards
Available in McFarland or Colorimeter Standard Sets
  spinner