Susceptibility Testing Systems
Thermo Scientific™ alamarBlue™ Dye
Allows three assays to be performed with just one dye, including cell growth determination, in vitro cytotoxicity and cytokine assays.
Thermo Scientific™ Remel™ Blank Paper Disk
Thermo Scientific™ Remel Blank Paper Disk can be impregnated with reagents for the detection, differentiation and identification of microorganisms.
BD BBL™ Prompt™ Inoculation System Available on GSA/VA Contract for Federal Government customers only.
For the rapid, inoculum standardization of 0.5 McFarland turbidity level of bacterial inocula using Kirby-Bauer procedures for antimicrobial susceptibility testing
BD BBL™ Crystal™ ID Systems Available on GSA/VA Contract for Federal Government customers only.
Miniaturized microbial ID systems use modified conventional, fluorogenic and chromogenic substrates
Thermo Scientific™ Oxoid™ Inoculum Water Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Oxoid Inoculum Water is for use with ANA MIC and FAS MIC Panels.
Hardy Diagnostics™ Coagulase Cyro™
The ready-to-use Coagulase Cryo is used to differentiate Staphylococcus species based on the ability to produce coagulase
  spinner