Cell Culture Dishes, Plates and Flasks
Corning™ 1L and 3L Disposable Plastic Spinner Flasks Available on GSA/VA Contract for Federal Government customers only.
Ready-to-use system comes with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly
Corning™ Disposable Polycarbonate Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear polycarbonate for optimum visibility and durability. Corning™ Disposable Polycarbonate Erlenmeyer Flasks are ideal for shaker cultures applications.
Corning™ U-Shaped Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Designed to improve usability while maintaining the same environment for cell growth as previous designs. Corning™ U-Shaped Cell Culture Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Corning™ TC-Treated Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Tissue-culture (TC) treated for optimum attachment and growth of anchorage-dependent cells. Corning™ TC-Treated Culture Dishes provide an ideal solution for growing and maintaining cells in vitro
Corning™ Cell Culture Treated Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear virgin polstyrene for optimum visibility and distortion-free microscopic viewing. Corning™ Cell Culture Treated Flasks are available in a variety of sizes, designs and cap styles to meet laboratory diagnostic needs.
Corning™ Disposable Spinner Flask with Top Cap and Angled Sidearms Available on GSA/VA Contract for Federal Government customers only.
Ready-to-use system comes with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly
Corning™ Costar™ Flat Bottom Cell Culture Plates Available on GSA/VA Contract for Federal Government customers only.
Tissue culture-treated plates for superior cell attachment. Corning™ Costar™ Flat Bottom Cell Culture Microplates provide an excellent device for diagnostic testing when cell adherence is essential.
PYREX™ Fernbach Flasks Available on GSA/VA Contract for Federal Government customers only.
Fernsbach culture flasks offer wide surface area is ideal for growth of organisms requiring a large surface area-to-volume ratio
Corning™ HYPERFlask™ M Cell Culture Vessels Available on GSA/VA Contract for Federal Government customers only.
Designed specifically for manual use, high-yield and high-performance. Specifically designed for manual use. Corning™ HYPERFlask™ M Cell Culture Vessel uses a multi-layer gas permeable growing surface for efficient gas exchange.
Corning™ Two- and Three-Liter Disposable Polycarbonate Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear polycarbonate for optimum visibility and durability. Corning™ Two-and Three-Liter Disposable Polycarbonate Erlenmeyer Flasks are ideal for shaker cultures applications.
PYREX™ Culture Media Flasks Available on GSA/VA Contract for Federal Government customers only.
Suitable for mixing and storing culture media
Corning™ Costar™ Ultra-Low Attachment Microplates Available on GSA/VA Contract for Federal Government customers only.
Featuring a covalently bound hydrogel layer that effectively inhibits cellular attachmment. Corning™ Costar™ Ultra-Low Attachment Microplates greatly reduce binding of attachment and serum proteins, minimizing cell attachment and spreading.
Corning™ CELLine Disposable Bioreactor Available on GSA/VA Contract for Federal Government customers only.
Cultivation flask for secreted product, recombinant protein expression and high-yield monoclonal antibody (MAb) production
DWK Life Sciences Kimble™ KIMAX™ Baffled Culture Flasks
For fermentation, cell culture growth and cultivation, and genetic engineering procedures
Fisherbrand™ Shaker Flasks
Reduce the risk of cross-contamination.
Corning™ Closed System Solutions Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Premium quality conical centrifuge tubes manufactured from high-quality, ultra-clear resins
DWK Life Sciences Wheaton™ CELLine™ Bioreactor Flask
Membrane cell culture flasks ideal for antibody and protein production
Corning™ Baffled Polycarbonate Flasks with Caps Available on GSA/VA Contract for Federal Government customers only.
Designed with baffles to provide premium agitation at lower speed for gentler, more efficient aeration. Corning™ Baffled Polycarbonate Flasks with Caps are ideal for shaker culture applications.
Falcon™ Polystyrene Microplates Available on GSA/VA Contract for Federal Government customers only.
Constructed of crystal grade virgin polystyrene for optimum visibility. Corning™ Falcon™ Polystyrene Microplates are available with tissue culture-treated and nontreated surfaces.
Falcon™ Standard Tissue Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Constructed of flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells. Corning Falcon™ Standard Tissue Culture Dishes works with most attachment-dependent cell lines.
Corning™ Falcon™ Cell Culture Multi Flasks Available on GSA/VA Contract for Federal Government customers only.
Grow cells faster and easier, increasing productivity in cell culture workflow. Corning™ Falcon™ Cell Culture Mini Flasks provide even distribution of media across all layers for homogeneous cell growth.
Corning™ 384-Well Solid Black or White Polystyrene Microplates Available on GSA/VA Contract for Federal Government customers only.
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Corning™ Closed System Solutions Preassembled Disposable Spinner Flasks Available on GSA/VA Contract for Federal Government customers only.
Ready to use with paddle and integrated magnet, eliminating need for time-consuming assembly or cleaning and reassembly
Thermo Scientific™ Nunc™ TripleFlask™ Treated Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Perform large cell mass applications such as drug screening and scale up with these cell-culture treated TripleFlasks.
Corning™ Polycarbonate 5L Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
Made of noncytotoxic USP Class VI polycarbonate that provides optical clarity and mechanical strength. Corning™ Polycarbonate 5L Erlenmayer Flask are ready-to-use for efficient scale-up of suspension cultures with space efficiency.
  spinner