Cell Culture Dishes
Corning™ TC-Treated Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Tissue-culture (TC) treated for optimum attachment and growth of anchorage-dependent cells. Corning™ TC-Treated Culture Dishes provide an ideal solution for growing and maintaining cells in vitro
Corning™ Ultra-Low Attachment Dishes Available on GSA/VA Contract for Federal Government customers only.
Unique covalently bonded hydrogel surface is hydrophilic and neutrally charged
Falcon™ Standard Tissue Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Constructed of flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells. Corning Falcon™ Standard Tissue Culture Dishes works with most attachment-dependent cell lines.
Corning™ Falcon™ Tissue Culture Dish with Grid Available on GSA/VA Contract for Federal Government customers only.
Features flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells. Corning™ Falcon™ Easy-Grip Tissue Culture Dishes are uniquely designed to simplify lifting one dish or a small stack of dishes.
Thermo Scientific™ BioLite Cell Culture Treated Dishes Available on GSA/VA Contract for Federal Government customers only.
Perform general cell culture procedures more economically with Thermo Scientific™ BioLite Cell Culture Treated Dishes, guaranteed to be sterile, non-pyrogenic and non-cytotoxic.
Falcon™ Organ Culture Dish Available on GSA/VA Contract for Federal Government customers only.
Vacuum-gas plasma tissue culture treated polystyrene dish
Thermo Scientific™ Nunc™ Cell Culture/Petri Dishes Available on GSA/VA Contract for Federal Government customers only.
Perform a variety of applications across a broad range of sizes and styles to suit a variety of experimental needs.
Thermo Scientific™ Nunc™ OmniTray™ Available on GSA/VA Contract for Federal Government customers only.
Screen libraries, DNA hybridization, standard petri dish techniques and bacterial growth with Thermo Scientific™ Nunc™ OmniTrays™.
Corning™ BioCoat™ Collagen I, Rat Tail Culture Dish Available on GSA/VA Contract for Federal Government customers only.
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Thermo Scientific™ Nunc™ MicroWell™ MiniTrays Available on GSA/VA Contract for Federal Government customers only.
Create more space in lab freezers and inubators with Nunc MicroWell MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
Corning™ Falcon™ Easy-Grip Tissue Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Features flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells. Corning™ Falcon™ Easy-Grip Tissue Culture Dishes are uniquely designed to simplify lifting one dish or a small stack of dishes.
Corning™ Untreated 245mm Square BioAssay Dishes Available on GSA/VA Contract for Federal Government customers only.
Designed with a generous growth area of 500cm2 to allow for larger assays and better overall use of space in incubators. Corning™ Untreated 245mm Square Low Profile BioAssay Dishes provide a superior option for applications where cell attachment is not desired.
Corning™ Primaria™ Tissue Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Modified polystyrene surface for enhanced cell culture. Corning™ Primaria™ Tissue Culture Dishes feature flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells.
Thermo Scientific™ Nunc™ Rectangular Dishes Available on GSA/VA Contract for Federal Government customers only.
Perform molecular biology applications with non-treated Thermo Scientific™ Nunc™ Rectangular Dishes, which have ANSI™ standard dimensions and are optimized for robotics.
Corning™ BioCoat™ Poly-D-Lysine Culture Dish Available on GSA/VA Contract for Federal Government customers only.
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Corning™ Untreated Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Binds bio-molecules through passive interactions and hydrophobic in nature. Corning™ Untreated Culture Dishes provide an ideal environment for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.
Thermo Scientific™ Nunc™ Glass Bottom Dishes
Using Nunc Glass Bottom Dishes in your research applications gives you the flexibility to convert your culture device into an imaging device.
Thermo Scientific™ Nunc™ Dishes with UpCell™ Surface Available on GSA/VA Contract for Federal Government customers only.
Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with Thermo Scientific™ Nunc™ Dishes with UpCell™ Surface.
Thermo Scientific™ Nunc™ Non-Treated Multidishes Available on GSA/VA Contract for Federal Government customers only.
Perform suspension cell cultures with Thermo Scientific™ Nunc™ Non-Treated Multidishes.
Thermo Scientific™ Nunc™ Poly-D-Lysine or Collagen I Coated Multidishes Available on GSA/VA Contract for Federal Government customers only.
Enhance cell attachment to the culture surface with the extracellular matrix coating on Thermo Scientific™ Nunc™ Poly-D-Lysine or Collagen I Coated Multidishes.
MP Biomedicals™ PlantCon™ Plant Tissue Culture Container
Sterilized Base for PLANTCON™ Plant Tissue Culture Container. Use PlantCon™ containers with liquid or agar solidified media. They can be used for many plant cell culture procedures that currently utilize glass containers.
Corning™ CellBIND™ Surface Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically-clear virgin polystyrene for excellent visibility. Corning™ CellBIND™ Suface Culture Dishes are treated for optimal cell attachment to help cells adapt to reduced serum or serum-free media.
Thermo Scientific™ Nunc™ MiniTrays with Nunclon™ Delta surface Available on GSA/VA Contract for Federal Government customers only.
Acheieve excellent cell attachment and growth rates with Nunc MiniTrays with Nunclon Delta Surface.
Corning™ PureCoat™ Carboxyl Surface Dishes
Negatively charged surface improves attachment. increases proliferation and improved differentiation for a broad range of cells that exhibit poor attachment properties in serum-free or serum-reduced conditions
Corning™ BioCoat™ Poly-D-Lysine Culture Dishwith Coverslip Bottom
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Corning™ BioCoat™ Fibronectin TC-treated Culture Dish Available on GSA/VA Contract for Federal Government customers only.
Matrix components are applied as an even, optically clear coating
Corning™ BioCoat™ Collagen IV TC-Treated Culture Dishes
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
Corning™ BioCoat™ Poly-L-Lysine Culture Dishes
Provide excellent substrate for cell attachment and differentiation of primary neurons, glial, neuroblastoma and glioma cells
  spinner