Cell Culture Flasks
Corning™ U-Shaped Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Designed to improve usability while maintaining the same environment for cell growth as previous designs. Corning™ U-Shaped Cell Culture Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Corning™ Disposable Spinner Flask with Top Cap and Angled Sidearms Available on GSA/VA Contract for Federal Government customers only.
Ready-to-use system comes with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly
Corning™ Disposable Polycarbonate Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear polycarbonate for optimum visibility and durability. Corning™ Disposable Polycarbonate Erlenmeyer Flasks are ideal for shaker cultures applications.
Corning™ Cell Culture Treated Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear virgin polstyrene for optimum visibility and distortion-free microscopic viewing. Corning™ Cell Culture Treated Flasks are available in a variety of sizes, designs and cap styles to meet laboratory diagnostic needs.
Corning™ 1L and 3L Disposable Plastic Spinner Flasks Available on GSA/VA Contract for Federal Government customers only.
Ready-to-use system comes with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly
PYREX™ Fernbach Flasks Available on GSA/VA Contract for Federal Government customers only.
Fernsbach culture flasks offer wide surface area is ideal for growth of organisms requiring a large surface area-to-volume ratio
PYREX™ Pear-Shaped Flasks
With a single standard taper outer joint
Corning™ HYPERFlask™ M Cell Culture Vessels Available on GSA/VA Contract for Federal Government customers only.
Designed specifically for manual use, high-yield and high-performance. Specifically designed for manual use. Corning™ HYPERFlask™ M Cell Culture Vessel uses a multi-layer gas permeable growing surface for efficient gas exchange.
Corning™ Two- and Three-Liter Disposable Polycarbonate Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear polycarbonate for optimum visibility and durability. Corning™ Two-and Three-Liter Disposable Polycarbonate Erlenmeyer Flasks are ideal for shaker cultures applications.
PYREX™ Delong Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Heavy, fire-polished, long necks reduce splashing
DWK Life Sciences Wheaton™ CELLine™ Bioreactor Flask
Membrane cell culture flasks ideal for antibody and protein production
Corning™ Closed System Solutions Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Premium quality conical centrifuge tubes manufactured from high-quality, ultra-clear resins
DWK Life Sciences Kimble™ KIMAX™ Baffled Culture Flasks
For fermentation, cell culture growth and cultivation, and genetic engineering procedures
Fisherbrand™ Shaker Flasks
Reduce the risk of cross-contamination.
PYREX™ Baffled Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Usable with shakers—three baffles provide optimal and consistent agitation
Corning™ Polycarbonate 5L Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
Made of noncytotoxic USP Class VI polycarbonate that provides optical clarity and mechanical strength. Corning™ Polycarbonate 5L Erlenmayer Flask are ready-to-use for efficient scale-up of suspension cultures with space efficiency.
Corning™ Baffled Polycarbonate Flasks with Caps Available on GSA/VA Contract for Federal Government customers only.
Designed with baffles to provide premium agitation at lower speed for gentler, more efficient aeration. Corning™ Baffled Polycarbonate Flasks with Caps are ideal for shaker culture applications.
PYREX™ Culture Media Flasks Available on GSA/VA Contract for Federal Government customers only.
Suitable for mixing and storing culture media
Corning™ Falcon™ Cell Culture Multi Flasks Available on GSA/VA Contract for Federal Government customers only.
Grow cells faster and easier, increasing productivity in cell culture workflow. Corning™ Falcon™ Cell Culture Mini Flasks provide even distribution of media across all layers for homogeneous cell growth.
Corning™ 500mL Disposable Spinner Flask Available on GSA/VA Contract for Federal Government customers only.
PTFE Paddle Bushing was life tested for 2.5 months/24 hr./day at 125rpm
Falcon™ Tissue Culture Treated Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear virgin polystyrene for easy viewing of sample or cells. Corning™ Falcon™ Tissue Culture Treated Flasks features vacuum-gas plasma tissue culture treatment to ensure consistent growth surface.
Thermo Scientific™ Nunc™ TripleFlask™ Treated Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Perform large cell mass applications such as drug screening and scale up with these cell-culture treated TripleFlasks.
Fisherbrand™ Surface Treated Sterile Tissue Culture Flasks, Vented Cap
Perfect for cell growth, cell yields aim on little or medium input volume.
Corning™ Disposable Spinner Flask Aseptic Transfer Caps Available on GSA/VA Contract for Federal Government customers only.
For use with Corning disposable spinner flasks with top cap and angled sidearms
DWK Life Sciences Wheaton™ Magna-Flex™ Spinner Flasks Available on GSA/VA Contract for Federal Government customers only.
Produce high-yield cultures by maintaining even circulation while keeping microcarriers in suspension
Thermo Scientific™ Nunc™ EasYFlask™ Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Enhance cell attachment, growth and differentiation with Thermo Scientific™ Nunc™ Cell Culture Treated EasYFlasks™.
  spinner