Glass Erlenmeyer Style Cell Culture Flasks
PYREX™ Fernbach Flasks Available on GSA/VA Contract for Federal Government customers only.
Fernsbach culture flasks offer wide surface area is ideal for growth of organisms requiring a large surface area-to-volume ratio
PYREX™ Delong Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Heavy, fire-polished, long necks reduce splashing
DWK Life Sciences Kimble™ KIMAX™ Baffled Culture Flasks
For fermentation, cell culture growth and cultivation, and genetic engineering procedures
PYREX™ Baffled Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Usable with shakers—three baffles provide optimal and consistent agitation
PYREX™ Culture Media Flasks Available on GSA/VA Contract for Federal Government customers only.
Suitable for mixing and storing culture media
PYREX™ 50mL Long Neck Shaker Erlenmeyer Flask
Longer necks reduce the amount of splashed solution reaching the closure. PYREX™ 50mL Long Neck Shaker Erlenmeyer Flask are ideal for shaker cultures
PYREX™ Trypsinizing Flasks Available on GSA/VA Contract for Federal Government customers only.
Extra-deep baffles provide vigorous agitation
PYREX™ Low-Form Culture Flask Available on GSA/VA Contract for Federal Government customers only.
For culturing organisms requiring a large surface area-to-volume ratio
PYREX™ Straight-Neck Flasks Available on GSA/VA Contract for Federal Government customers only.
Ideal for shaker cultures
DWK Life Sciences Kimble™ Kontes™ Incubation Flasks
For metabolic studies of biological materials
DWK Life Sciences Wheaton™ Trypsinizing Flasks Available on GSA/VA Contract for Federal Government customers only.
Deep fluted sides enhance agitation
DWK Life Sciences Wheaton™ Nephelo Culture Flask with Cleanout Port and Caps Available on GSA/VA Contract for Federal Government customers only.
Features depressed side arm and side clean-out port
  spinner