Plastic Erlenmeyer Style Cell Culture Flasks
Corning™ Disposable Polycarbonate Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear polycarbonate for optimum visibility and durability. Corning™ Disposable Polycarbonate Erlenmeyer Flasks are ideal for shaker cultures applications.
PYREX™ Pear-Shaped Flasks
With a single standard taper outer joint
Corning™ Two- and Three-Liter Disposable Polycarbonate Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear polycarbonate for optimum visibility and durability. Corning™ Two-and Three-Liter Disposable Polycarbonate Erlenmeyer Flasks are ideal for shaker cultures applications.
Fisherbrand™ Shaker Flasks
Reduce the risk of cross-contamination.
Corning™ Polycarbonate 5L Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
Made of noncytotoxic USP Class VI polycarbonate that provides optical clarity and mechanical strength. Corning™ Polycarbonate 5L Erlenmayer Flask are ready-to-use for efficient scale-up of suspension cultures with space efficiency.
Corning™ Baffled Polycarbonate Flasks with Caps Available on GSA/VA Contract for Federal Government customers only.
Designed with baffles to provide premium agitation at lower speed for gentler, more efficient aeration. Corning™ Baffled Polycarbonate Flasks with Caps are ideal for shaker culture applications.
Thermo Scientific™ Nalgene™ Polycarbonate Baffled Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Increase mixing efficiency with these polycarbonate flasks, an alternative to glass flasks that produce equivalent growth curves with E. coli and S. cerevisiae.
Corning™ Polycarbonate Erlenmeyer Flask with Dip Tube Available on GSA/VA Contract for Federal Government customers only.
Erlenmeyer flasks now available as a closed system solution
Thermo Scientific™ Nalgene™ Polycarbonate Fernbach Culture Flask Available on GSA/VA Contract for Federal Government customers only.
Increase aeration for better growth of culture in liquid media with the wide surface area of these Nalgene Fernbach culture flasks.
Falcon™ Erlenmeyer Flasks
High performance polycarbonate Erlenmeyer flask for suspension culture and storage
IBI Scientific™ TUNAIR™ Shake Flask System
Provides optimum growth conditions for aerobic microorganisms, mammalian cells and plant cells
Greiner Bio-One Erlenmeyer Shaker Flasks
Ideal for suspension culture of mammalian cells, plants, microbes and microorganisms. Each flask is manufactured utilizing an optically clear, inert polycarbonate resin which can be disposed of after single use or be made reusable when autoclaved.
Greiner Bio-One Erlenmeyer Shaker Flasks
Ideal for suspension culture of mammalian cells, plants, microbes and microorganisms. Each flask is manufactured utilizing an optically clear, inert polycarbonate resin which can be disposed of after single use or be made reusable when autoclaved.
Corning™ Vent Cap for 5L Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
Cap, for use with 5L Erlenmeyer flask
DWK Life Sciences Wheaton™ Erlenmeyer Shake Flasks, Baffled Base
Manufactured with virgin, non-leaching Lexan resin that retains clarity after gamma irradiation. Wheaton Shake Flasks, Baffled are certified pyrogen-, DNase- and RNase-free. They have outstanding mechanical, optical, electrical and thermal properties and
  spinner