Cell Culture Microplates
Corning™ Costar™ Flat Bottom Cell Culture Plates Available on GSA/VA Contract for Federal Government customers only.
Tissue culture-treated plates for superior cell attachment. Corning™ Costar™ Flat Bottom Cell Culture Microplates provide an excellent device for diagnostic testing when cell adherence is essential.
Corning™ Costar™ Ultra-Low Attachment Microplates Available on GSA/VA Contract for Federal Government customers only.
Featuring a covalently bound hydrogel layer that effectively inhibits cellular attachmment. Corning™ Costar™ Ultra-Low Attachment Microplates greatly reduce binding of attachment and serum proteins, minimizing cell attachment and spreading.
Corning™ 384-Well Solid Black or White Polystyrene Microplates Available on GSA/VA Contract for Federal Government customers only.
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Falcon™ Polystyrene Microplates Available on GSA/VA Contract for Federal Government customers only.
Constructed of crystal grade virgin polystyrene for optimum visibility. Corning™ Falcon™ Polystyrene Microplates are available with tissue culture-treated and nontreated surfaces.
Corning™ HTS Transwell™ 24-Well Permeable Support System Available on GSA/VA Contract for Federal Government customers only.
Run 24 automated, high-throughput drug transport studies at once. HTS Costar™ HTS Transwell™ 24-Well Permeable Support System is a microplate support system for an array of 24 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 24 inserts to be handled as a single unit.
Corning™ Costar™ HTS Transwell™ -96 Permeable Support with Polyester Membrane Available on GSA/VA Contract for Federal Government customers only.
An array of 96 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit
Greiner Bio-One CELLSTAR™ Cell-Repellent Surface Cell Culture Multiwell Plates
Ideal for suspension, spheroid cultures and stem cell aggregate formation. Greiner Bio-One CELLSTAR™ Cell-Repellent Surface Cell Culture Multiwell Plates effectively inhibit cell adhesion.
Fisherbrand™ Surface Treated SterileTissue Culture Plates
Ideal for comparison and analysis of cell growth, yield in multiple platforms.
Greiner Bio-One CELLSTAR™ 384 Well Polystyrene Cell Culture Microplates
Improved cell adhesion through physical surface treatment. Greiner Bio-One CELLSTAR™ 384 Well Polystyrene Cell Culture Microplates are suitable for optimal microscopic examinations.
Thermo Scientific™ Nunc™ Cell-Culture Treated Multidishes Available on GSA/VA Contract for Federal Government customers only.
Reduce the risk of contamination with these multidishes, which feature a raised well rim and a special surface treatment.
Thermo Scientific™ BioLite Multidishes and Microwell Plate Available on GSA/VA Contract for Federal Government customers only.
Perform general cell culture procedures more economically with Thermo Scientific™ BioLite Cell Culture Treated Plates, guaranteed to be sterile, non-pyrogenic and non-cytotoxic.
Greiner Bio-One 384-Well Storage Microplates
Bind negligible amounts of proteins or active substances.
Greiner Bio-One CELLSTAR™ Cell Culture Multiwell Microplates
For high-throughput screening in cell based test systems.
Falcon™ Solid Opaque 96-Well Tissue Culture Plates Available on GSA/VA Contract for Federal Government customers only.
For cell-based bioluminescent, chemiluminescent, or fluorescent assays
Corning™ CellBIND™ Surface Microplates Available on GSA/VA Contract for Federal Government customers only.
Treated with CellBIND™ to help cells adapt to reduced serum or serum-free media and improve cell attachment for increased cell growth and yield. Corning™ CellBIND™ Surface Microplates are constructed of clear polystyrene for optimum visibility and are available in multiple well formats.
Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates Available on GSA/VA Contract for Federal Government customers only.
Perform automated applications more precisely with these round-bottom plates, which are available with and without cell culture treatment.
Falcon™ HTS Multiwell Insert System Available on GSA/VA Contract for Federal Government customers only.
Save time and labor in HTS protocols—grow, handle, and analyze 24 samples simultaneously. Corning™ Falcon™ HTS Multiwell Insert System features a generous, automation-compatible sampling port which allows sampling above and below membranes.
Greiner Bio-One SensoPlate™ 96-Well Glass Bottom Microplates
Designed for FCS (fluorescence correlation spectroscopy) and demanding microscopic applications. Greiner Bio-One SensoPlate™ 96-Well Glass Bottom Microplates facilitates use of lenses with low working distance and high aperture.
EMD Millipore™ Millicell™ 24-Well Cell Culture Microplate Assemblies Available on GSA/VA Contract for Federal Government customers only.
Provides twice the membrane area of standard 24-well plates
Corning™ HTS Transwell™ -96 Well Permeable Support System Available on GSA/VA Contract for Federal Government customers only.
Boasts membrane area for cell growth or migration of 0.143cm2. Corning™ HTS Transwell™ 96-Well Permeable Support System is a microplate support system for an array of 96 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.
Corning™ 96 Well Ultra-Low Attachment Treated Spheroid Microplate Available on GSA/VA Contract for Federal Government customers only.
Ultra low attachment surface with covalently bonded hydrogel minimizes cell attachment, protein absorption, enzyme activation and cellular activation
Corning™ 384 Well Ultra-Low Attachment Treated Spheroid Microplate Available on GSA/VA Contract for Federal Government customers only.
Ultra low attachment surface with covalently bonded hydrogel minimizes cell attachment, protein absorption, enzyme activation and cellular activation
Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Storage Microplates Available on GSA/VA Contract for Federal Government customers only.
Lower spending when storing compounds for small volume applications by using Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Microplates.
Thermo Scientific™ MIC 2000 Microtiter microplate Available on GSA/VA Contract for Federal Government customers only.
For support of microbiolgical applications
EMD Millipore™ MultiScreen™ Cell Culture Trays Available on GSA/VA Contract for Federal Government customers only.
For feeding cells during membrane-based cell culture
Falcon™ FluoroBlok™96 Well HTS Insert Systems Available on GSA/VA Contract for Federal Government customers only.
Simplify detection of migrating fluorescently labeled cells. Corning™ Falcon™ FluoroBlok™96 Well HTS Insert Systems are designed with a unique light-tight PET membrane that blocks the transmission light within the range of 400 to 700nm.
Corning™ 384-Well Optical Imaging Microplates Available on GSA/VA Contract for Federal Government customers only.
Ideal for whole cell or subcellular high content screening
  spinner