Dishes
Thermo Scientific™ Nalgene™ Large Polypropylene Sterilizing Pans Available on GSA/VA Contract for Federal Government customers only.
Dry and sterilize instruments and labware with Thermo Scientific™ Nalgene™ Large Polypropylene Sterilizing Pans.
Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Pans Available on GSA/VA Contract for Federal Government customers only.
Repeatedly autoclave instruments and labware with Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Pans, featuring heavy-duty construction.
Thermo Scientific™ Coplin Staining Jar
Hold 10 slides back-to-back with the Thermo Scientific™ Plastic Coplin Staining Jar.
Thermo Scientific™ Nalgene™ HDPE Pans Available on GSA/VA Contract for Federal Government customers only.
Soak labware or form ice baths using Thermo Scientific™ Nalgene™ HDPE Pans, which are tapered for nesting and chemically resistant to most acids, bases and alcohols.
Thermo Scientific™ RA Lamb Glass Coplin Jar, holds 5 slides (10 back to back)
Hold up to 10 slides back-to-back with Thermo Scientific™ RA Lamb molded glass coplin jars. Jars have a drop-on lid and integral large diameter base.
Fisherbrand™ Polystyrene Antistatic Weighing Dishes
Disposable dishes are ideal for weighing, dispensing, or storing applications
PYREX™ Crystallizing Dishes Available on GSA/VA Contract for Federal Government customers only.
Reusable borosilicate glass dishes with heavy duty rims ideal for storage and crystallization
PYREX™ Reusable Petri Dishes: Complete Available on GSA/VA Contract for Federal Government customers only.
Flat, clear dishes with beaded edges withstand repeated sterilization (wet or dry)
Fisherbrand™ Petri Dishes with Clear Lid Available on GSA/VA Contract for Federal Government customers only.
Feature a new and improved design and packaging
PYREX™ Glass Drying Trays Available on GSA/VA Contract for Federal Government customers only.
Designed for use as drying, staining, or developing trays
Fisherbrand™ Aluminum Weighing Dishes
Aluminum dishes for weighing and dispensing
Fisherbrand™ Glass Staining Dish for 10 Slides, with Cap Available on GSA/VA Contract for Federal Government customers only.
Hold up to 10 standard 3 × 1 in. (75 × 25mm) slides back-to-back
Bel-Art™ SP Scienceware™ Conway Diffusion Cells Available on GSA/VA Contract for Federal Government customers only.
For Obrink method of determining carbon dioxide, ammonia or alcohol in blood or serum by diffusion method. Bel Art™ SP Scienceware™ Conway Diffusion Cells are steam autoclavable at 121°C (250°F).
Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray with Cover Available on GSA/VA Contract for Federal Government customers only.
Reduce breakage and chipping of glass pipette tips with the stackable and lightweight Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray.
Fisherbrand™ Glass Staining Dishes for 16 Slides Available on GSA/VA Contract for Federal Government customers only.
Staining jar holds 3 × 1 in. (75 × 25mm) slides vertically
Mettler Toledo™ Moisture Analyzer Sample Pans Available on GSA/VA Contract for Federal Government customers only.
For halogen and infrared moisture analyzers
DWK Life Sciences Wheaton™ Accessories and Replacement Parts for 20-Slide Glass Staining Dish Available on GSA/VA Contract for Federal Government customers only.
Featuring an open bottom to facilitate rapid immersion and drainage, reducing carryover. Wheaton™ Accessories and Replacement Parts for 20-Slide Glass Staining Dish are manufactured from soda-lime glass.
Bel-Art™ SP Scienceware™ Polypropylene Sterilizing Trays and Covers Available on GSA/VA Contract for Federal Government customers only.
Corrosion-proof polypropylene plastic trays and covers (sold separately. Bel Art™ SP Scienceware™ Polypropylene Sterilizing Trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.
Fisherbrand™ Heavy Duty Polypropylene Lab Trays
Fabricated with 3/16 in. thick polypropylene.
Fisherbrand™ Hexagonal Polystyrene Weighing Dishes
Hexagonal Polystyrene Weighing Dishes are used for handling solids or liquids during weighing. Design provides greater balance, protection and safety.
Thermo Scientific™ Plastic Spill-Free Slide Jar
Hold 10 slides, one per slot with the Thermo Scientific™ Plastic Spill-Free Slide Jar.
Bel-Art™ SP Scienceware™ Polypropylene Coplin Staining Jar Available on GSA/VA Contract for Federal Government customers only.
Improves on the classic tapered square design with a larger opening for easier placement and removal of slides. Bel Art™ SP Scienceware™ Polypropylene Coplin Staining Jar makes working with slides easier.
Fisherbrand™ Low- and Tall-Form PTFE Evaporating Dishes
Inert and usable to 280°C (536°F).
  spinner