Special Purpose Dishes
PYREX™ Crystallizing Dishes Available on GSA/VA Contract for Federal Government customers only.
Reusable borosilicate glass dishes with heavy duty rims ideal for storage and crystallization
Bel-Art™ SP Scienceware™ Conway Diffusion Cells Available on GSA/VA Contract for Federal Government customers only.
For Obrink method of determining carbon dioxide, ammonia or alcohol in blood or serum by diffusion method. Bel Art™ SP Scienceware™ Conway Diffusion Cells are steam autoclavable at 121°C (250°F).
DWK Life Sciences Kimble™ KIMAX™ Crystallizing Dishes
Ideal for storage and crystallization
Dual Manufacturing Company Aluminium Dishes Non-distribution product as customer accommodation.
Heavy guage aluminum construction
CoorsTek™ Porcelain Combustion Boats
AD-998 High-Alumina labware
DWK Life Sciences DURAN™ Crystallizing Dishes with Spout
An excellent option for crystallization, heating and cooling baths and microscale experiments in the laboratory.
Eisco™ Flat Bottom Glass Crystallizing Dishes with Spout Products for Science Education
Flat bottom glass crystallizing dishes with spouts for easy, mess free pouring. Ideal for storage and crystallization. Reusable and autoclavable.
Lab-Aids™ Chemplates™ Reaction Plate Products for Science Education
An inexpensive way to conduct safe and effective laboratory experiments.
United Scientific Supplies Porcelain Casseroles Products for Science Education
Porcelain casserole dishes with porcelain handles.
  spinner