Boiling Flasks
PYREX™ Flasks with standard taper Joints, Short Necks, Round Bottom Available on GSA/VA Contract for Federal Government customers only.
Excellent chemical durability for contamination-free results
PYREX™ Flat Bottom Flasks with Long Necks Available on GSA/VA Contract for Federal Government customers only.
Usual globe-shaped Florence boiling flasks
Kimble™ Kontes™ Antibiotic Moisture Content Flasks
For determining moisture loss on drying in tablets, troches, or capsules
Kimble™ Kontes™ Pear-Shaped Heavy Wall Boiling Flask
Single-neck flasks with a Standard Taper outer joint at the top manufactured from 33 expansion, low extractable borosilicate glass.
PYREX™ VISTA™ Flat Bottom Boiling Flask Available on GSA/VA Contract for Federal Government customers only.
Designed for shock resistance
PYREX™ Heavy-Wall Boiling Flasks, Round Bottom Available on GSA/VA Contract for Federal Government customers only.
Feature 25 to 30% greater wall thickness than standard flasks for mechanical strength
PYREX™ Pear-Shaped Flasks Available on GSA/VA Contract for Federal Government customers only.
With a single standard taper outer joint
Kemtech America™ Synthware™ Two Neck Pear Shaped Flasks
Used for filtration followed by recrystallization on opposite side of disc of air sensitive compounds
Kimble™ Kontes™ Flat Bottom Heavy-Wall Boiling Flasks
Single neck with a standard tapered outer joint
Kimble™ Ray-Sorb™ Brand Flat Bottom Flasks
Provide protection to solutions sensitive to light of shorter wavelengths
Kemtech America™ Synthware™ Three-Neck Flasks, Angled
With standard taper outer joints on both center and angled side necks
Kemtech America™ Synthware™ Single Neck Round Bottom Flasks, Full Length Joint
Hand-blown from tubing to ensure uniform wall thickness
Kemtech America™ Synthware™ Single Neck Round Bottom Flasks, Medium Length Joint
Hand-blown from tubing to ensure uniform wall thickness
PYREX™ VISTA™ Round Bottom Boiling Flask Available on GSA/VA Contract for Federal Government customers only.
Designed for great shock resistance
Kemtech America™ Synthware™ Flat Bottom Flask, Single Neck
All sizes are hand blown from tubing to ensure a uniform wall thickness
PYREX™ Flasks with Short Ring Necks Available on GSA/VA Contract for Federal Government customers only.
Sometimes called “balloon flasks”
Kimble™ Kontes™ Round Bottom Heavy Wall Boiling Flasks
Single neck flask with a standard outer joint
Kemtech America™ Synthware™ Round Bottom Flasks, 4-Neck
With standard taper outer joints on both the center and vertical side necks
Kimble™ Kontes™ Round Bottom with SJ Joint, Heavy Wall Boiling Flasks
Useful as rotary evaporator condensate collection flask
Kimble™ Kontes™ Boiling Flask for Microflex Kit
For use with standard taper14/10 joint microscale apparatus
  spinner