Flat Bottom Boiling Flasks
PYREX™ Flat Bottom Flasks with Long Necks Available on GSA/VA Contract for Federal Government customers only.
Usual globe-shaped Florence boiling flasks
DWK Life Sciences Kimble™ Kontes™ Antibiotic Moisture Content Flasks
For determining moisture loss on drying in tablets, troches, or capsules
PYREX™ VISTA™ Flat Bottom Boiling Flask Available on GSA/VA Contract for Federal Government customers only.
Designed for shock resistance
Synthware™ Flat Bottom Flask, Single Neck
All sizes are hand blown from tubing to ensure a uniform wall thickness
United Scientific Supplies Boiling Flasks, Flat Bottom, Ground Glass Joint, Borosilicate Glass
Boiling, flat round bottom flasks with ground glass joints (24/40).
DWK Life Sciences DURAN™ Narrow Neck Flat Bottom Flasks
With beaded rim; Uniform wall thickness
Eisco 500mL Florence Boiling Flask; Flat bottom; Premium Borosilicate Glass; Narrow neck; Beaded Rim
500ml Flat bottom Florence boiling flask made of heavy duty borosilicate glass. Flask has a flat bottom. Long neck, Stopper not included. The flask is ideal for dealing with liquids at different temperatures due to its thick-walled design.
DWK Life Sciences Kimble™ Ray-Sorb™ Brand Flat Bottom Flasks
Provide protection to solutions sensitive to light of shorter wavelengths
DWK Life Sciences Kimble™ ValueWare™ Flat-Bottom Florence Boiling Flasks Products for Science Education
Low coefficient of expansion to resist thermal shock.
Dynalon™ Polypropylene Flasks Products for Science Education
Your best choice for basic chemistry labs.
PYREX™ VISTA™ Flat-Bottom Boiling Flasks Products for Science Education
Designed for great shock resistance.
  spinner