Flat Sided Cell Culture Flasks and Accessories
Corning™ U-Shaped Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Designed to improve usability while maintaining the same environment for cell growth as previous designs. Corning™ U-Shaped Cell Culture Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Corning™ Cell Culture Treated Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear virgin polstyrene for optimum visibility and distortion-free microscopic viewing. Corning™ Cell Culture Treated Flasks are available in a variety of sizes, designs and cap styles to meet laboratory diagnostic needs.
Corning™ HYPERFlask™ M Cell Culture Vessels Available on GSA/VA Contract for Federal Government customers only.
Designed specifically for manual use, high-yield and high-performance. Specifically designed for manual use. Corning™ HYPERFlask™ M Cell Culture Vessel uses a multi-layer gas permeable growing surface for efficient gas exchange.
DWK Life Sciences Wheaton™ CELLine™ Bioreactor Flask
Membrane cell culture flasks ideal for antibody and protein production
Corning™ Falcon™ Cell Culture Multi Flasks Available on GSA/VA Contract for Federal Government customers only.
Grow cells faster and easier, increasing productivity in cell culture workflow. Corning™ Falcon™ Cell Culture Mini Flasks provide even distribution of media across all layers for homogeneous cell growth.
Falcon™ Tissue Culture Treated Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear virgin polystyrene for easy viewing of sample or cells. Corning™ Falcon™ Tissue Culture Treated Flasks features vacuum-gas plasma tissue culture treatment to ensure consistent growth surface.
Thermo Scientific™ Nunc™ TripleFlask™ Treated Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Perform large cell mass applications such as drug screening and scale up with these cell-culture treated TripleFlasks.
Fisherbrand™ Surface Treated Sterile Tissue Culture Flasks, Vented Cap
Perfect for cell growth, cell yields aim on little or medium input volume.
Thermo Scientific™ Nunc™ EasYFlask™ Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Enhance cell attachment, growth and differentiation with Thermo Scientific™ Nunc™ Cell Culture Treated EasYFlasks™.
Thermo Scientific™ Nalgene™ Vented HDPE Closures for Sterile Single Use Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.
Ensure a sterile gas exchange for shaker and suspension cell culture by using Thermo Scientific™ Nalgene™ Vented HDPE Closures for Sterile Single Use Erlenmeyer Flasks.
Corning™ CELLine Disposable Bioreactor Available on GSA/VA Contract for Federal Government customers only.
Cultivation flask for secreted product, recombinant protein expression and high-yield monoclonal antibody (MAb) production
Thermo Scientific™ Nunc™ Nunclon™ Δ Cell Factory Components and Replacement Parts Available on GSA/VA Contract for Federal Government customers only.
Accessories for filling and emptying the cell factories with media
Corning™ U-Shaped Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Designed to improve usability while maintaining the same environment for cell growth as previous designs. Corning™ U-Shaped Cell Culture Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Thermo Scientific™ BioLite Cell Culture Treated Flasks Available on GSA/VA Contract for Federal Government customers only.
Perform general cell culture procedures more economically with Thermo Scientific™ BioLite Cell Culture Treated Flasks, guaranteed to be sterile, non-pyrogenic and non-cytotoxic.
Corning™ Ultra-Low Attachment Flasks Available on GSA/VA Contract for Federal Government customers only.
Features a covalently bound hydrogel layer that minimizes cell attachment, protein absorption and cellular activation. Corning™ Ultra-Low Attachment Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Greiner Bio-One CELLSTAR™ TC Treated Cell Culture Flasks
Improved cell adhesion through physical surface treatment. Greiner Bio-One CELLSTAR™ TC Treated Cell Culture Flasks are free of detectable DNase, RNase and human DNA.
Corning™ U-Shaped Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Designed to improve usability while maintaining the same environment for cell growth as previous designs. Corning™ U-Shaped Cell Culture Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Corning™ Cell Culture Untreated Flasks Available on GSA/VA Contract for Federal Government customers only.
Printed with lot numbers for ease in traceability
Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap Available on GSA/VA Contract for Federal Government customers only.
Pre-coated with Collagen I for improved cell attachment and an increased proliferation rate of transfected mammalian cells, primary cells and stem cells. Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap provides highly consistent and biologically functional precoated surfaces to more closely mimic in vivo environments for cell culture applications.
Greiner Bio-One CELLSTAR™ TC Treated Lo-Profile Cell Culture Flasks
Improved cell adhesion through physical surface treatment. Greiner Bio-One CELLSTAR™ TC Treated Lo-Profile Cell Culture Flasks is free of detectable DNase, RNase and human DNA.
Corning™ CellBIND™ Surface Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Increases suface wettability for more even and consistent cell attachment. Corning™ CellBIND™ Surface Cell Culture are available in a variety of sizes, designs and cap styles to meet all laboratory needs.
Thermo Scientific™ Nunc™ Non-treated Flasks Available on GSA/VA Contract for Federal Government customers only.
Minimize the chance of contamination with these high-quality, optically clear polystyrene Thermo Scientific™ Nunc™ Non-treated Flasks for suspension cell cultures.
Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Filter Caps Available on GSA/VA Contract for Federal Government customers only.
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Filter Caps.
Greiner Bio-One CELLCOAT™ Collagen Type I Coated Cell Culture Flask
Ideal for cell cultivation in serum-free or serum-reduced medium
Falcon™ Nontreated Tissue Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Similar to the surface of a bacteriological Petri dish
Corning™ Cell Culture Flask with Bar Code Available on GSA/VA Contract for Federal Government customers only.
Features nonrepeating bar code for use in automation
Greiner Bio-One CELLSTAR™ Suspension Culture Flask with Filter Cap
Ideal for suspension cultures, hybridoma and embryonic stem cells. Greiner Bio-One CELLSTAR™ Suspension Culture Flask with Filter Cap is free of detectable DNase, RNase and human DNA.
Corning™ Primaria™ Tissue Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Improves attachment, spreading and growth for many primary cells or cell lines. Corning™ Primaria™ Tissue Culture Flasks are suitable for growing cells that grow poorly on conventional, negatively charged, standard tissue culture surfaces—including tumor cells, endothelial cells and hepatocytes.
Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask Available on GSA/VA Contract for Federal Government customers only.
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology. Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask is ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.
  spinner