384 Well Assay Microplates
Thermo Scientific™ 384-Well Conical Bottom Polypropylene Plates Available on GSA/VA Contract for Federal Government customers only.
Minimize costs when storing compounds for small volume applications. Lower binding capacity means proteins or DNA will not bind, allowing for complete sample recovery.
Thermo Scientific™ 384-Well Full Skirted PCR Plates Available on GSA/VA Contract for Federal Government customers only.
Reduce well-to-well variability with 384-Well PCR Plate Full Skirted PCR Plates, thin walled for efficient heat transfer and compatible with most major thermal cyclers.
Thermo Scientific™ Armadillo™ 384-Well PCR Plates Available on GSA/VA Contract for Federal Government customers only.
Optimize high throughput robotic PCR and qPCR applications with these uItra-rigid 384-well PCR plates with poylcarbonate frames and polypropylene wells.
Applied Biosystems™ MicroAmp™ Optical 384-Well Reaction Plate with Barcode Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification
Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 384-Well Clear GPLE Reaction Plates with Barcode Non-distribution product as customer accommodation.
Validated on Applied Biosystems thermal cyclers, real-time thermal cyclers, and genetic analyzers for optimal fit and performance
Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 384-Well Plates Available on GSA/VA Contract for Federal Government customers only.
Carry out colormetric assays with reliable and reproducible results using these clear flat-bottom immuno nonsterile 384-well plates, offered with a choice of surface.
Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 384-Well Clear Reaction Plates with Barcode Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Ideal solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments
Corning™ 384 Well Flat Clear Bottom Polystyrene TC-Treated Microplates, low flange, with lid, sterile Available on GSA/VA Contract for Federal Government customers only.
Can be used for both top and bottom reading instruments. Corning™ 384 Well Flat Clear Bottom Polystyrene TC-Treated Microplates have black wall plates for low background fluorescence, minimal light scatter, and reduced crosstalk.
Corning™ 384-Well Solid Black or White Polystyrene Microplates Available on GSA/VA Contract for Federal Government customers only.
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Corning™ 384-Well Spheroid Microplate Available on GSA/VA Contract for Federal Government customers only.
Treated with an ultra-low attachment surface to enable uniform and reproducible 3D multicellular spheroid formation. Corning™ 384-Well Spheroid Microplate permit direct microscopic viewing and readings for top and bottom reading instruments.
Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 384-Well Green Reaction Plates with Barcode Non-distribution product as customer accommodation.
Ideal solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments
Eppendorf™ twin.tec™ 384-Well PCR Plate Available on GSA/VA Contract for Federal Government customers only.
Unique, “twin-shot” process combines virgin polypropylene wells and polycarbonate plate deck and skirt in a one-piece plate
Axygen™ 384-well Skirted PCR Microplates Available on GSA/VA Contract for Federal Government customers only.
Single and double notch 384-well PCR plates
Corning™ 384-Well Low Flange Polystyrene Microplates Available on GSA/VA Contract for Federal Government customers only.
For use with fluorescent assays with detection microplate reader. Corning™ 384-Well Low Flange Polystyrene Microplates have low background fluorescence, minimal light scatter and reduced crosstalk.
Greiner Bio-One™ 384-Well Polypropylene Conical Bottom Microplates
Resistant to many standard laboratory chemicals such as DMSO. Greiner Bio-One 384 Well Polypropylene Conical Bottom Non-Sterile Microplate are compatible with standard equipment and automated systems.
Thermo Scientific™ Nunc™ 384-Well Optical Bottom Plates Available on GSA/VA Contract for Federal Government customers only.
View microscopic plate contents using luminescence and fluorescence with these 384-well optical bottom plates.
Corning™ 384 Well Flat Clear Bottom Polystyrene Untreated Microplates, low flange, no lid, nonsterile Available on GSA/VA Contract for Federal Government customers only.
Untreated (medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions. Corning™ 384 Well Flat Clear Bottom Polystyrene Untreated Microplates are ideally suited for fluorescent assays using either top or bottom detection plate readers.
Corning™ 384-Well Clear Polystyrene Microplates Available on GSA/VA Contract for Federal Government customers only.
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ 364-Well Clear Polystyrene Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 364-well microplates for laboratory miniaturization and automation.
Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 384-Well Red Reaction Plates with Barcode Non-distribution product as customer accommodation.
Ideal solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments
Greiner Bio-One™ Small Volume™ 384 Deep Well Polypropylene Microplates
Ensures accurate pipetting, reduces dead volume. Greiner Bio-One Small Volume™ 384 Deep Well Polypropylene Microplates are specifically designed to accommodate heat-sealing systems.
Axygen™ LightCycler 480 Compatible PCR Plates Available on GSA/VA Contract for Federal Government customers only.
384-well plate is designed to fit the Roche LightCycler™ 480
Thermo Scientific™ Nunc™ 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces Available on GSA/VA Contract for Federal Government customers only.
Reduce the volume of reagent needed with Thermo Scientific™ Nunc™ 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces.
Corning™ CellBIND™ 384 Well Flat Clear Bottom Polystyrene Microplates, with lid, sterile, 10/bag Available on GSA/VA Contract for Federal Government customers only.
Feature CellBIND™ surface treated for optimal cell attachment. Corning™ CellBIND™ 384 Well Flat Clear Bottom Polystyrene Microplates can be used for both top and bottom reading instruments. Their black wall plates have low background fluorescence, minimal light scatter, and reduced crosstalk.
Greiner Bio-One™ 384 Well Polypropylene Flat Bottom Non-Sterile Microplates
Resistant to many standard laboratory chemicals such as DMSO. Greiner Bio-One 384 Well Polypropylene Flat Bottom Non-Sterile Microplates are compatible with standard equipment and automated systems.
Corning™ Low-Volume 384-Well Polystyrene Plates Available on GSA/VA Contract for Federal Government customers only.
High-base design is compatible with automation and bar coding
Thermo Scientific™ WebSeal Plate+ 384-Well Glass-Coated Microplates Available on GSA/VA Contract for Federal Government customers only.
Get chemical resistance similar to glass while retaining the handling advantages of polypropylene with these 384-well glass-coated polypropylene microplates.
Corning™ Falcon™ Optilux 384-Well Microtest Microplates Available on GSA/VA Contract for Federal Government customers only.
Manufactured from polystyrene and treated with tissue culture to optimize cell growth. Corning™ Falcon™ Optilux 384-Well Microtest Microplates are ideal for use in high throughput systems.
Greiner Bio-One™ 384-Well Small Volume™ HiBase Polystyrene Microplates
Features round wells with a conical geometry. Greiner Bio-One 384-Well Small Volume™ HiBase Polystyrene Microplates is free of detectable DNase, RNase, human DNA.
Fisherbrand™ 384-Well Skirted PCR Plates
Offer minimized warp and bowing for better robotic handling
  spinner