Filtering Microplates
MilliporeSigma™ MultiScreen™ 96-Well Assay Plates for Sample Preparation Available on GSA/VA Contract for Federal Government customers only.
For the isolation, purification, and preparation of samples prior to testing in drug discovery processes and protein and DNA applications
MilliporeSigma™ MultiScreenHTS Durapore™ 96-Well Filter Plates Available on GSA/VA Contract for Federal Government customers only.
Designed for full automation compatibility. MilliporeSigma™ MultiScreenHTS Durapore™ 96-Well Filter Plates are innovative plates ideal for bead based assays. Opaque Barex™ and clear, sterile or non-sterile styrene and racks are available in a range of pore sizes.
MilliporeSigma™ MultiScreen™ 96-Well Assay Plates for ELISPOT Assays Available on GSA/VA Contract for Federal Government customers only.
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
MilliporeSigma™ MultiScreen™-Mesh Filter Plates with 96-Well Receiver Plates Available on GSA/VA Contract for Federal Government customers only.
Ideal for target screening and other applications that evaluate new compounds using multicellular organisms as the in vivo model
GE Healthcare Whatman™ UniFilter™ 384-Well Microplates Available on GSA/VA Contract for Federal Government customers only.
Microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples.
MilliporeSigma™ MultiScreenHTS 96-Well Filter Plates
For filtration-based solubility determination. MultiScreenHTS 96-Well Filter Plates are specifically developed for use with automation.
GE Healthcare Whatman™ UniFilter™ Microplates, 24-Well, 10mL
Ideal for applications that require larger sample or reagent volumes (e.g. biomolecular purification by solid-phase extraction and organic synthesis
MilliporeSigma™ MultiScreen™ 96-Well Assay Plates for Cell-Based Assays
Offer formats that allow a wide range of functional cell-based and whole-organism assays to be performed
Corning™ FiltrEX Filter Plates Available on GSA/VA Contract for Federal Government customers only.
Individual filter discs eliminate liquid cross-contamination
MilliporeSigma™ MultiScreen™ HTS 96-Well Filter Plates with MCE Membrane Available on GSA/VA Contract for Federal Government customers only.
Commonly used for Elispot assays, bacterial screening, ELISA or viral infection studies
MilliporeSigma™ MultiScreenHTS 384-Well Filter Plates
Use in automated, high throughput applications, including biological assays and sample preparation. MilliporeSigma™ MultiScreenHTS 384-Well Filter Plates are non-sterile, single use plates available in several mesh materials and pore sizes to meet the needs of different applications.
MilliporeSigma™ MultiScreen™ High Volume Filter Plates
Multiwell solutions for discovery research and sample prep
MilliporeSigma™ MultiScreen™ 96-Well Assay Systems
Perform cell growth, feeding, and analysis in one plate
MilliporeSigma™ MultiScreenHTS Solubility Filter Plate
96-well filter plate designed and optimized for aqueous solubility assays
MilliporeSigma™ MultiScreenHTS 96-Well Filter Plates
Receptor/ligand binding assays, sample prep, enzyme assays with precipitation, DNA purification
MilliporeSigma™ MultiScreen™ Solvinert Filter Plates
Optimized for drug discovery applications. MilliporeSigma™ MultiScreen™ Solvinert Filter Plates are solvent compatible and feature a unique one-piece design. 96-well plates are available in deep well (2mL) format (with or without pre-filter) and standard (500μL) format, with hydrophobic or hydrophilic membranes.
MilliporeSigma™ MultiScreenHTS 96-Well Filter Plates
Receptor/ligand binding assays, sample prep,TCA precipitation
MilliporeSigma™ MultiScreenHTS 96-Well Filter Plates
Non-sterile, clear 96-well filter plate with 0.45μm pore size Hydrophobic PVDF membrane for protein interaction assays, Avidin-biotin Linkages, DNA-binding proteins.
MilliporeSigma™ MultiScreen™ Harvest Plates
This Opaque 96-well Cell Harvesting Plates has a Glass Fiber membrane used for Cell harvesting.
MilliporeSigma™ MultiScreen™HTS 96-Well Filter Plates with Negatively Charged Membrane
Binds positive charge—ideal for kinase assays with peptide substrates and enzyme assays
MilliporeSigma™ MultiScreen™ Centrifuge Alignment Frame
Allows easy and accurate transfer of samples
MilliporeSigma™ MultiScreenHTS 96-Well Filter Plates
For filtration-based solubility determination. MultiScreenHTS 96-Well Filter Plates are specifically developed for use with automation
MilliporeSigma™ MultiScreen™ 96-Well Assay Plates
Provide tools to perform high-throughput studies of ADME processes—the investigation of the adsorption, distribution, metabolism, and excretion of potential drugs
MilliporeSigma™ MultiScreen™ Multiple Punch Kits
Includes punch with punch carrier plate and punch distributor
MilliporeSigma™ MultiScreen™ Column Loaders
No need to pipet slurries, gels, or resins; load all 96 wells simultaneously and uniformly
MilliporeSigma™ MultiScreen™HTS 96-Well Filter Plates for Clearing
Designed to meet specific performance criteria for sample preparation methodology
MilliporeSigma™ MultiScreen™ High-Volume Manifold Collar
For use with Millipore MultiScreen high-volume filter plates
  spinner