Abietic Acid
Abietic acid, 85%, ACROS Organics™
Chemical Name or Material: Abietic acid Name Note: 85% CAS: 514-10-3 CAS Min %: 80.0 CAS Max %: 100.0 Purity Grade: Technical Grade Assay: ≥80% Assay Percent Range: 85% Molecular Formula: C20H30O2 Formula Weight: 302.45 MDL Number: MFCD03423567
  spinner