Acylation Reagents
Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI) Acylation Reagents Available on GSA/VA Contract for Federal Government customers only.
Conduct acylation of hydroxyl groups and primary and secondary amines with the Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI).
Thermo Scientific™ Pentafluoropropanol Acylation Reagent Available on GSA/VA Contract for Federal Government customers only.
Improve detection with the Thermo Scientific™ Pentafluoropropanol Acylation Reagent for derivatization.
Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA) Acylation Reagents Available on GSA/VA Contract for Federal Government customers only.
Prepare volatile and stable fluoracyl derivatives for GC/MS with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA).
Thermo Scientific™ MBTFA Acylation Reagent Available on GSA/VA Contract for Federal Government customers only.
Trifluoroacylate primary and secondary amines, hydroxyl and thiol groups, and carbohydrates with the Thermo Scientific™ MBTFA Acylation Reagent.
  spinner