Adipic Acid
Adipic acid, 99%, ACROS Organics™
Chemical Name or Material: Adipic acid CAS: 124-04-9 CAS Min %: 98.5 CAS Max %: 100.0 Assay: ≥99% Assay Percent Range: 99% Molecular Formula: C6H10O4 Linear Formula: HO2C(CH2)4CO2H Formula Weight: 146.14 MDL Number: MFCD00004420 Synonym: Hexanedioic acid
Adipic Acid (Powder/Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Adipic Acid CAS: 124-04-9 Purity Grade: Certified Grade Physical Form: Solid Molecular Formula: C6H10O4 Linear Formula: HOOC(CH2)4COOH Formula Weight: 146.15 MDL Number: MFCD00004420 Synonym: 1, 4-Butanedicarboxylic Acid
Adipic Acid Products for Science Education
Chemical Name or Material: Adipic Acid CAS: 124-04-9 Linear Formula: HO2C(CH2)4CO2H Formula Weight: 146.14 Synonym: 1, 4-Butanedicarboxylic Acid
Adipic Acid, Free Acid, MP Biomedicals™
Chemical Name or Material: Adipic acid Name Note: Free Acid CAS: 124-04-9 Physical Form: Crystalline Molecular Formula: C6H10O4 Formula Weight: 146.1 Synonym: 1, 4-Butanedicarboxylic acid
Adipic Acid Products for Science Education
Chemical Name or Material: Adipic Acid CAS: 124-04-9 Linear Formula: C6H10O4 Formula Weight: 146.15
Adipic Acid, OR, Macron Fine Chemicals™
Chemical Name or Material: Adipic Acid CAS: 124-04-9 Physical Form: Powder or Crystals Molecular Formula: HOCO(CH2)4COOH Formula Weight: 146.14
  spinner