Alanine -Ala and Derivatives
DL-Alanine 99%, ACROS Organics™
Chemical Name or Material: DL-Alanine Name Note: 99% CAS: 302-72-7 Assay: 98.5 to 101.5% (on dry substance) Assay Percent Range: 99% Molecular Formula: C3H7NO2 Linear Formula: CH3CH(NH2)CO2H Formula Weight: 89.09 MDL Number: MFCD00064408
2-Bromo-4-fluoroaniline, 99%, ACROS Organics™
Chemical Name or Material: 2-Bromo-4-fluoroaniline CAS: 1003-98-1 CAS Min %: 98.5 CAS Max %: 100.0 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C6H5BrFN Linear Formula: BrC6H3(F)nH Formula Weight: 190.02 MDL Number: MFCD00042462
BOC-D-Alanine, 99+%, ACROS Organics™
Chemical Name or Material: BOC-D-Alanine Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C8H15NO4 Linear Formula: (CH3)3COC(O)nHC(CH3)COOH Formula Weight: 189.21 MDL Number: MFCD00063123 Synonym: N-(tert-Butoxycarbonyl)-D-alanine
D-Alanine, +99%, ACROS Organics™
Chemical Name or Material: D-Alanine Assay: 99.0 to 101.0% (on dry substance), 1% max. (L-content) Assay Percent Range: 99+% Molecular Formula: C3H7NO2 Linear Formula: CH3CH(NH2)CO2H Formula Weight: 89.09 Synonym: D-2-Aminopropionic acid
beta-Alanine, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: β-Alanine CAS: 107-95-9 CAS Min %: 98.5 CAS Max %: 100.0 Assay: ≥98.5% (on dry substance) Assay Percent Range: 99% Molecular Formula: C3H7NO2 Linear Formula: H2NCH2CH2CO2H Formula Weight: 89.09 MDL Number: MFCD00008200 Synonym: 3-Aminopropanoic acid
β-Alanine ethyleester hydrochloride, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: β-Alanine ethyleester hydrochloride CAS: 4244-84-2 CAS Min %: 97.5 CAS Max %: 100.0 Assay: 97.5 to 102.5% (Separat. Techn, GC ≥98.5%) Assay Percent Range: 98% Molecular Formula: C5H11NO2·HCl Linear Formula: HNCHCHCOCH5·HCl Formula Weight: 153.61 MDL Number: MFCD00012909
L-Alanine, 99%, ACROS Organics™
Chemical Name or Material: L-Alanine Assay: 98.5 to 101.5% (on dry substance) Assay Percent Range: 99% Molecular Formula: C3H7NO2 Linear Formula: CH3CH(NH2)CO2H Formula Weight: 89.09 MDL Number: MFCD00064410 Synonym: Ala
3-(2-Naphthyl)-L-alanine, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 3-(2-Naphthyl)-L-alanine CAS: 58438-03-2 CAS Min %: 97.5 CAS Max %: 100.0 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C13H13NO2 Formula Weight: 215.25 MDL Number: MFCD00066087
3-(3,4-Dihydroxyphenyl)-L-alanine, 99%, ACROS Organics™
Chemical Name or Material: 3-(3, 4-Dihydroxyphenyl)-L-alanine Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C9H11NO4 Linear Formula: (HO)2C6H3CH2CH(NH2)CO2H Formula Weight: 197.19 MDL Number: MFCD00002598 Synonym: L-DOPA
DL-Alanine methyl ester hydrochloride, 98+%, ACROS Organics™
Chemical Name or Material: DL-Alanine methyl ester hydrochloride Assay: ≥98.0% Assay Percent Range: 98+% Molecular Formula: C4H9NO2·HCl Linear Formula: CH3CH(NH2)COOCH3·HCl Formula Weight: 139.58 MDL Number: MFCD00035523
N-BOC-4-chloroaniline, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N-BOC-4-Chloroaniline CAS: 18437-66-6 CAS Min %: 97.5 CAS Max %: 100.0 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C11H14ClNO2 Linear Formula: ClC6H4NHCOOC(CH3)3 Formula Weight: 227.69 MDL Number: MFCD00816772
L-Alanine, 99%, specified according to the requirements of Ph.Eur., ACROS Organics™
Chemical Name or Material: L-Alanine Name Note: specified according to the requirements of Ph.Eur. CAS: 56-41-7 Assay Percent Range: 99% Molecular Formula: C3H7NO2 Formula Weight: 89.09 MDL Number: MFCD00064410 Synonym: Ala
L-Alanine tert-butyl ester hydrochloride, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 13404-22-3 Physical Form: Solid or Crystalline Powder Assay Percent Notes: 99% Molecular Formula: C7H15NO2·HCl Formula Weight: 181.66 MDL Number: MFCD00035524
L-Alanine, 99%
Chemical Name or Material: L-Alanine CAS: 56-41-7 Molecular Formula: C3H7NO2 Formula Weight: 89.09 MDL Number: MFCD00064410 Beilstein: 1720248 Merck Index: 14,204
BOC-L-Alanine, 99+%, ACROS Organics™
Chemical Name or Material: BOC-L-Alanine Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C8H15NO4 Linear Formula: (CH3)3COC(O)NHC(CH3)COOH Formula Weight: 189.21 MDL Number: MFCD00037225 Synonym: N-(tert-Butoxycarbonyl)-L-alanine
DL-Alanine Products for Science Education
Chemical Name or Material: DL-Alanine CAS: 302-72-7 Linear Formula: CH3CH(NH2)CO2H Formula Weight: 89.09 Synonym: D-2-Aminopropionic Acid
L-Alanine Products for Science Education
Chemical Name or Material: L-Alanine CAS: 56-41-7 Assay Percent Range: >98.5% w/v Linear Formula: CH3CH(NH2)CO2H Formula Weight: 89.09 Synonym: (S)-Alanine
NKH 477, Tocris
Chemical Name or Material: NKH 477 Assay Percent Range: >98% Molecular Formula: C27H43NO8·HCl Formula Weight: 546.1 Synonym: N,N-Dimethyl-(3R,4aR,5S,6aS,10S,10aR,10bS)-5-(acetyloxy)-3-ethenyldodecahydro-10,10b-dihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-1H-naphtho[2,1-b]pyran-6-yl ester β-alanine hydrochloride
L-(-)-α-Methyldopa, Tocris
Chemical Name or Material: L-(-)-α-Methyldopa CAS: 555-30-6 Assay Percent Range: >98% Molecular Formula: C10H13NO4 Formula Weight: 211.22 Synonym: 3-(3, 4-Dihydroxyphenyl)-α-methyl-L-alanine
GSK J4, Tocris™
Chemical Name or Material: GSK J4 Assay Percent Range: >99% Molecular Formula: C24H27N5O2 Formula Weight: 417.5 Synonym: N-[2-(2-Pyridinyl)-6-(1,2,4,5-tetrahydro-3H-3-benzazepin-3-yl)-4-pyrimidinyl]-β-alanine ethyl ester
GSK J1, Tocris™
Chemical Name or Material: GSK J1 Assay Percent Range: >99% Molecular Formula: C22H23N5O2 Formula Weight: 389.45 Synonym: N-[2-(2-Pyridinyl)-6-(1,2,4,5-tetrahydro-3H-3-benzazepin-3-yl)-4-pyrimidinyl]-β-alanine
L-Alanine, Tocris
Chemical Name or Material: L-Alanine CAS: 56-41-7 Assay Percent Range: >99% Molecular Formula: C3H7NO2 Formula Weight: 89.09
L-Alanine (White Crystals or Crystalline Powder), Fisher BioReagents
Chemical Name or Material: L-Alanine CAS: 56-41-7 Physical Form: Solid Assay: 98.5 to 101.0% Assay Percent Range: 98.5 to 101.0 % Molecular Formula: C3H7NO2 Formula Weight: 89.09 Synonym: L-α-Amino Propionic Acid
β-Alanine, Tocris
Chemical Name or Material: β-Alanine CAS: 107-95-9 Assay Percent Range: >98% Molecular Formula: C3H7NO2 Formula Weight: 89.09
L-BMAA hydrochloride, Tocris
Chemical Name or Material: L-BMAA hydrochloride CAS: 16012-55-8 Assay Percent Range: >98% Molecular Formula: C4H10N2O2·HCl Formula Weight: 154.6 Synonym: 3-(Methylamino)-L-alanine hydrochloride
Poly-DL-alanine, 95%, MP Biomedicals™
Chemical Name or Material: Poly-DL-alanine CAS: 25281-63-4 Physical Form: Powder Assay Percent Range: 95% Molecular Formula: C3H7NO2 Formula Weight: 1000 to 5000 MDL Number: MFCD00166369
L-Alanine, 99%, MP Biomedicals™
Chemical Name or Material: L-Alanine CAS: 56-41-7 Physical Form: Powder Assay Percent Range: 0.99 Molecular Formula: C3H7NO2 Formula Weight: 89.1 MDL Number: MFCD00064410 Synonym: 2-Aminopropionic acid
Nalpha-BOC-beta-Amino-L-alanine, 97%, ACROS Organics™
Chemical Name or Material: Nalpha-BOC-beta-Amino-L-alanine CAS: 73259-81-1 Assay Percent Range: 97% Molecular Formula: C8H6N2O4 Formula Weight: 204.23 MDL Number: MFCD00236843 Synonym: BOC-Dap-OH
β-Alanine, 99+%, ACROS Organics™
Chemical Name or Material: β-Alanine Name Note: 99+% CAS: 107-95-9 CAS Min %: 99.0 CAS Max %: 100.0 Assay: 99.0 to 101.0% (on dry substance) Assay Percent Range: 99+% Molecular Formula: C3H7NO2 Linear Formula: H2NCH2CH2CO2H Formula Weight: 89.09 MDL Number: MFCD00008200 Synonym: 3-Aminopropanoic acid
β-Alanine, 98.05%, MP Biomedicals™
Chemical Name or Material: β-Alanine CAS: 107-95-9 Physical Form: Crystalline Powder Molecular Formula: C3H7NO2 Formula Weight: 89.09 Synonym: 3-Aminopropanoic acid
  spinner