Amino Alcohols
D-Cystine 98%, ACROS Organics™
Chemical Name or Material: D-Cystine Name Note: 98% CAS: 349-46-2 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C6H12N2O4S2 Linear Formula: [-SCH2CH(NH2)CO2H]2 Formula Weight: 240.29 MDL Number: MFCD00002610 Synonym: D(+)-3, 3';-Dithiobis(2-aminopropanoic acid)
L-Aspartyl-L-phenylalanine, 95%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-Aspartyl-L-phenylalanine Name Note: 95% CAS: 13433-09-5 Assay: ≥94% (HPLC) Assay Percent Range: 95% Molecular Formula: C13H16N2O5 Linear Formula: HO2CCH2CH(NH2)CONHCH(CH2C6H5)CO2H Formula Weight: 280.28 MDL Number: MFCD00063155
N-BOC-D-Valinol, 98%, ACROS Organics™
Chemical Name or Material: N-BOC-D-Valinol CAS: 106391-87-1 CAS Min %: 97.5 CAS Max %: 100.0 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C10H21NO3 Formula Weight: 203.28 MDL Number: MFCD00235960
L-Methioninol, 99+%, ACROS Organics™
Chemical Name or Material: L-Methioninol CAS: 2899-37-8 Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C5H13NOS Formula Weight: 135.22 MDL Number: MFCD00004735 Synonym: (S)-2-Amino-4-(methylthio)-1-butanol
(S)-(+)-2-Phenylglycinol, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: (S)-(+)-2-Phenylglycinol CAS: 20989-17-7 CAS Min %: 97.5 CAS Max %: 100.0 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C8H11NO Linear Formula: C6H5CH(NH2)CH2OH Formula Weight: 137.18 MDL Number: MFCD00064404
(S)-tert-Leucinol, 95%, ACROS Organics™
Chemical Name or Material: (S)-tert-Leucinol CAS: 112245-13-3 CAS Min %: 94.0 CAS Max %: 100.0 Assay: ≥94% Assay Percent Range: 95% Molecular Formula: C6H15NO Linear Formula: (CH3)3CCH(NH2)CH2OH Formula Weight: 117.19 MDL Number: MFCD00192250
L-Cystine, 99%, ACROS Organics™
Chemical Name or Material: L-Cystine Assay: ≥98.5% (Acid) Assay Percent Range: 99% Molecular Formula: C6H12N2O4S2 Linear Formula: [-SCH2CH(NH2)CO2H]2 Formula Weight: 240.3 MDL Number: MFCD00064228 Synonym: L(-)-3, 3';-Dithiobis(2-aminopropanoic acid)
L-Cystine, 99%
Chemical Name or Material: L-Cystine CAS: 56-89-3 Molecular Formula: C6H12N2O4S2 Formula Weight: 240.3 MDL Number: MFCD00064228 Beilstein: 1728094 Merck Index: 14,2782
L(-)-Histidinol dihydrochloride, 99%, ACROS Organics™
Chemical Name or Material: L(-)-Histidinol dihydrochloride CAS: 1596-64-1 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C6H11N3O·2HCl Formula Weight: 214.1 MDL Number: MFCD00078058 Synonym: L-β-Amino-1H-imidazole-4-propanol dihydrochloride
L(+)-Leucinol, 98%, ACROS Organics™
Chemical Name or Material: L(+)-Leucinol Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C6H15NO Linear Formula: (CH3)2CHCH2CH(NH2)CH2OH Formula Weight: 117.19 MDL Number: MFCD00063676
DL-Ethionine, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: DL-Ethionine Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C6H13NO2S Linear Formula: C2H5SCH2CH2CH(NH2)CO2H Formula Weight: 163.23 MDL Number: MFCD00063102 Synonym: DL-2-Amino-4-(ethylthio)butyric acid
L-β-Homoproline hydrochloride, 98% , ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-β-Homoproline hydrochloride CAS: 53912-85-9 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C6H11NO2·ClH Formula Weight: 165.62 MDL Number: MFCD07363484 Synonym: (S)-2-(2-Pyrrolidinyl)acetic acid hydrochloride
N-BOC-L-Valinol, 96%, ACROS Organics™
Chemical Name or Material: N-BOC-L-Valinol CAS: 79069-14-0 CAS Min %: 95.0 CAS Max %: 100.0 Assay Percent Range: 96% Molecular Formula: C10H21NO3 Formula Weight: 203.28 MDL Number: MFCD00082635
L(+)-Prolinol, 95%, ACROS Organics™
Chemical Name or Material: L(+)-Prolinol CAS: 23356-96-9 Assay: ≥94% (GC) Assay Percent Range: 95% Molecular Formula: C5H11NO Formula Weight: 101.15 MDL Number: MFCD00005255 Synonym: (S)-(+)-2-Hydroxymethylpyrrolidine
D(-)Prolinol, 99%, ACROS Organics™
Chemical Name or Material: D(-)Prolinol Name Note: 99% CAS: 68832-13-3 Assay: ≥98.5% (GC) Assay Percent Range: 99% Molecular Formula: C5H11NO Formula Weight: 101.15 MDL Number: MFCD00064321 Synonym: (R)-(-)-2-Hydroxymethylpyrrolidine
N-BOC-L-Leucinol, 97%, ACROS Organics™
Chemical Name or Material: N-BOC-L-Leucinol CAS: 82010-31-9 CAS Min %: 96.0 CAS Max %: 100.0 Assay: ≥96.0% Assay Percent Range: 97% Molecular Formula: C11H23NO3 Formula Weight: 217.31 MDL Number: MFCD00076950
L-Homoserine, 97%, ACROS Organics™
Chemical Name or Material: L-Homoserine CAS: 672-15-1 Assay: ≥96.0% Assay Percent Range: 97% Molecular Formula: C4H9NO3 Formula Weight: 119.12 MDL Number: MFCD00063090 Synonym: L-2-Amino-4-hydroxybutyric acid
DL-Homoserine, 99%, ACROS Organics™
Chemical Name or Material: DL-Homoserine Name Note: 99% CAS: 1927-25-9 Assay: 98.5 to 101% Assay Percent Range: 99% Molecular Formula: C4H9NO3 Linear Formula: HOCH2CH2CH(NH2)CO2H Formula Weight: 119.12 MDL Number: MFCD00002618 Synonym: DL-2-Amino-4-hydroxybutyric acid
L-Cystine (White Crystals or Cryst. Powder), Fisher BioReagents
Chemical Name or Material: L-Cystine CAS: 56-89-3 Physical Form: Powder/Solid Assay: 98.5 to 101.0% Assay Percent Range: 98.5 to 101.0 % Molecular Formula: C6H12N2O4S2 Formula Weight: 240.29
L-Citrulline, 99.3%, MP Biomedicals™
Chemical Name or Material: L-Citrulline CAS: 372-75-8 Physical Form: Crystalline Powder Assay Percent Range: 99.3% Molecular Formula: C6H13N3O3 Formula Weight: 175.19 Synonym: L -2-Amino-5-ureidovaleric acid
L-Cystine, 99%, MP Biomedicals™
Chemical Name or Material: L-Cystine Name Note: Metal binding, protein structure, antioxidant CAS: 56-89-3 Purity Grade: Cell Culture Reagent Grade Physical Form: Crystals Assay Percent Range: 99% Molecular Formula: C6H12N2O4S2 Formula Weight: 240.3 MDL Number: MFCD00064228 Synonym: Cystein disulfide
H-Ser-Gly-OH, MP Biomedicals™
CAS: 687-63-8 Physical Form: Powder Assay Percent Range: >99% Molecular Formula: C5H10N2O4 Formula Weight: 162.14 MDL Number: MFCD00037778 Synonym: SER-GLY
DL-Homocystine, MP Biomedicals
Chemical Name or Material: Dl-Homocystine CAS: 870-93-9 Formula Weight: 268.36
L-Mimosine, 98.2%, MP Biomedicals™
Chemical Name or Material: L-Mimosine CAS: 500-44-7 Physical Form: Powder Assay Percent Range: ≥98% Molecular Formula: C8H10N2O4 Formula Weight: 198.2 Synonym: [N-(3-Hydroxy-4-pyridone)-aminopropionic acid
  spinner