Ammonium Hydroxide
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 28-30 wt% solution of NH3 in water CAS: 1336-21-6 CAS: 7732-18-5 Purity Grade: For analysis Molecular Formula: H5NO Linear Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00066650
Ammonium hydroxide, ACS reagent, 28-30% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ammonium hydroxide CAS: 1336-21-6 CAS: 7732-18-5 Purity Grade: ACS Reagent Grade Assay: 28.0 to 30.0% (HCl-titration (as NH3)) Molecular Formula: H5NO Linear Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00066650
Ammonia, 2M solution in methanol, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ammonia CAS: 7664-41-7 CAS: 67-56-1 Molecular Formula: H3N Linear Formula: NH3 Formula Weight: 17.03 MDL Number: MFCD00011418
Ammonium hydroxide, extra pure, 25% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ammonium hydroxide CAS: 1336-21-6 CAS: 7732-18-5 Purity Grade: Extra Pure Assay: ≥25% (as NH3) (total base) Molecular Formula: H5NO Linear Formula: NH4OH Formula Weight: 35.05
Ammonium Hydroxide (Optima™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ammonium Hydroxide CAS: 1336-21-6 Purity Grade: OPTIMA™ Linear Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00066650 Synonym: Aqueous Ammonia
Ammonium Hydroxide (Certified ACS Plus), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ammonium Hydroxide Name Note: about 14.8N CAS: 7664-41-7 CAS: 7732-18-5 Purity Grade: Certified ACS Plus Grade Assay: 28.0 to 30.0% (w/w) Assay Percent Range: Assay: 28.0 to 30.0 w/w % Molecular Formula: H5NO Linear Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00011418 Synonym: Aqueous Ammonia
Ammonium hydroxide, for analysis, 25% solution in water, ACROS Organics™
Chemical Name or Material: Ammonium hydroxide CAS: 1336-21-6 CAS: 7732-18-5 Purity Grade: For analysis Molecular Formula: H5NO Linear Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00066650
Ammonium hydroxide, for analysis, 20% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ammonium hydroxide CAS: 1336-21-6 CAS: 7732-18-5 Purity Grade: For analysis Assay: ≥20% (Acidimetry) Molecular Formula: H5NO Linear Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00066508
Ammonium hydroxide, 25% NH3, 99.99% (metals basis), Alfa Aesar™
Chemical Name or Material: Ammonium hydroxide CAS: 1336-21-6 Physical Form: Liquid Assay Percent Range: 99.99% (metals basis) Molecular Formula: H5NO Formula Weight: 35.05 MDL Number: MFCD00066650
Ammonium Hydroxide (TraceMetal™ Grade), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ammonium Hydroxide CAS: 1336-21-6 Purity Grade: TraceMetal Linear Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00066650 Synonym: Aqueous Ammonia
Ammonium Hydroxide, ACS, 28.0-30.0%NH™, Alfa Aesar
Chemical Name or Material: Ammonium hydroxide CAS: 1336-21-6 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: 28.0-30.0% NH3 Molecular Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00066650
Ammonium Hydroxide Solution, 14.8M Products for Science Education
Chemical Name or Material: Ammonium Hydroxide Solution CAS: 1336-21-6 Assay Percent Range: < 30% w/v Linear Formula: NH4OH Formula Weight: 35.04 Synonym: Ammonium Hydrate
Ammonia, ca. 7N solution in methanol, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ammonia Name Note: ca. 7N Solution in Methanol CAS: 7664-41-7 CAS: 67-56-1 Molecular Formula: H3N Linear Formula: NH3 Formula Weight: 17.03 MDL Number: MFCD00011418
Ammonium Hydroxide, 1M Products for Science Education
Chemical Name or Material: Ammonium Hydroxide Solution, 1M CAS: 1336-21-6 Physical Form: Liquid Assay Percent Range: < 30% w/v Linear Formula: NH4OH Formula Weight: 35.04 Synonym: Ammonium Hydrate
Ammonia Solution Products for Science Education
Chemical Name or Material: Ammonia Solution CAS: 7664-41-7 CAS: 7732-18-5 Physical Form: Liquid Assay Percent Range: < 5% Formula Weight: 17.03 Synonym: Ammonia hydroxide water solution
Ammonium Hydroxide (26° Bé/NF/FCC), Fisher Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 7664-41-7 CAS: 7732-18-5 Purity Grade: FCC/NF Assay: 27.0 to 31.0% (NF) Molecular Formula: H5NO Linear Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00066650 Synonym: Aqueous Ammonia
Ammonium Hydroxide, 20% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 1336-21-6 (Ammonium Hydroxide) CAS: 7732-18-5 (Water, Deionized) Physical Form: Liquid
Ammonium Hydroxide 20%, Riccal Chemical
Chemical Name or Material: Ammonium Hydroxide 20%, Ricca Chemical CAS: 1336-21-6 (Ammonium Hydroxide) CAS: 7732-18-5 (Water, Deionized)
Ammonium hydroxide, 30%, J.T.Baker™
Chemical Name or Material: Ammonium hydroxide CAS: 1336-21-6 Physical Form: Liquid Molecular Formula: NH4OH Formula Weight: 35.04 Synonym: Ammonia water
Ammonium Hydroxide, 2.00 Normal, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 7732-18-5 CAS Min %: 87.0 CAS Max %: 91.0 CAS: 1336-21-6 CAS Min %: 10.83 CAS Max %: 11.27 Physical Form: Liquid
Ammonium Hydroxide, 0.500 Normal, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 1336-21-6 (Ammonium Hydroxide) CAS: 7732-18-5 (Water, Deionized) Physical Form: Liquid
Ammonium Hydroxide, 5.00 Normal, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 7732-18-5 CAS Min %: 75.0 CAS Max %: 78.0 CAS: 1336-21-6 CAS Min %: 23.22 CAS Max %: 24.17 Physical Form: Liquid
Ammonium Hydroxide, 6.00N, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide, 6.00N, Ricca Chemical CAS: 7732-18-5 CAS Min %: 71.0 CAS Max %: 74.0 CAS: 1336-21-6 CAS Min %: 26.6 CAS Max %: 27.69
Ammonium Hydroxide, 25% (v/v) Aqueous Solution (1+3), Ricca Chemical
CAS: 7732-18-5 Physical Form: Liquid Assay Percent Range: 25% (v/v) Aqueous Solution (1 + 3)
Ammonium Hydroxide, 16.7% (v/v) Aqueous Solution, Electronic Grade, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 7732-18-5 CAS Min %: 83.0 CAS Max %: 84.0 CAS: 1336-21-6 CAS Min %: 16.0 CAS Max %: 17.0 Purity Grade: Electronic Grade Physical Form: Liquid
Ammonium Hydroxide, 50% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 7732-18-5 CAS Min %: 49.0 CAS Max %: 51.0 CAS: 1336-21-6 CAS Min %: 49.0 CAS Max %: 51.0 Physical Form: Liquid
Ammonium Hydroxide Solution, 10% (w/v) NH3, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide Solution CAS: 7732-18-5 CAS Min %: 72.0 CAS Max %: 75.0 CAS: 1336-21-6 CAS Min %: 9.0 CAS Max %: 11.0 Physical Form: Liquid
Ammonium Hydroxide, 6.00 Normal, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 7732-18-5 CAS Min %: 71.0 CAS Max %: 74.0 CAS: 1336-21-6 CAS Min %: 26.6 CAS Max %: 27.69 Physical Form: Liquid
Ammonium Hydroxide, ACS Reagent Grade, 28.0-30.0% as NH3, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 1336-21-6 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7732-18-5 (Water, Deionized) Purity Grade: ACS Reagent Grade Physical Form: Liquid
  spinner